Fast 5
From time to time Thermo Fisher Scientific releases firmware upgrades for our gas analyzers. For the iQ series analyzers, this firmware upgrade can be accomplished in a couple of different ways.
Option one
Download the firmware upgrade file from our website to a USB drive then insert it into the USB port on the front of your iQ analyzer. Firmware upgrade files can be found on our Enviromental and Process Monitoring Services page, for your specific model of instrument, under Firmware / Software.
Using the USB method is the most simple and intuitive. With the firmware downloaded to the USB drive, insert it into the USB port on your analyzer and follow the self-guided instructions. Do not remove it until the analyzer reboots as part of the upgrade process.
Note: Do not change the name of the file in any way as the iQ series unit will not recognize it if changed.
Option two
Use Thermo Scientific ePort software to communicate with your instrument and download the latest firmware upgrade file.
Available: ePort Software
The latest version of our ePort software (v4.0.5) for remote communication to your instrument can be found on our Enviromental and Process Monitoring Services page, for your specific model of instrument, under Firmware / Software.
If your instrument has Version 11 firmware installed (V1.6.11.xxxxx), ePort software must be used to upgrade the firmware. To check the version of firmware installed of your iQ series instrument, go to Settings (accessible via the Home Screen) → Health Check. The installed firmware version will be displayed.
For further information, please review the Technical Bulletin, iQ Instrument V1.6.11 Firmware Update Using ePort Software.
ePort Software Security Improvements
New security improvements were released for firmware versions V8 (V1.6.8.xxxxx) and higher. To connect with the ePort software remotely, a security password must now be entered. Please review the Technical Bulletin, iQ Series Instrument Security Improvements, describing the password procedure.
Need Service or Support?
Customer Service/Orders
1 (866) 282-0430, Option 1
customerservice.aqi@thermofisher.com 
orders.aqi@thermofisher.com
Field Service
1 (866) 282-0430, Option 3 
epm.emfieldservice@thermofisher.com
Technical Support
1 (866) 282-0430, Option 2 
epm.techsupport@thermofisher.com
Factory Depot Service
1 (866) 282-0430, Option 3 
orders.aqi@thermofisher.com
To schedule your analyzer, calibrator, probe controller or probe for a return to factory depot repair for complete maintenance or factory specification service, please contact us today ›
Was this newsletter forwarded to you?
Sign up to get your own Fast5 newsletter delivered to your inbox. Sign up ›
Interested in past Fast5 newsletters? Visit the Fast5 Archive ›
Have an idea for a Fast5 newsletter topic? Let us know ›
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.