Fast5 bannner
January/February 2018
Volume V, Edition 1
The  iQ Series of gas analyzers, the latest platform of Thermo Scientific gas analyzers, is now available for purchase and quote. Predictive Diagnostics is a standard feature of the iQ Series platform which is designed to anticipate your instrument’s maintenance needs, identify possible component failures, reduce downtime and reduce troubleshooting time. If a predictive diagnostic event is flagged, your instrument can proactively notify you to this unusual or subpar performance condition affording you the opportunity to resolve the condition before your instrument enters an alarm state. The functionality is intended to be scalable by refining existing diagnostic parameters based upon historical performance or building out additional parameters. Customers will be able to upgrade their instruments to take advantage of new Predictive Diagnostic parameters in the future with a routine firmware upgrade. 
Let’s take a 5 minute look at how Predictive Diagnostics work:
Thermo Scientific™ 48iQ Carbon Monoxide Analyzer
S/R – By design, the S/R ratio will slowly decrease as the filter wheel leaks carbon monoxide (CO). Typically, this can take 3-5 years before the filter wheel would need to be replaced.
With Predictive Diagnostics, the instrument will look at the S/R ratio every time a zero calibration is performed & take note of the value.  If it is predicted that the S/R will be <1.14 within 60 days, an alert will be triggered to inform you to change the filter wheel before you experience unplanned downtime.
A modular subsystem
Have an idea for a future Fast5 issue? Let us know ›
Support
Factory repair
Schedule your analyzer, calibrator, probe controller or probe for a Return Material Authorization (RMA) to factory depot repair for complete maintenance and factory specification service.
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.