Search Thermo Fisher Scientific
Nicolet Summit FTIR Spectrometers
Generate fast, reliable answers with less effort—even in busy, multi-user QC labs or teaching labs with our compact, powerful FTIR spectrometers for the name you trust in FTIR–Nicolet. Today’s busy QA/QC and teaching laboratories demand fast answers with high confidence. When you can’t afford to slow down or make a mistake, rely on our compact Thermo Scientific Nicolet Summit FTIR Spectrometers to help you identify, verify, or quantify materials faster than ever before.
The Nicolet Summit is available in four different models—LITE, X, PRO, and OA—offering you a range of options for the solution that fits your needs.
|
||||
LightDrive Optical Engine (laser, source, interferometer) |
|
|
|
|
Warranty on LightDrive laser, source and interferometer |
5-year |
10-year |
10-year |
10-year |
OMNIC Paradigm Software |
|
|
|
|
Pharmacopeia workflows |
|
|
|
|
Detector |
LiTaO3 |
DTGS |
TEC-DTGS |
DTGS |
Included library/reference spectra |
850 |
5,000 |
10,000 |
|
User Interface |
USB External PC |
USB External PC |
Integrated Windows 10 Computer |
Integrated Windows 10 Computer |
Wi-Fi enabled |
|
|
|
|
Touchscreen available |
|
|
|
|
Internal motorized aperture | ![]() |
The Thermo Scientific Nicolet Summit X FTIR Spectrometer can help you reach new heights in productivity, so you can accomplish more in less time. It is ideal for both single-user labs and multi-user labs. Simple, high-functionality software allows easier operation for new and experienced users alike. For the ultimate in reliability and accuracy, the Nicolet Summit X FTIR Spectrometer is designed with our unique LightDrive Optical Engine. The user-friendly interface of the Thermo Scientific OMNIC Paradigm Software makes life in the lab less complicated with programmed applications that streamline data acquisition, processing, and interpretation.
Reduced workflow set-up time. An intuitive, visual workflow creator with a drag-and-drop functionality guides you through the workflow creation.
Audit-ready cGxP compliance. Achieve a high level of data integrity with our database infrastructure, data security software, and audit manager applications that keep you compliant with Pharmacopeia (Ph. Eur., USP, JP, and CP) instrument qualification. 21 CFR Part 11 compliance and data security packages are available.
Data analysis anytime, anywhere. With full Wi-Fi connectivity on your PC, you can send data to your Thermo Fisher Connect account in the Cloud and analyze data away from the lab. Share data and collaborate with colleagues anywhere in the world to accelerate your research. Keep students engaged with data they can access in your teaching lab or in their dorm room.
OMNIC Paradigm Desktop Software. Find what you need quickly from the Dashboard screen, create libraries, or perform multi-component searching.
With no time to waste, you can move quickly from one sample to the next and have your colleagues repeat your success—that’s the power of the Nicolet Summit FTIR Spectrometer combined with the OMNIC Paradigm software.
Nicolet Summit X FTIR Spectrometer product walkthrough
Explore OMNIC Paradigm software's touchscreen interface, including the home screen, spectral view, and workflows.
Explore OMNIC Paradigm software's desktop interface, including the dashboard, spectral view, and workflow editor.
The multi-colored LED light bar on the Nicolet Summit FTIR Spectrometer provides pass/fail indicators of product quality with just a quick glance. Monitor the status of your instrument (idle, collect, alert).
Need more proof?
What would it be like if you could cut your analysis time in half? Smart Background is a proprietary technology that completely removes the background collection step from your workflow.
Intuitive software that drives projects to completion
The Nicolet Summit FTIR Spectrometer is powered by our revolutionary OMNIC Paradigm Software to help you keep pace with all the daily demands on your lab. The user-friendly dashboard in OMNIC Paradigm Software helps you view instrument status and recent work, process your spectra, conduct multi-component searching, and create new libraries. Designed with lab managers and science educators in mind, this software helps automate workflows using an intuitive drag-and-drop workflow creator.
Achieve the highest degree of data integrity and surpass your auditors’ expectations. Our database infrastructure, streamlined data security software, and Audit Manager applications keep your data compliant and well-protected. Manage multiple user permissions, digitally sign important files, and track detailed user activity using the OMNIC Paradigm Security Suite Software.
Achieve a high level of data integrity with our database infrastructure, data security software, and audit manager applications that keep you complaint with Pharmacopeia (Ph. Eur., USP, JP, and CP) instrument qualification. 21 CFR Part 11 compliance and data security packages are available.
Verify raw materials and specifications of solid dosage drugs and identify contaminants to help protect your company’s reputation. The Nicolet Summit FTIR Spectrometer is portable, so you can bring it to the production line for at-line process analysis or station an instrument at your incoming raw materials location.
Understand what chemical properties you need to control when designing new materials with advanced features. Catch changes in additive formulations, degradation of multi-layer polymer composites, weakened packaging materials, and the effects of process curing times on polymer specifications.
Keep students engaged with this intuitive spectrometer and lesson plans that help you demonstrate different functional groups of the IR spectra. Have students quickly measure samples in a crowded teaching lab and then analyze data later, on or off campus, using the OMNIC Anywhere. Learn more with our Introduction to Lab Spectrometers for Your Classroom.
Confidently identify different grades of jade or uncover the composition of a mineral sample using the Nicolet FTIR Summit Spectrometer. Find out how the Gemological Institute of America (GIA) uses FTIR spectroscopy for identifying treated versus untreated gemstones.
After optimizing your lithium battery alternatives (e.g., Li–S, Li–O2, Na-ion) and electrolyte formulations, inspect product quality and collect chemical information that assures specifications have been met. The non-destructive nature of FTIR spectroscopy makes this an ideal technique for quickly catching changes that can affect battery life and safety.
From leather interiors to dashboard components to fuel additives, use the Nicolet Summit FTIR Spectrometer to help you improve profitability, performance, and appeal of your automotive design. Reveal the chemistry and composition of materials used to manufacture today’s fuel-efficient cars and trucks for a cleaner, safer world.
Save time, boost proficiency, and speed results by bundling with equipment purchase. When time is on the line, busy QA/QC and academic teaching laboratories require accuracy and speed—no exceptions. That’s why bundling the Nicolet Summit FTIR Spectrometer along with Unity Lab Services Installation/Education Service augments abilities and ignites results.
Standard |
Premium |
Validation |
Installation/Orientation |
Installation/Education |
Optional |
|
|
Required for regulated environments, our Validation Option is available for both Standard and Premium services and includes the necessary Professional Qualification Service with all Certificates and documentation. |
Reserve your installation/education services when you buy your equipment—it's as simple as adding a line to your purchase order. You can also add it separately up until your unit ships.
Access Nicolet Summit FTIR Spectrometer resources at any time and keep your lab operating at peak. Whether you are a novice to spectroscopy or an experienced professional, helpful technical information for your Nicolet Summit FTIR Spectrometer is just a click away.
Nicolet Summit support center has:
For Research Use Only. Not for use in diagnostic procedures.