Pierce™ Gentle Ag/Ab Binding and Elution Buffer Kit
Pierce™ Gentle Ag/Ab Binding and Elution Buffer Kit
Thermo Scientific™

Pierce™ Gentle Ag/Ab Binding and Elution Buffer Kit

Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such asRead more
Have Questions?
Catalog NumberQuantity
21030200 mL
Catalog number 21030
Price (USD)
178.00
Each
-
Add to cart
Quantity:
200 mL
Request bulk or custom format
Price (USD)
178.00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads. The Gentle Ag/Ab Binding and Elution Buffer system uses near-neutral, high-salt conditions for elution, enabling even sensitive and labile antibodies (or other proteins) to be eluted from an affinity system without denaturation and inactivation. The phosphate-free binding buffer in this system is a necessary substitute for the Protein A binding buffer (pH 8), which precipitates with the Gentle Ag/Ab Elution Buffer.

Shared features of all Thermo Scientific Pierce IgG Binding and Elution Buffers:

Ready to use—carefully formulated and prefiltered 1X solutions
Three binding buffers—optimized for Protein A, Protein G or Protein A/G
Two elution buffer options—acidic (pH 2.8) or high salt (near-neutral)
Versatile—elution buffers are appropriate for antibody-antigen affinity purification

Item-Specific Features:

Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA
Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA
Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide
IgG Elution Buffer: pH 2.8; amine-based
Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide
Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

Related Products
Pierce™ Gentle Ag/Ab Binding Buffer, pH 8.0
Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Gentle Ag/Ab Binding and Elution Buffer Kit
FormLiquid
Quantity200 mL
Reagent TypeElution Buffer, Binding Buffer
FormatKit
Product LinePierce™
Product TypeBinding and Elution Buffer
pH8 (Binding Buffer), 6.6 (Elution Buffer)
Unit SizeEach
Contents & Storage
Sufficient For: Binding and near-neutral elution of antibodies with Protein A or G
• Pierce Gentle Ag/Ab Binding Buffer, 100 mL
• Pierce Gentle Ag/Ab Elution Buffer, 100 mL

Store at 4°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3212663ACertificate of AnalysisMay 19, 202521030
3212663Certificate of AnalysisMay 19, 202521030
ZL400445Certificate of AnalysisJan 31, 202521030
ZL400445ACertificate of AnalysisJan 31, 202521030
ZE381149Certificate of AnalysisMay 22, 202421030
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

This varies with the stability of the immobilized protein and the type of elution buffer used. Columns can typically be reused at least 10 times without significant loss of activity. If the immobilized molecule is sensitive to low pH, consider using our Pierce Gentle Ag/Ab Elution Buffer (Cat. No. 21013, 21027 or 21030). For alternative elution buffers, please refer to this Tech Tip(http://tools.thermofisher.com/content/sfs/brochures/TR0027-Elution-conditions.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Share catalog number, name or link

1x1 image pixel for data collection