All-in-one solution for time-lapse imaging
The second-generation Invitrogen EVOS Onstage Incubator (OSI-2) can be paired with the EVOS M7000 or EVOS M5000 imaging systems for long-term monitoring of cell cultures and time-lapse imaging at high resolution. The environmental chamber enables precise control over temperature, humidity, and gas (O₂, CO₂ or NO₂) levels for incubating cells under physiological and nonphysiological conditions over long periods of time, making it excellent for demanding live-cell imaging experiments including hypoxia and neurite outgrowth studies.
Features
Intuitive UI
Simplifies experiment setup and monitoring of experimental conditions and enables adjustments while experiments are running
Compact
Small footprint and sleek design saves lab space 
Onboard air compressor
Eliminates need for separate air tank; OSI-2 is also quiet at 42 dB 
User-defined controls
Precise control over the incubation temperature, humidity, and gasses facilitates relatively uniform live-cell envirnoment (see image on right)
Chart showing OSI-2 temperature uniformity within a 6-well plate
Temperature uniformity for OSI-2 with a 37°C setpoint.
Featured images and videos
OSI-2 maintains the optimal live-cell environment for your live-cell experiment, including neurite outgrowth, cell proliferation, and hypoxia. Images and time-lapse movies can be further analyzed and additional visualization options are available with Celleste 6.0 Image analysis software.
Video Player is loading.
Current Time 0:00
Duration 1:54
Loaded: 0%
Stream Type LIVE
Remaining Time 1:54
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Overview and general operation
    Learn more about the general operation of the EVOS Onstage Incubator following installation by a qualified field representative.
    Video Player is loading.
    Current Time 0:00
    Duration 3:27
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 3:27
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Unboxing and installation
      Learn how to unbox and install the EVOS Onstage Incubator.
      Video Player is loading.
      Current Time 0:00
      Duration 0:22
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 0:22
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Lysosomal localization and degradation
        Time-lapse imaging over 65 hours of lysosomal localization and degradation with a LysoLight Deep Red conjugated antibody using the EVOS Onstage Incubator and EVOS M7000 microscope. SKBR3 cells were treated with 1 µg/mL of LysoLight Deep Red conjugated to the chemotherapy antibody, trastuzumab and high-resolution images were captured every 15 minutes over 3 days.
        Video Player is loading.
        Current Time 0:00
        Duration 0:10
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 0:10
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          Cell proliferation
          Time-lapse imaging of U2OS cells with an EVOS M7000 Imaging System equipped with an EVOS Onstage Incubator (OSI-2). U2OS treated with CellLight GFP-tubulin (C10613) and imaged every 15 minutes over 48 hours at 37oC, 5% CO2, and 80% humidity with an EVOS M7000 Imaging System (Cat. No. AMF7000) equipped with an OSI-2 (Cat. No. AMC2000) and a 20X objective (Cat. No. AMEP4734).
          Neurite outgrowth
          Microscopic view of cultured rat hippocampus neurons
          Microscopic view of cultured rat hippocampus neurons
          Video Player is loading.
          Current Time 0:00
          Duration 0:25
          Loaded: 0.00%
          Stream Type LIVE
          Remaining Time 0:25
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • default, selected
            Time-lapse imaging of neurite outgrowth from neurons over several days using an EVOS M7000 Imaging System equipped with an EVOS Onstage Incubator (OSI-2). Rat hippocampal neurons plated on Poly-D-Lysine (Cat. No. A38904) coated plates in Neurobasal Plus medium (Cat. No. A3582901) with B-27 Plus supplement (Cat. No. A35828) incubated in an OSI-2 (Cat. No AMC2000) with 5% CO2, 37°C, and 80% humidity and imaged every 15 minutes for 48 hours with an EVOS M7000 Imaging System (Cat. No. AMF7000) equipped with a 20x objective (Cat. No. AMEP4734). Left image: day 1 neurite outgrowth; right image: day 3 neurite outgrowth.
            Hypoxia
            Microscopic view hypoxic HeLa cells indicated by red fluorescence at 20 percent oxygen
            Microscopic view hypoxic HeLa cells indicated by red fluorescence at 1 percent oxygen
            Real-time detection of hypoxia with Image-iT Hypoxia Red using an EVOS M7000 Imaging System equipped with an EVOS Onstage Incubator (OSI-2). A549 cells were labeled with Image-iT Hypoxia Red (Cat. No. H10498) and incubated in the OSI-2 (Cat. No. AMC2000) at 20% oxygen for 1 hour (left image) then oxygen was decreased to 1% oxygen for 1 hour (right image) with 5% CO2, 37oC, and 80% humidity held constant. Imaging performed with an EVOS M7000 Imaging System (Cat. No. AMF7000), equipped with a custom EVOS filter cube with GFP excitation (470/22) and RFP emission (593/40) and a 20x objective (Cat. No. AMEP4734).
            Specifications
            EVOS vessel holders and stage plates
            To help ensure proper operation of the EVOS Onstage Incubator, we recommend choosing vessels designed specifically for live-cell imaging. Conventional plastic plates can generate optical problems or even autofluorescence. Additionally, it is important to use only EVOS Onstage Incubator compatible vessel holders and stage plates.
            Onstage incubator media
            Many cell culture media are carbonate buffered and, once removed from the incubator, the relatively lower CO2 concentration of air will cause an increase in the medium pH within an hour. The optically clear, physiological solution (Live Cell Imaging Solution, or LCIS) maintains physiological pH and osmolarity for up to 4 hours at ambient atmosphere and temperature. This is especially useful for imaging outside of an incubator but can also help to maintain pH while using the OSI-2.
            For longer-term imaging, particularly in a microscope-stage incubator, we offer a clear cell culture medium, Gibco FluoroBrite DMEM, that is based on DMEM to help preserve cell health while also providing low background fluorescence (comparable to that of phosphate-buffered saline and 90% lower than standard phenol red–free DMEM).
            Ordering information
            Catalog # Name Size Price (USD) Qty
            AMC2000 Each -