Element Crystal orth
Neptunium • Actinide Rare Earth
Symbol: Np
Date of discovery: 1940
Name origin: planet Neptune
Appearance: silver
Discoverer: E.M. McMillan
Obtained from: man-made
Melting point: 910 K
Boiling point: 4273 K
Density[kg/m3]: 20450
Molar volume: 11.59 × 10-6 m3/mol
Protons/Electrons: 93
Neutrons: 144
Shell structure: 2,8,18,32,22,9,2
Electron configuration: [Rn]5f46d17s2
Oxidation state: 3,4,5,6
Crystal structure: orthorhombic
Neptunium is a silvery metallic element named after the planet Neptune. The first synthetic transuranium element of the actinide series, neptunium was discovered by E. McMillan and P.H. Abelson in 1940 by bombarding uranium with slow-moving neutrons. While neptunium is naturally found in only minute quantities as decay products of transmutation reactions in uranium ores, this ductile element can be extracted in kilogram quantities from nuclear reactors. 237Np, the most stable of 17 isotopic forms, can be used as a component in neutron detection instruments.
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.