Suntec Singapore Convention & Exhibition Centre | Level 3 – 1 Raffles Blvd, Singapore 039593
Thank you to everyone who joined us for our lunch session and visited our booth at Dioxin 2024 in Singapore. We were excited to be part of this prestigious gathering with leading experts, researchers, and professionals.
Please download the assets for our presentations and posters below and we look forward to seeing you at the next Dioxin conference.
Interested in finding out more about our solutions?  Please let us know and we’ll put you in touch with one of our experts.
Beyond dioxins - Obtaining accurate compound identification in environmental samples with GC Orbitrap MS
In this presentation hear from Dr. Kjell Hope from Pacific Rim Labs on their approach to non-targeted screening and quantitation using GC Orbitrap MS. Environmental samples were extracted with the aim of preserving sample integrity whilst sufficiently cleaning the sample of background matrix. Extract analysis was performed using high-resolution accurate mass GC Orbitrap MS in full scan mode at 60,000 resolution. Analysis in full scan (untargeted) allows for a complete chemical profile to be collected from the sample. This data was then processed to screen for an unlimited number of compounds, thereby increasing scope of analysis beyond what is possible using targeted methods such as GC-MS/MS or GC Magnetic Sector instrumentation. In this study, samples were processed and compared using Thermo Scientific Compound Discoverer Software to identify features of interest. These suspect features were then screened against NIST 2023 and a high-resolution accurate mass environmental contaminants library. The complete workflow, which aims for fast and accurate compound identification, will be discussed.
Speaker:
Kjell Hope, Technical research chemist, Pacific Rim Laboratories
Which system do I need for dioxin analysis: GC Magnetic Sector MS, GC-MS/MS or GC Orbitrap MS?
For Dioxins and POPs analysis, the DFS Magnetic Sector GC-HRMS System has long been recognized as the instrumentation for meeting worldwide regulations and compliance. However, techniques such as GC-MS/MS or GC Orbitrap MS provide alternative instrumental approaches to this challenging application. As laboratories strive for high throughput, increased scope of analysis, and reduced cost of ownership, it is important to consider the merits and limitations of all three approaches for various sample types and performance criteria. Join our application specialists as they discuss, with real-world data examples, the advantages of each technique in meeting regulatory criteria.
Speakers:
Heinz Mehlmann, Application Manager DFS Magnetic Sector GC-HRMS, Thermo Fisher Scientific
Nicholas Warner, Product Applications Specialist GC-MS, Thermo Fisher Scientific
About the speakers
Kjell Hope
Kjell Hope has been working in chemical analysis for 8 years. He obtained a master’s degree in chemistry from Orebrö University in Sweden analysing PFAS in water collected off the east coast of Africa. He has contributed to many projects in private industry and academia, including the UNEP global monitoring of persistent organic pollutants, project Campania Trasparente, screening PCBs in various foodstuffs, and ongoing MS/MS method development at Pacific Rim Labs.
Heinz Mehlmann
Dr. Heinz Mehlmann is a Senior Product Specialist for the GC-HRMS. His background is in Electronics, and he has a Ph.D. in Organic Chemistry. He started at Thermo Fisher Scientific as a Global Support Specialist and was involved in the development of the DFS magnetic Sector GC-HRMS. He has 20 years of experience in high resolution mass spectroscopy particularly in the field of Dioxin and POPs analysis.
Nicolas Warner
Nicholas is a product specialist with a strong scientific background in GC and GC-MS applications. Originally from Canada, he has worked as an environmental researcher for 14 years within Norway before moving to join the Thermo Fisher Scientific team in Bremen, Germany in 2021. With a background in Environmental/Analytical chemistry, he is a specialist in the analysis of emerging harmful substances in the environment using gas chromatography mass spectrometry and high-resolution mass spectrometry.
DFS Magnetic Sector GC-HRMS
DFS Magnetic Sector GC-HRMS
Reasons and technical background why the Thermo Scientific DFS Magnetic Sector GC-HRMS is the gold standard for performing Dioxin and Persistent Organic Pollutants (POPs) analysis.
Learn more
NeverVent Technology
NeverVent Technology
NeverVent technology maximizes GC-MS instrument productivity and eliminates unwanted instrument downtime. Equipped across the entire portfolio of GC-MS instrumentation, routine maintenance is easily achieved without venting the mass spectrometer. Column exchange and ion source removal can be performed while maintaining instrument vacuum. The combination of our Advanced Electron Ionization (AEI) source and NeverVent technology will be demonstrated in recent applications including POPs and pesticide analysis showing sensitivity with robust instrument uptime.
Learn more
Orbitrap Exploris GC
Orbitrap Exploris GC
Since its launch in 2015, Orbitrap Exploris GC has enabled scientists across all applications to gain a deeper understanding of their samples. With the current Orbitrap Exploris GC series at a mass resolving power of up to 240,000, this provides unprecedented analytical performance for confident identification and quantitation of compounds in complex matrices. In this short introduction hear from our applications specialist who will introduce the key hardware features and the performance it delivers in environmental applications.
Learn more
HeSaver-H2Safer
HeSaver-H2Safer
The Thermo Scientific HeSaver-H2Saver SSL injection module provides laboratories the same robustness and performance of the standard SSL injection, but with the added benefit of drastically reducing carrier gas consumption during analysis. An overview of this gas saving technology and its application towards of Dioxins and pesticides in various environmental matrices will be discussed as well as performance when using alternative carrier gases (i.e., hydrogen).
Learn more
* Required field
*
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
CMD Form Code v3.0
Previous years
Dioxin 2023
Lunchtime seminars
Monday, Sep 11 | 12:15 - 1:15 p.m. CEST | Room 0.2 Berlin | Level-1
Breaking Down POPs & PFAS : Sample to Solution looking at sample preparation and PFAS analysis, specifically untargeted analysis
New advancements have been made which allows for a more efficient and controlled extraction of persistent organic pollutants (POPs), using parallel gas assisted accelerated solvent extraction, in-line cleanup and automated solvent concentration in the same platform. We present here performance data from soil samples for different POPs. Unlike traditional methods such as liquid-solvent extraction followed by N2 stream evaporation, the fully automated solvent extraction, in-line cleanup and evaporation system saves time, solvent, and labor, while ensuring high reproducibility and productivity for analytical testing.
One of the main emerging POPs of concern is PFAS (Per- and Polyfluoroalkyl Substances) and the vast amount and variety of PFAS compounds having different chemical properties creates analysis challenges and opportunities to apply multiple techniques like GC- and LC-MS for the analysis. To screen and/or quantitate the relevant PFAS compounds appropriate sample preparation, chromatography and instrument settings, and experiments are required due to the nature of different matrices. We will present how to analyze large number of compounds utilizing full scan with high resolution and has the advantage to be able to review data retrospectively.
Presentors: Michael Ebitson, Product Marketing Manager Sample Preparation, Thermo Fisher Scientific; Frans Schoutsen, Application Support Sr. Specialist, Thermo Fisher Scientific
Tuesday, Sep 12 | 12:15 - 1:15 p.m. CEST | Room 0.2 Berlin | Level-1
Latest advancements in Dioxin & POPs analysis – focusing on the DFS magnetic sector GC-HRMS, Orbitrap technology and the helium saver
Latest advancements in Dioxin & POPs analysis with our complete Thermo Fisher Scientific GC-MS portfolio.
System advancements on the Thermo Scientific DFS Magnetic Sector GC-HRMS.
Customer insights on Dioxin & POPs analysis using the Thermo Scientific Orbitrap Exploris GC-MS.
Save money with reduced helium usage with unique solutions.
Presentors: Dr. Kerstin Kraetschmer - Wageningen Food Safety Research, Netherlands; Heinz Mehlmann, Sr. Product Specialist, Thermo Fisher Scientific
Posters
Customer solutions
Learn how our customers solve their analytical challenges with Thermo Scientific technology over a wide range of applications involving POP analysis.
Featured solutions
GCMS POPs and Dioxins portfolio
With unrivaled expertise in the world of Dioxins and POPs analysis, allow Thermo Scientific to be your partner and enable you to surpass your analytical challenges. Each solution offers advantages for the analysis POPs in environmental and food samples
DFS Magnetic Sector GC-HRMS system
World-Wide compliance for the lowest regulatory limits for Dioxins
Orbitrap Exploris GC Mass Spectrometer
Rapid instrument set up and broad scope POPs screening
TSQ 9610 GC-MS
Ready to implement methods for POPs and compliant with EU regulations for Dioxin food and feed
GC Consumables for POPs analysis
Dedicated columns for ultra trace POPs determination
LCMS POPs portfolio
Continual changes in POPs research and regulatory requirements drive the need for eve-lower limits of detection and quantitation. The emerging contaminants, PFAS, are extremely persistent in the environment and resistant to typical environmental degradation processes.
Use of LC-MS/MS has enabled more sensitive determinations of individual PFOS and PFOA.
The unique features of Thermo Scientific LC-MS/MS platforms allow sensitive detection and quantitation in matrix to address these more stubbornly resilient POPs compounds
Vanquish Flex UHPLC system
TSQ Plus triple quadrupole mass spectrometers
Orbitrap Exploris 120 mass spectrometer
POPs Extraction portfolio
Coming soon!
Thermo Scientific EXTREVA ASE Accelerated Solvent Extractor
Walk away, sample to vial, all-in-one sample extraction, in-cell cleanup, and evaporation set you free. Reproducible and efficient parallel extractions cut manual sample preparation steps from hours to minutes, increase productivity, minimize errors, and dramatically reduce solvent use to meet laboratory sustainability and cost goals.
Dionex AutoTrace 280 PFAS
Increase throughput and reduced risk of contamination
Software for POPs analysis
Chromeleon 7.3.1 Chromatography Data Systems (CDS)
Simplify meeting latest regulatory requirements
TraceFinder Software
Simplifying high-throughput screening and quantification
Compound Discoverer Software
Turn data into knowledge
39th International Symposium on Halogenated Persistent Organic Pollutants
The most comprehensive portfolio for Dioxin and POPs analysis
Presentations at Dioxin Conference 2019
Lunch Seminar: Routine, regulatory analysis of dioxins and dioxin-like compounds in food and feed samples using a triple quadrupole GC-MS/MS system with an advanced EI source
27 August | 12:15 – 12:30
Recent advances in GC-MS/MS technology have enabled lower detection limits and improved automated selected reaction monitoring (SRM) performance. This, in turn, has led the European Commission to update regulations to permit the use of GC-MS/MS, in addition to magnetic sector technology, as a confirmatory method for enforcing maximum levels (ML) and action levels (AL) for dioxins/furans and dl-PCBs in foodstuffs and animal feeds. Attend the lunch seminar and discover more about the Dioxin Analyzer. Not only does it take advantage of the new GC-MS/MS technical advancements, but significantly improves the ease of use and accelerates the implementation in routine laboratories.
Lunch Seminar: Are you being served? The benefits of DualData Acquisition in a Routine Dioxin Lab
27 August | 12:30 – 12:45
Looking for more performance for Dioxin and POPs analysis? Join this seminar and listen to the experts of NofaLab presenting their real-life experience with the DualData XL module of the DFS Magnetic Sector GC-HRMS
38th International Symposium on Halogenated Persistent Organic Pollutants
Discover the most comprehensive GC-MS portfolio for Dioxin and POPs analysis
Presentations at Dioxin Conference 2018
Dioxin, POPs, and Emerging Contaminants with Thermo Scientific GC-MS and LC-MS solutions
26 August | 12:00 – 13:30
From research to routine, pesticides and POPs analysis require comprehensive GC-MS and/or LC-MS solutions enabling sensitive performance and precise results. Attend the seminar and discover real applications, and the latest advances on GC-MS technologies.
Open User Meeting - DFS Magnetic Sector GC-HRMS
26 August | 14:00 – 15:30
Join the user community of Thermo Scientific DFS Magnetic Sector GC-HRMS and discover applications and examples of how it is used in research and routine analysis for dioxin and POPs determinations.
Lunch Seminar: DFS Magnetic Sector GC-HRMS
28 August | 12:20 – 13:05
Looking for more performance for Dioxin and POPs analysis? Join this seminar and listen to expert presentions on DFS Magnetic Sector GC-HRMS applications.
  •  
Lunch Seminar: New Source Technology and Software for Analyzing POPs in Various Matrices
30 August | 12:20 – 13:05
Challenging applications need sensitivity without compromising on ease-of-use. Register and attend this seminar to learn about new source technology and new software tools for analyzing POPs in various matrices.
Featured solutions
Choose from the Thermo Scientific GC-MS portfolio for your analytical challenges. Each solution offers you specific capabilities and advantages, and collectively they represent the most complete and complimentary GC-MS portfolio available on the market for Dioxins and POPs analysis.
37th International Symposium on Halogenated Persistent Organic Pollutants
Discover the Most Comprehensive GC-MS Portfolio and Complete Analysis Workflows
Learn more about latest developments in the field of Dioxin and POPs Analysis.
Seminars at Dioxin Conference 2017
Dioxin, POPs and Emerging Contaminant Workshop: From Sample Prep to Data Analysis
Sunday, 20 August | 13:00 – 14:30
Go through the steps of GC-MS/MS and LC-MS analytical workflows for emerging contaminants determinations.
Speaker: Dave Hope, CEO Pacific Rim Lb. Inc.
Dioxins & POPs: Discover the Most Comprehensive GC-MS Portfolio
Tuesday, 22 August | 12:00 – 12:45
Experts in the field of Dioxin and POPs analysis will present their research in the field of Dioxin and POPs analysis. Register and come by the Thermo Fisher Scientific booth to collect your entrance card. Lunch will be provided.
CMD Wide-format style fixes
Style Sheet for Komodo Tabs
Style Sheet for Global Design System