Alpha-Gal Syndrome Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
The recently identified alpha-Gal syndrome (AGS)—aka red meat allergy, tick-bite meat allergy, alpha-Gal allergy—is a type of serious, potentially life-threatening allergic reaction to red meat and other products made from mammals.1,2 Most patients that become allergic to alpha-Gal have tolerated red meat for years before being sensitized by a tick bite.3 Thus, the condition typically begins when a tick bites someone and transmits alpha-Gal, a sugar molecule found in most mammals.2 This may trigger an immune response, which on subsequent exposure can cause mild to severe reactions after consumption of red meat, organ meats, and other products made from mammals.2,4
Prevalence of alpha-Gal syndrome is increasing, and several tick species have been implicated.3 One study noted that out of 143 healthy blood donors from the tick-prevalent Stockholm area, 10 percent had immunoglobulin E (IgE) (i.e., an antibody associated with an immune-system response) to alpha-Gal.5,6 A report from Denmark and Spain showed similar results.6
Alpha-Gal reactions are typically caused by ingestion of red meat (e.g., beef, pork, lamb, rabbit, horse, goat, venison, bear); organ meats (e.g., intestines, hearts, livers, kidneys), and other products made from mammals.2,4 Additional foods that may pose a risk for those with alpha-Gal syndrome include soup-stock cubes, gravy packages, flavor ingredients in prepackaged products, meat extracts used in flavoring, dairy products (e.g., milk, cheese, yogurt, butter), canned tuna (which can be contaminated by dolphin or whale), chicken or fish cooked on a grill contaminated with red meat, pork-gut casings for sausages, pork fat (used for cooking and often found in pastries and venison burgers), and suet.4,8 Meanwhile, fish, reptiles, and birds typically don’t contain alpha-Gal.1
Gelatin, which is normally derived from the collagen in the skin or hooves of large mammals, may also be problematic. In fact, most patients allergic to red meat are sensitized to gelatin, which is the main ingredient in items such as jelly babies (aka gummies), marshmallows, and jello.4,10 While rare, case reports show reactions to topical or oral gelatin can occur from items such as shampoo, collagen implants, catgut sutures, and collagen-derived contact lenses. Additionally, gelatin may pose a risk as a binding agent in tablets, capsules, suppositories, and confectionaries.10 Those with antibodies related to alpha-Gal syndrome also can have reactions to the cancer treatment cetuximab.2
Nonfood sources that may cause a reaction include some forms of monoclonal antibodies, vaccines, gelatin used as a plasma expander, enzyme replacements, bovine or porcine heart valves, antivenom, heparin (i.e., a glycosaminoglycan derived from pig intestines), and stearic acid and/or magnesium stearate (e.g., various tablets including acetaminophen, oxycodone, lisinopril, oxycontin).4
Cooking meat at high temperatures fails to destroy the allergenicity of several red meat proteins, and even a small amount of red meat can cause a severe reaction.7,8
Your healthcare provider may recommend a plan that includes the following.1,2,4,8,11,12
Your healthcare provider may direct you to take one of the following medications to improve your allergy symptoms:
If someone you’re with is having an allergic reaction and showing signs of shock, act quickly. Look for a weak, rapid pulse; trouble breathing; cool, clammy, and pale skin; confusion; and loss of consciousness. Immediately do the following:
After eating red meat, people don’t experience symptoms of alpha-Gal syndrome for at least two hours, and many reactions appear three to five hours or more after consumption. However, many patients don’t have any symptoms, and if symptoms present, they may not occur with every exposure to red meat.7 Over time, symptoms may lessen or even disappear if sensitized individuals don’t receive additional bites from ticks carrying alpha-Gal.8
Symptoms of alpha-Gal syndrome can include:2
Alpha-Gal syndrome may also cause anaphylaxis, which can include symptoms such as:2,9
Skin-prick tests with extracts of pork or beef have been unreliable in diagnosing alpha-Gal syndrome. While intradermal skin tests for several food allergens may be useful, few clinics perform such testing. Thus, most allergists rely on a blood test for IgE to identify alpha-Gal.4
Together with your symptom history, specific IgE blood testing can help determine if you are sensitized to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
The clinical spectrum of symptoms for alpha-Gal syndrome includes anaphylaxis in up to 60 percent of individuals.11 If you experience a severe allergic reaction, you may need an emergency injection of epinephrine and a visit to the emergency room. Once you’ve been diagnosed with alpha-Gal syndrome, your healthcare provider or allergist likely will prescribe an epinephrine autoinjector.8