bf5a- Apps/Techniques/Products/Resources/Contact Us
Polymers play a vital role in our daily lives, making up everything from household products and textiles to transportation, aviation, and aerospace components that move the world. As the polymer microstructure determines the overall performance of these vital functional products, they must be studied and optimized at corresponding scales.
Electron microscopy (EM) plays an indispensable role in the characterization and analysis of polymers. It provides a range of critical information such as; the morphology and composition of new materials, the identity of foreign elements (to trace contamination in the production process), and the failure analysis of components (enabling the creation of improved materials with better performance).
Thermo Fisher Scientific offers a comprehensive solution for polymer analysis across our entire portfolio, including X-ray microtomography (microCT), desktop- and floor-model scanning electron microscopy (SEM), Thermo Scientific DualBeam instruments (focused ion beam-SEM), and transmission electron microscopy (TEM). Together these tools easily fulfill the polymer characterization needs of R&D, technical service, and near-line quality control laboratories.
Video Player is loading.
Current Time 0:00
Duration 2:40
Loaded: 6.25%
Stream Type LIVE
Remaining Time 2:40
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    3D reconstruction of a polymer filter membrane. Data acquired on the Thermo Scientific Apreo Volumescope SEM with a nominal slicing thickness of 50 nm. Backscatter electron images were obtained under a 50 Pa environment.
    Resources
    Applications
    Process control using electron microscopy
    Process control using electron microscopy
    Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
    Quality control and failure analysis using electron microscopy
    Quality control and failure analysis
    Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
    Fundamental Materials Research_R&D_Thumb_274x180_144DPI
    Fundamental Materials Research
    Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
    Aluminum mineral grain found with SEM during parts cleanliness testing
    Technical Cleanliness
    More than ever, modern manufacturing necessitates reliable, quality components. With scanning electron microscopy, parts cleanliness analysis can be brought inhouse, providing you with a broad range of analytical data and shortening your production cycle.
    Style Sheet for Komodo Tabs
    Techniques
    Atomic-Scale Elemental Mapping with EDS
    Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
    Learn more ›
    EDS Analysis with ChemiSEM Technology
    Energy dispersive X-ray spectroscopy for materials characterization.
    Learn more ›
    Cross-sectioning
    Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
    Learn more ›
    Particle analysis
    Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
    Learn more ›
    Differential Phase Contrast Imaging
    Modern electronics research relies on nanoscale analysis of electric and magnetic properties. Differential phase contrast STEM (DPC-STEM) can image the strength and distribution of magnetic fields in a sample and display the magnetic domain structure.
    Learn more ›
    (S)TEM Sample Preparation
    DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
    Learn more ›
    Imaging Hot Samples
    Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
    Learn more ›
    3D Materials Characterization
    Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
    Learn more ›
    Environmental SEM (ESEM)
    Environmental SEM allows materials to be imaged in their native state. This is ideally suited for academic and industrial researchers who need to test and analyze samples that are wet, dirty, reactive, outgassing or otherwise not vacuum compatible.
    Learn more ›
    EDS Elemental Analysis
    Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
    Learn more ›
    SIMS
    The TOF-SIMS (time-of-flight secondary ion mass spectrometry) detector for focused ion beam scanning electron microscopy (FIB-SEM) tools enables high-resolution analytical characterization of all elements in the periodic table, even at low concentrations.
    Learn more ›
    Multi-scale analysis
    Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
    Learn more ›
    3D EDS Tomography
    Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
    Learn more ›
    X-Ray Photoelectron Spectroscopy
    X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
    Learn more ›
    Atomic-Scale Elemental Mapping with EDS
    Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
    Learn more ›
    EDS Analysis with ChemiSEM Technology
    Energy dispersive X-ray spectroscopy for materials characterization.
    Learn more ›
    Cross-sectioning
    Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
    Learn more ›
    Particle analysis
    Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
    Learn more ›
    Differential Phase Contrast Imaging
    Modern electronics research relies on nanoscale analysis of electric and magnetic properties. Differential phase contrast STEM (DPC-STEM) can image the strength and distribution of magnetic fields in a sample and display the magnetic domain structure.
    Learn more ›
    (S)TEM Sample Preparation
    DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
    Learn more ›
    Imaging Hot Samples
    Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
    Learn more ›
    3D Materials Characterization
    Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
    Learn more ›
    Environmental SEM (ESEM)
    Environmental SEM allows materials to be imaged in their native state. This is ideally suited for academic and industrial researchers who need to test and analyze samples that are wet, dirty, reactive, outgassing or otherwise not vacuum compatible.
    Learn more ›
    EDS Elemental Analysis
    Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
    Learn more ›
    SIMS
    The TOF-SIMS (time-of-flight secondary ion mass spectrometry) detector for focused ion beam scanning electron microscopy (FIB-SEM) tools enables high-resolution analytical characterization of all elements in the periodic table, even at low concentrations.
    Learn more ›
    Multi-scale analysis
    Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
    Learn more ›
    3D EDS Tomography
    Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
    Learn more ›
    X-Ray Photoelectron Spectroscopy
    X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
    Learn more ›
    Products
    Style Sheet for Instrument Cards Original
    Spectra 300
    • Highest-resolution structural and chemical information at the atomic level
    • Flexible high-tension range from 30-300 kV
    • Three lens condenser system
    Spectra 200
    • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
    • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
    • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
    Talos F200i TEM
    • Compact design with X-TWIN objective lens
    • Available with S-FEG, X-FEG, and X-CFEG
    • Flexible and fast EDS options for comprehensive elemental analysis
    Talos F200S TEM
    • Intuitive and easy-to-use automation software
    • Available with Super-X EDS for rapid quantitative chemical analysis
    • High-throughput with simultaneous multi-signal acquisition
    Talos F200X TEM
    • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
    • X-FEG and X-CFEG available for the highest brightness and energy resolution
    • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
    Thermo Scientific Talos F200C transmission electron microscope (TEM)
    Talos F200C TEM
    • High-contrast and high-quality TEM and STEM imaging
    • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
    • Large pole piece gap and multiple in situ options
    Thermo Scientific Talos L120C transmission electron microscope (TEM)
    Talos L120C TEM
    • High versatility and stability
    • 4k x 4k Ceta CMOS camera for speed and large FOV
    • TEM magnification range of 25X to 650kX
    • EDS and STEM options for compositional analyses
    Thermo Scientific Helios Hydra plasma focused ion beam scanning electron microscope (DualBeam)
    Helios Hydra DualBeam
    • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
    • Ga-free TEM sample preparation
    • Extreme high resolution SEM imaging
    Helios 5 PFIB DualBeam
    • Gallium-free STEM and TEM sample preparation
    • Multi-modal subsurface and 3D information
    • Next-generation 2.5 μA xenon plasma FIB column
    Helios 5 HX/Helios 5 UX/Helios 5 FX DualBeam
    • Fully automated, high-quality, ultra-thin TEM sample preparation
    • High throughput, high resolution subsurface and 3D characterization
    • Rapid nanoprototyping capabilities
    Thermo Scientific Scios 2 plasma focused ion beam scanning electron microscope (DualBeam)
    Scios 2 DualBeam
    • Full support of magnetic and non-conductive samples
    • High throughput subsurface and 3D characterization
    • Advanced ease of use and automation capabilities
    Thermo Scientific Verios 5 XHR scanning electron microscope (SEM)
    Verios 5 XHR SEM
    • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
    • Easy access to beam landing energies as low as 20 eV
    • Excellent stability with piezo stage as standard
    Thermo Scientific Quattro E scanning electron microscope (SEM)
    Quattro ESEM
    • Ultra-versatile high-resolution FEG SEM with unique environmental capability (ESEM)
    • Observe all information from all samples with simultaneous SE and BSE imaging in every mode of operation
    Thermo Scientific Prisma E scanning electron microscope (SEM)
    Prisma E SEM
    • Entry-level SEM with excellent image quality
    • Easy and quick sample loading and navigation for multiple samples
    • Compatible with a wide range of materials thanks to dedicated vacuum modes
    Thermo Scientific VolumeScope 2 scanning electron microscope (SEM)
    VolumeScope 2 SEM
    • Isotropic 3D data from large volumes
    • High contrast and resolution in high and low vacuum modes
    • Simple switch between normal SEM use and serial block-face imaging
    Thermo Scientific Apreo 2 scanning electron microscope (SEM)
    Apreo 2 SEM
    • High-performance SEM for all-round nanometer or sub-nanometer resolution
    • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
    • Excellent performance at long working distance (10 mm)
    Phenom Pharos G2 Desktop FEG-SEM
    • FEG source with 1 – 20 kV acceleration voltage range
    • <2.0 nm (SE) and 3.0 nm (BSE) resolution @ 20 kV
    • Optional fully integrated EDS and SE detector
    Phenom XL G2 Desktop SEM
    • For large samples (100x100 mm) and ideal for automation
    • <10 nm resolution and up to 200,000x magnification; 4.8 kV up to 20 kV acceleration voltage
    • Optional fully integrated EDS and BSE detector
    Phenom ProX G6 Desktop SEM
    • High performance desktop SEM with integrated EDS detector
    • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
    • Optional SE detector
    Phenom Pure G6 Desktop SEM
    • Entry level desktop SEM
    • Resolution <15 nm; magnification up to 175,000x
    • Longlife CeB6 source
    ESCALAB QXi XPS
    • High spectral resolution
    • Multi-technique surface analysis
    • Extensive sample preparation and expansion options
    Nexsa G2 XPS
    • Micro-focus X-ray sources
    • Unique multi-technique options
    • Dual-mode ion source for monoatomic & cluster ion depth profiling
    K-Alpha XPS
    • High resolution XPS
    • Fast, efficient, automated workflow
    • Ion source for depth profiling
    Thermo Scientific Aquilos 2 cryo focused ion beam (FIB)
    Aquilos 2 Cryo-FIB
    • Automation enables production of multiple lamellas
    • Target and extract your structure of interest with lift-out nano-manipulator
    • 3D visualization for high-resolution tomography
    Athena Software
    Imaging Data Management
    • Ensure traceability of images, data, metadata and experimental workflows
    • Simplify your imaging workflow​
    • Improve collaboration
    • Secure and manage data access​
    Standard Sample Holder
    • Compact stage allowing analysis of samples of up to 100 mm x 100 mm
    • Can be extended with 3 types of resin or metallurgical mount inserts
    • Used with Phenom Desktop SEM
    Thermo Scientific Auto Slice and View 4.0 serial section electron microscopy software
    Auto Slice and View 4.0 Software
    • Automated serial sectioning for DualBeam
    • Multi-modal data acquisition (SEM, EDS, EBSD)
    • On-the-fly editing capabilities
    • Edge based cut placement
    Resin Mount Inserts
    • A unique sample holder concept
    • Available in 3 models for supporting standard sized samples of 25 mm (~1 inch), 32 mm (~1 ¼ inch) and 40 mm (~1 ½ inch) diameter
    Style Sheet to change H2 style to p with em-h2-header class
    Contact us
    Style Sheet for Support and Service footer
    Style Sheet for Fonts
    Style Sheet for Cards
    Electron microscopy services for
    the materials science
    To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.