Flow measurement and process control are critical aspects of producing, handling, and transporting hydrocarbons around the world. With our Thermo Scientific suite of flow computers and flow meters we enable our customers to reduce cost, decrease lost material, automate, and monitor critical points in processes. From field to control room and upstream to downstream, our products provide greater control, confidence, and reliability.
If you are unsure which flow computer is right for you, please view our product selection guide.
View selection guide
Thermo Scientific™ AutoFLEX Flow Computer
New Case Study!
Learn how Thermo Fisher Scientific and Vector Pipeline collaborated to use the Thermo Scientific™ AutoFLEX Flow Computer to establish safe and effective pipelines through real-time hydrocarbon measurement.
Download the Case Study now
Petroleum and natural gas workflows
Petroleum and natural gas workflows
upstream oil and gas sector
Exploration and production
The upstream oil and gas sector encompasses both the exploration and production of hydrocarbon products. Upstream includes the extraction of oil, gas, and water from underground formations in a cost-effective manner. Typical applications include plunger lift, gas lift, separator control, well testing, and automation.
Minimizing production costs and increasing production via enhanced oil recovery (EOR) are critical parameters in the upstream oil and gas sector. Intelligent, customizable, and easy-to-implement solutions are needed in order to maximize operations monitoring.
Recommended products
Transport, control, and store your product reliably with the Thermo Scientific AutoFLEX Flow Computer. From a single measurement point to large delivery points, the AutoFLEX flow computer coupled with the AutoCONFIG interface helps oil and gas transporters maximize flow throughput with quick field deployment and enhanced station control.
Designed on the basis of the Thermo Scientific AutoXP Gas and Liquid Flow Computer, the Thermo Scientific Smart Multivariable Transmitter (SMV) provides accuracy and reliability in hydrocarbon measurement and control. Meet the rigorous demands of the oil and gas industry from production to the refinery. 
  • Ethernet connectivity
  • Bluetooth connectivity
  • Historical archiving
  • Plug and play interface
Read FAQs
The Thermo Scientific AutoXP Gas and Liquid Flow Computer provides ultimate flexibility by providing a unit that can be used as a single-run flow computer or smart multi-variable transmitter. The AutoXP can be configured for both gas and liquid applications utilizing today’s most common primary devices, all while meeting Class I Div 1 requirements.
  • Rugged, robust unit for well testing
  • Ease of connectivity out in the field via Bluetooth
  • Preprogrammed functions simplify automation
  • Through glass keypad interface for enhanced worker safety
  • Designed to meet Class I Div 1 requirements
AutoPILOT PRO Multi-Run Hydrocarbon Flow Computer
Easily integrate the Thermo Scientific AutoPILOT PRO Multi-Run Hydrocarbon Flow Computer into SCADA networks to facilitate data access. The AutoPILOT PRO scales from a simple, single-run measurement application to a multi-run unit with plunger control capabilities, gas lift function, and customizable user interface.
  • Configurable MODBUS Protocol mapping
  • Increase capacity with modular expansion boards
  • Eight plunger methods available
  • Simple connectivity to other instrumentation such as tank levels and gas chromatographs (GCs)
AutoMITTER PRO Smart Multi-Variable Transmitter
The Thermo Scientific AutoMITTER PRO Smart Multi-variable Transmitter features a single board, user-friendly design to simplify maintenance and troubleshooting. Automatic detection via AutoCONFIG software allows for quick and easy field deployment.
  • Single board design
  • In/Out connectors
  • Intrinsically safe
  • Rotary switch addressing
I/P Transducers
Setting the standard in the industry for electronic input signal conversion, the Thermo Scientific STD5000 and STD 6000 Transducers ensure accuracy, reliability and ease-of-use to facilitate operations and increase productivity.
Featured resources
Thermo Scientific™ AutoCONFIG™ Software Introduction
Find, install and download AutoCONFIG software when used with the Thermo Scientific™ AutoPILOT PRO or AutoEXEC flow computers.
Video Player is loading.
Current Time 0:00
Duration 6:33
Loaded: 2.55%
Stream Type LIVE
Remaining Time 6:33
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextThermo Scientific™ AutoCONFIG™ Software Introduction
        Find, install and download AutoCONFIG software when used with the Thermo Scientific™ AutoPILOT PRO or AutoEXEC flow computers.
      2. Now Playing
        Up Next AutoSERIES Flow Computers for Upstream Sector
        The Thermo Scientific AutoXP and AutoPILOT PRO flow computers, built on the AutoCONFIG interface that simplifies configurations and eliminates the need for in depth programming.
      3. Now Playing
        Up NextPlunger Lift
        PlungerLift_8.16.11
    Electronic Flow Measurement at the Wellhead Ensures Productivity
    Electronic flow measurement at the wellhead ensures productivity
    Maximizing well production is the key to maximizing profits. If well pressure drops and water flows from the formation to the well bore and accumulates, it creates a gradual increase in backpressure. 
    Read more
    midstream oil and gas sector
    Gathering and processing
    The midstream oil and gas sector refers to the gathering, processing, and storage of natural gas, crude, or refined petroleum products. Pipelines and other means are used to gather crude oil and natural gas from production sites to processing plants and to downstream customers. Typical applications include custody transfer, storage, gas control, system balance, and automation.
    Successfully managing lost and unaccounted (L&U) levels is a key driver in midstream applications. Instruments that offer effective means of balancing your gathering system will provide accurate, repeatable measurements and will have advanced analytics capabilities.
    Recommended products
    Transport, control, and store your product reliably with the Thermo Scientific AutoFLEX Flow Computer. From a single measurement point to large delivery points, the AutoFLEX flow computer coupled with the AutoCONFIG interface helps oil and gas transporters maximize flow throughput with quick field deployment and enhanced station control.
    Designed on the basis of the Thermo Scientific AutoXP Gas and Liquid Flow Computer, the Thermo Scientific Smart Multivariable Transmitter (SMV) provides accuracy and reliability in hydrocarbon measurement and control. Meet the rigorous demands of the oil and gas industry from production to the refinery. 
    • Ethernet connectivity
    • Bluetooth connectivity
    • Historical archiving
    • Plug and play interface
    Read FAQs
    The Thermo Scientific AutoXP Gas and Liquid Flow Computer provides ultimate flexibility by providing a unit that can be used as a single-run flow computer or smart multi-variable transmitter. The AutoXP can be configured for both gas and liquid applications utilizing today’s most common primary devices, all while meeting Class I Div 1 requirements.
    • Rugged, robust unit for well testing
    • Ease of connectivity out in the field via Bluetooth
    • Preprogrammed functions simplify automation
    • Through glass keypad interface for enhanced worker safety
    • Designed to meet Class I Div 1 requirements
    AutoEXEC 32-Run Expandable Gas and Liquid Flow Computer
    Increase productivity and efficiency using the Thermo Scientific AutoEXEC 32-Run Expandable Gas and Liquid Flow Computer. It is the industry's first flow computer that measures natural gas and petroleum liquids simultaneously. Rugged and easy to use, the AutoEXEC calculates flow at a rate of up to 10 times per second on four to 32 runs to reduce capital expenditures.
    • Complicated AGA and API equations solved rapidly and accurately
    • Preloaded default configuration
    • API 21.2 Custody Transfer Compliance for liquids
    AutoMITTER PRO Smart Multi-Variable Transmitter
    The Thermo Scientific AutoMITTER PRO Multi-variable Transmitter features a single board, user-friendly design to simplify maintenance and troubleshooting. Automatic detection via AutoCONFIG software allows for quick and easy field deployment.
    • Single board design
    • In/Out connectors
    • Intrinsically safe
    • Rotary switch addressing
    I/P Transducers
    Setting the standard in the industry for electronic input signal conversion, the Thermo Scientific STD5000 and STD 6000 Transducers ensure accuracy, reliability and ease-of-use to facilitate operations and increase productivity.
    Featured video
    Video Player is loading.
    Current Time 0:00
    Duration 6:33
    Loaded: 2.50%
    Stream Type LIVE
    Remaining Time 6:33
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      downstream oil and gas sector
      Refining, petrochemical, energy
      The downstream oil and gas sector covers the refining of petroleum crude oil, the processing and purifying of raw natural gas, and the distribution and commerce of petroleum products or natural gas. Typical applications include custody transfer, tank level, flow control, and plant balance.
      Accurate, reliable measurements are critical to realizing maximum profitability of the petroleum or gas product. Instruments that provide precise and repeatable measurements out of the box, as well as units that ensure worker safety, are key factors when considering a solution in the downstream sector.
      Recommended products
      Transport, control and store your product reliably with the Thermo Scientific AutoFLEX Flow Computer. From a single measurement point to large delivery points, the AutoFLEX flow computer coupled with the AutoCONFIG interface helps oil and gas transporters maximize flow throughput with quick field deployment and enhanced station control.
      Designed on the basis of the Thermo Scientific AutoXP Gas and Liquid Flow Computer, the Smart Multivariable Transmitter (SMV) provides accuracy and reliability in hydrocarbon measurement and control. Meet the rigorous demands of the oil and gas industry from production to the refinery. 
      • Ethernet connectivity
      • Bluetooth connectivity
      • Historical archiving
      • Plug and play interface
      Read FAQs
      The Thermo Scientific AutoXP Gas and Liquid Flow Computer provides ultimate flexibility by providing a unit that can be used as a single-run flow computer or smart multi-variable transmitter. The AutoXP can be configured for both gas and liquid applications utilizing today’s most common primary devices, all while meeting Class I Div 1 requirements.
      • Rugged, robust unit for well testing
      • Ease of connectivity out in the field via Bluetooth
      • Preprogrammed functions simplify automation
      • Through glass keypad interface for enhanced worker safety
      • Designed to meet Class I Div 1 requirements
      AutoEXEC 32-Run Expandable Gas and Liquid Flow Computer
      Increase productivity and efficiency using the Thermo Scientific AutoEXEC 32-Run Expandable Gas and Liquid Flow Computer. It is the industry's first flow computer that measures natural gas and petroleum liquids simultaneously. Rugged and easy to use, the AutoEXEC calculates flow at a rate of up to 10 times per second on four to 32 runs to reduce capital expenditures.
      • Complicated AGA and API equations solved rapidly and accurately
      • Preloaded default configuration
      • API 21.2 Custody Transfer Compliance for liquids
      AutoMITTER PRO Smart Multi-Variable Transmitter
      The Thermo Scientific AutoMITTER PRO multi-variable transmitter features a single board, user-friendly design to simplify maintenance and troubleshooting. Automatic detection via AutoCONFIG software allows for quick and easy field deployment.
      • Single board design
      • In/Out connectors
      • Intrinsically safe
      • Rotary switch addressing
      I/P Transducers
      Setting the standard in the industry for electronic input signal conversion, the Thermo Scientific STD5000 and STD 6000 Transducers ensure accuracy, reliability and ease-of-use to facilitate operations and increase productivity.
      Featured video
      Video Player is loading.
      Current Time 0:00
      Duration 6:33
      Loaded: 2.50%
      Stream Type LIVE
      Remaining Time 6:33
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Thermo Scientific AutoCONFIG Software is the robust interface for the Thermo Scientific AutoSERIES of Flow Computers. Designed to empower the user, the interface is simple to configure eliminating the need for in depth programming. 
        Learn more
        AutoCONFIG Software Overview Demo Video
        AutoCONFIG Software overview demo video
        Experience the field-proven platform designed to empower the user. In this demo learn how to configure communication parameters, understand the navigation bar and use the help viewer feature.
        Watch demo
        Infographic: 5 Benefits to Using AutoCONFIG Software in Flow Measurement and Control
        Infographic: 5 benefits to using AutoCONFIG Software in flow measurement and control
        Download infographic
        Video Player is loading.
        Current Time 0:00
        Duration 6:33
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 6:33
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Related applications & support
          Confident Answers at Critical Points in the Oil Refining Process
          Oil and Gas Testing
          Webinar: Fugitive emissions and best practices for leak detection and repair (LDAR)
          environmental monitoring product support and spare parts
          Product support and spares
          Thermo Fisher Scientific Service Solutions are a vital component of your product’s lifecycle. These comprehensive solutions are exclusively created to drive optimal performance of your time-critical instruments.
          When you partner with Thermo Fisher Scientific, you gain access to engineers trained within the same factory your instruments were manufactured. We only ship spare parts that were designed specifically for your instrument. Our expert service team is truly an integrated part of your product’s life cycle.
          Resources
          Contact us
           Yes, I would like to schedule a demo of AutoCONFIG
           No thanks
          *
          *
          *
          *
          *
          *
          *
          *
           I would like to receive marketing and promotional email messages about Thermo Scientific products and services.
          This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.