System overview
SureTect Food Safety PCR System
Visibly simple food pathogen and quality testing—simple workflows designed to get results.
Rapidly and accurately detect foodborne pathogens, and other important food safety and quality targets with the Thermo Scientific™ SureTect Food Safety PCR System.
Empower your operations with simple, automatable workflows and rapid reporting to enable faster product release and interventions. Our food safety PCR assays offer detection of the major pathogens and other important targets in food, pet food and animal feed, primary production samples, and samples from environmental manufacturing.
Whether your laboratory is looking to introduce PCR technology for the first time, or you want to expand the PCR testing services you offer, we can help you find the right solution.
Streamlined workflow
Enrichment
 Report/Confirm
Single enrichment step
Sample preparation and lysis
Run, read and report
Confirm PCR positives
8-24 hours
Single enrichment step of 8-24 hours (depending on target and matrix).
20-40 minutes
Fully automated using the SureTect™ Automation Workflow, or manual preparation with direct lysis in the Applied Biosystems™ SimpliAmp™ Thermal Cycler.
~80 minutes
Perform PCR with the Applied Biosystems QuantStudio™ Instrument using the RapidFinder Analysis Software. Report PCR-negative results immediately (results are automatically interpreted).
24 hours
Confirm PCR-positive results, after overnight incubation, using rapid chromogenic culture media methods.
See how we have innovated to improve workflow & handling:
Learn how you can reduce hands-on time whilst increasing accuracy with the SureTect Automation Workflow:
Video Player is loading.
Current Time 0:00
Duration 2:08
Loaded: 7.64%
Stream Type LIVE
Remaining Time 2:08
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video Player is loading.
    Current Time 0:00
    Duration 2:22
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 2:22
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      For labs adopting PCR, one of the biggest questions surrounds the selection of an automated or manual sample preparation workflow. Our free eBook explores both testing approaches, helping you to select the right strategy for your food lab. Get your copy today!  
      Download eBook
      SureTect PCR Assays
      Validated for a broad range of targets and matrices according to ISO 16140-2:2016 (AFNOR validation) and AOAC certification requirements, the SureTect PCR Assays provide the ultimate solution for all your foodborne pathogen detection requirements.
      • Pre-dispensed lysis and PCR reagents, reducing handling steps and risk of operational error
      • NEW color-coded frames for lysis and PCR stripe well plates for easy assay target identification
      • NEW clearly oriented lysis and assay plates reducing human error
      • NEW color indicator in Proteinase K Reagent enabling tracking of pipetted wells
      • Thin-walled polypropylene wells ensure superior thermal cycling performance for consistent results run after run
      • Internal Positive Control (IPC) in every well providing confidence in every result
      • NEW pierceable seals to reduce handling steps
      • All assays compatible with both manual and automated workflows
      Suretect PCR assays kit
      New features of the upgraded SureTect Assays include:
      Color-coded assay strip plates
       Color-coded Assay Strip Plates
      Easy, safe separation into 8-well strips
      Easy, safe separation of Lysis and Assay plates into 8-well strips
      Cut strips to required number of tubes
      Cut strips to required number of tubes
      Color-indicator in Proteinase K Reagent
      Color-indicator in Proteinase K Reagent
      Efficient plate sealing
      Efficient plate sealing
      Pierceable lysis tube seals
      Pierceable lysis tube seals
      SureTect assay list
      View the full range of SureTect PCR Assays:
      Featured Products
      Visible selective enrichment of Salmonella and Cronobacter species
      Visible selective enrichment of Salmonella and Cronobacter species
      In your busy Salmonella and Cronobacter testing workflows, there is no time for error. A common mistake, particularly in high throughout labs, is the accidental omission or duplication of adding a selective supplement during sample preparation.  For clearer selective enrichment of food and environmental samples, the new Thermo Scientific Oxoid PrecisBlue Supplement has been specifically designed to reduce workflow errors. The addition of an inert blue dye provides a clear visual reference, enabling easy identification of supplemented samples.
      Learn more
      SureCount Salmonella
      Salmonella control you can count on
      Simultaneously quantify & type with the innovative Thermo Scientific™ SureCount™ Salmonella species, Typhimurium and Enteritidis PCR Kit method. A simple, all-in-one solution designed to give you quantification and serotype results for the most common pathogenic Salmonella strains associated with meat and poultry matrices.
      Visit page
      Instruments & software
      MBD-SureTecct-landing-page-CyBio-FeliX-650x600
      Thermo Scientific™ SureTect™ Automation Workflow (using the CyBio™ FeliX)
      With the Thermo Scientific SureTect Automation Workflow you can take your food safety testing to the next level with automated lysis and PCR setup. Utilizing the CyBio FeliX Instrument, the automated workflow allows you to reduce hands-on time whilst increasing the accuracy and reproducibility of results.
      Fully validated by AFNOR and AOAC for use with all SureTect Assays, the instrument's flexible programs, user-friendly interface and rapid yet precise setup ensure maximum efficiency, whilst the compact design saves valuable bench space.
      Time saving
      • Fully automated lysis, PCR setup or complete workflow
      • Rapid 5-minute setup for loading reagents
      Intuitive software
      • Pre-programmed runs for quick setup
      • User-friendly interface with no specialist training required
      Flexible
      • Up to 6 different assays/96 samples on the same run
      • Integrated heating and cooling block
      • Choose between the CHOICE head for simultaneous 8-sample pipetting or the Head R 96
        for 96-sample pipetting
      • Choose from full or partial plate processing
      • Compact design: 58kg and 650x450x700mm
      Control
      • Control your consumable costs and wastage with the Flex Package option, allowing you to use the Head R96 Pipetting head as either a 96-channel pipetting or 8-channel pipetting, at any time  
      • Increase efficiency of operation with choice of runs, reducing service cost and time   
      • Be confident in your processes with the decontamination unit  
      Accurate and traceable
      • Barcoded sample entry for full traceability
      • Precise pipetting
      • Robust and factory calibrated instrument
      Order the SureTect Automation Workflow
      Applied Biosystems™ SimpliAmp™ Thermal Cycler
      Utilize the SimpliAmp Thermal Cycler in the SureTect workflow to prepare enriched samples for PCR by automating the incubation steps for cell lysis samples are ready for PCR in around 20 minutes with minimal hands-on steps required. The SimpliAmp Thermal Cycler will hold samples ready for use at 4 degrees °C at the end of the cycle.
      The SimpliAmp Thermal Cycler is an easy-to-use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow.
      Order the SimpliAmp Thermal Cycler
      Applied BioSystems™ QuantStudio™ 5 Food Safety Real-Time PCR System
      The SureTect PCR Assays are validated to run on the QuantStudio 5 Food Safety PCR system using Thermo Scientific RapidFinder Analysis Software for easy test set-up, results interpretation and data analysis.
      The QuantStudio 5 Food Safety PCR System is a high-performance benchtop platform, combining ease-of-use and system connectivity with powerful options that enable maximum data utility and control. Benefit from the flexibility of a 6-channel, 96-well, cloud-enabled open platform suitable for running our full range of PCR solutions for food safety, authenticity, and quality testing.
      Intuitive and easy-to-use for all levels of experience with touchscreen usability, allowing you to stay connected to your data easily.
      • Multiplex with ease—up to six excitation and six emission filters offer 21 different color combinations, allowing a broad range of detection chemistries and maximum multiplexing
      • Get up and running quickly—factory-calibrated for accuracy, quick installation, and immediate use; pre-optimized protocol templates help minimize training for new users
      • Maximize benchtop space—compact instrument can be configured as a stand-alone or with a computer
      • Open instrument—suitable for all our food safety, authenticity, and quality PCR tests
      • Intuitive, user-friendly software, avoiding subjective interpretation
      • Cloud-enabled, offering the potential for data sharing, analysis, and trending
      • Capacity for processing several hundred samples a day
      Order the QuantStudio 5 Food Safety PCR System
      Thermo Scientific RapidFinder Analysis Software
      An integral part of the SureTect workflow, the RapidFinder Analysis Software is a graphical, easy-to-use tool for food and environmental testing labs to optimize their pathogen detection workflow.
      Simple set-up
      • Intuitive sample set up
      • Flexible plate layout including automatic or customisable options
      • Pre-set, optimized thermal cycling conditions per target
      Automatic data interpretation
      • Fully automated amplification, detection, data collection, and data analysis
      • Quick interpretation of data with flags, notifications, and prompts
      • Detailed report generation for printing or export
      • Interoperability with LIMS or similar data management tool for importing plate set-up and exporting assay results
      • Automatically generates results (presence or absence of the target)
      • Offers interrogation of results for the whole plate, grouped by sample, or for individual PCR tubes
      Order the RapidFinder Analysis Software
      Handling tools
      Easy-to-use tools to simplify your workflow: designed specifically to enhance each step of the SureTect PCR Assay workflow, the new Thermo Scientific™ SureTect Tools enable ease of handling, reducing hands-on time and human error, providing a superior user handling experience.
       ProductDescriptionCat. No.
      PCR Strip Cutting Tool
      Cut Lysis and PCR Assay Strips when fewer than 8 tubes are needed.
      A55329
      PCR Lysis Tube Uncapping Tool
      For removal of Domed Strip Caps from Lysis Strips.
      A55328
      PCR Capping Tool
      For capping Lysis and PCR Assay Plate strips.
      A55327
      96-well PCR Base
      96-well base for Lysis and PCR Assay Strip Plates. Use in Lysis and PCR steps.
      A55330
      MicroAmp adhesive film applicator
      Pack of 5; for ease of applying the Pierceable Film to Lysis Plate strips
      4333183
      PCR Strip Cutting Tool:
      Video Player is loading.
      Current Time 0:00
      Duration 0:14
      Loaded: 67.57%
      Stream Type LIVE
      Remaining Time 0:14
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Capping Tool with Domed Lysis Tube Caps:
        Video Player is loading.
        Current Time 0:00
        Duration 0:12
        Loaded: 94.24%
        Stream Type LIVE
        Remaining Time 0:12
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          Uncapping Tool with Domed Lysis Tube Caps:
          Video Player is loading.
          Current Time 0:00
          Duration 0:11
          Loaded: 85.23%
          Stream Type LIVE
          Remaining Time 0:11
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • default, selected
            SureTect PCR Workflow video:
            Video Player is loading.
            Current Time 0:00
            Duration 8:09
            Loaded: 0.00%
            Stream Type LIVE
            Remaining Time 8:09
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              SureTect Trays
              ProductDescriptionCat. No.
              SimpliAmp Strip-Plate Tray
              For compatibility and ease of loading SureTect Lysis Tubes into the SimpliAmp Thermal Cycler
              A55333
              QuantStudio 5 Strip-Plate Tray
              For compatibility and ease of loading SureTect PCR Assay Tubes into the QuantStudio 5 PCR instrument
              A55332
              ABI 7500 Fast Precision Plate Adapter
              For compatibility and ease of loading SureTect PCR Assay Tubes into the 7500 FAST PCR instrument
              A46347
              SimpliAmp Strip Plate Tray Cover
              Pack of 10 For covering and securing the Lysis Tubes before pipetting
              A55971
              Accessories
              Each SureTect PCR Assay comes with all the reagents and PCR accessories needed to set up your SureTect PCR Assay tests to run on the QuantStudio 5 Food Safety PCR System. Additional SureTect Accessories are available to order separately, including Armadillo low-profile PCR strip plates for balancing when using the Applied Biosystems 7500 Fast Food Safety PCR System and domed lysis tube caps for use if preferred in place of the pierceable lysis seals.
              ProductDescriptionCat. No.
              Pierceable lysis seal
              Pack of 100 seals
              • 2 included with each kit
              • Alternative option to lysis caps
              • Printings around each well for positioning and orientation
              • Strip perforations
              A55331
              Lysis tube caps, domed, pack of 120 strips
              Pack of 120 strips: Optional domed caps that can be used in place of pierceable seals for the Lysis Tubes
              A56895
              Armadillo Low-Profile PCR Strip Plate, 96 well, clear
              For balancing of 7500FAST plates
              AB2696
              Ultra clear qPCR caps
              Additional PCR caps, if required
              AB0866
              SureTect workflow improvement starter pack
              Get up and running quickly and easily with the new, improved SureTect PCR Assay workflow with the convenient SureTect Workflow Starter Pack containing all of the tools and accessories you need.
              ProductDescriptionCat. No.
              SureTect Workflow Starter Pack
              Contains: 1x PCR Strip Cutting Tool, 1x PCR Strip Capping Tool, 1x PCR Strip Uncapping Tool, 2x 96-well PCR Base, 1x SimpliAmp Strip Plate Tray, 1x QuantStudio 5 Strip Plate Tray, 1x ABI 7500 Fast Precision Plate Adapter, 1x SimpliAmp strip plate tray cover, 1x MicroAmp Adhesive Film Applicator and 120x Lysis tube caps–domed
              A56898
              Validation & certification
              SureTect PCR Assay validated workflows
              SureTect PCR Assay workflows are validated for a broad range of matrices according to ISO 16140-2:2016 (AFNOR VALIDATION) and AOAC and fully supported by our dedicated and experienced technical teams.
              View certifications
              A guide to efficiently implementing a new, alternative food testing method in accordance with ISO 16140 - Part 3
              As the global food testing market continues to grow, businesses need faster, more efficient, and cost-effective solutions. That’s where alternative food testing approaches come in, offering rapid, reliable results that align with modern industry demands.
              Our comprehensive eBook, "A guide to efficiently implementing a new, alternative food testing method in accordance with ISO 16140 – Part 3" is your roadmap to navigating, selecting and verifying alternative workflows inline with an internationally recognised standard.
              Download eBook
              Resources
              Video Player is loading.
              Current Time 0:00
              Duration 4:01
              Loaded: 0%
              Stream Type LIVE
              Remaining Time 4:01
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • en (Main), selected
                SureTect workflow video
                Video Player is loading.
                Current Time 0:00
                Duration 8:09
                Loaded: 0.00%
                Stream Type LIVE
                Remaining Time 8:09
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • en (Main), selected
                  Style Sheet for Global Design System
                  Style Sheet for Komodo Tabs
                  Style Sheet for Cards
                  Style Sheet for Fonts
                  Style Sheet for Support and Service footer