Search Thermo Fisher Scientific
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
5 | 350, 488, 561, 594 | 605/20 | 300 | 603 | (in buffer) 5 | microscopy, flow cytometry |
Invitrogen Qdot 605 probe is an excellent red-fluorescent label with outstanding brightness and photostability for imaging and flow cytometry applications. With an emission maximum of ~605 nm, Qdot 605 probe can be excited using 350, 488, 561, or 594 nm laser lines or a standard TRITC filter. Optimum imaging results with brighter signals are achieved with a Qdot 605 filter or light cube.
Organic fluorophores and Qdot probes can be readily combined in multiplexed flow cytometry or imaging studies. Longer-wavelength Qdot probes are also well-suited for multiplexed western blots.
We offer Qdot 605 probe conjugated to a variety of antibodies, peptides, and proteins, optimized for cellular labeling and detection. In addition, reactive dye forms and antibody labeling kits are available to allow you to generate your own antibody conjugates or probes.
Qdot Label Conjugates for Cell and Tissue Staining Search Qdot 605 primary antibodies
Flow cytometry analysis of whole blood cells using streptavidin conjugates of Qdot 605 and Qdot 705. Whole blood cells were lysed with ammonium chloride, then sequentially stained with mouse anti-human CD3 (Biotin), Qdot 605 Streptavidin Conjugate, mouse anti-human CD56 (Biotin), and Qdot 705 Streptavidin Conjugate, with washes between each step. The cells were resuspended and analyzed by flow cytometry using 405 nm excitation, and detected using 605/20 nm and 710/20 nm emission bandpass filters. The dot plot shows Qdot 705 streptavidin conjugate on the Y-axis and Qdot 605 streptavidin conjugate on the X-axis.
Qdot probes are made from nanocrystals of semiconductor material and are broadly compatible with standard fluorescence techniques, but there are some novel aspects to their use that can be optimized to obtain the best possible results. The exceptional brightness and photostability of Qdot probes make it easier to detect low-abundance targets. With narrow and symmetric emission spectra, Qdot probes are also well-suited for multiplexed fluorescence detection using a combination of Qdot probes and organic dyes.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.