NuPAGE Tris-Acetate gels offer increased resolution and higher transfer efficiency of high molecular weight proteins than Tris-Glycine and Bis-Tris gels. In addition, the neutral pH environment helps preserve protein integrity, helping improve the separation and detection of high molecular weight proteins.
Key features of the NuPAGE Tris-Acetate Gels
  • High resolution—optimal separation of high molecular weight proteins
  • Great protein integrity—sample preparation process has been optimized to help preserve your proteins
  • Excellent transfer of large proteins—the specialized gel buffer and lower polyacrylamide concentration near the top of NuPAGE Tris-Acetate gradient gels allows larger proteins to transfer more efficiently
  • High-capacity wells—WedgeWellformat wells are wedge-shaped wells that double the well sample loading capacity and make it easy to load your sample (no specialized pipette tips required), available in mini and midi gel format
Available gel sizes Mini: 8 cm x 8 cm (1 or 1.5 mm thick)
Midi: 8 cm x 13 cm (1 mm thick)
Available well configurations
WedgeWell format, Midi (load up to 100 µL per well): 12+2, 20, 26 wells
WedgeWell format, Mini (load up to 60 µL per well): 10, 12, 15 wells
Midi: 12+2, 20, 26 wells
Mini: 10, 12, 15 wells
Storage conditions2–8°C
Shelf lifeup to 8 months
Recommended sample bufferSDS-PAGE: NuPAGE LDS Sample Buffer
Native-PAGE: Novex Tris-Glycine Native Sample Buffer
Recommended running bufferSDS-PAGE: NuPAGE Tris-Acetate SDS Running Buffer
Native-PAGE: Novex Tris-Glycine Native Running Buffer
Recommended transfer bufferNuPAGE Transfer Buffer
Gel chemistryTris-acetate
Available polyacrylamide concentrations7%, 3–8%
Separation range (denaturing)30–500 kDa
For use with (equipment) mini gelsMini Gel Tank or XCell SureLock Mini-Cell
For use with (equipment) midi gelsSureLock Tandem Midi Gel Tank, Invitrogen XCell4 SureLock Midi-Cell or Bio-Rad Criterion (with adapters only)
Mode of separationMolecular weight
ApplicationsSDS-PAGE, Native-PAGE
*Not all percentages are available in every well type
NuPAGE Tris-Acetate Gels do not contain SDS and so can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using LDS sample buffer and tris-acetate SDS running buffer. For native proteins, traditional Tris-glycine native sample buffer and a Tris-glycine native running buffer should be used.
The NuPAGE Tris-Acetate Gels use a discontinuous buffer system involving three ions:
  • Acetate (–) is from the gel buffer and serves as a leading ion due to its high affinity to the anode relative to other anions in the system. The gel buffer ions are tris (+) and acetate (–) (pH 7.0).
  • Tricine (–) is from the running buffer and serves as the trailing ion. The running buffer ions are tris (+), tricine (–) and dodecylsulfate (pH 8.3).
  • Tris (+) is the common ion present in the gel buffer and running buffer. The Tris-acetate system also operates at a significantly lower operating pH of 8.1 during electrophoresis.
Optimal separation and transfer of high-molecular weight proteins
Reliable separation and transfer of high molecular weight (HMW) proteins is a common challenge for life science researchers. A popular general-use gel is a 4–20% Tris-glycine gel, which can effectively separate a mixed range of proteins. However, HMW proteins will be compressed into a narrow region at the top of the gel. A better option for HMW proteins is a Tris-acetate gel. Tris-acetate gels maintain neutral pH (8.1) and separate HMW proteins with higher resolution than Bis-Tris or Tris-glycine gels. This increased resolution, paired with the easier protein movement afforded by low percentage polyacrylamide, leads to increased transfer efficiencies and higher sensitivity. In addition, the neutral pH environment helps minimize protein modification and results in sharper bands.
Figure 1. Tris-acetate gels enable the best separation of HMW proteins. (A) Optimal results are obtained in the yellow shaded areas. (B) Better transfer is seen using the tris-acetate gel over a 4–20% Tris-glycine gel: 9 ng is visualized on the Tris-acetate gel versus 620 ng visualized on the tris-glycine gradient gel.
Excellent sample integrity
Sample preparation is a crucial step in successful protein analysis. The pH of the traditional Laemmli-style sample buffer changes from 6.8 to 5.2 when heated at 100°C. This lower pH is known to induce aspartyl-prolyl (Asp-Pro) peptide bond cleavage, which leads to protein degradation. By maintaining a >7.0 pH environment, the Invitrogen NuPAGE LDS Sample Buffer preserves protein integrity by minimizing this Asp-Pro cleavage. Sample integrity is maintained throughout electrophoresis with the Invitrogen NuPAGE system; protein degradation is seen in samples prepared with Laemmli (tris-glycine) sample buffer. The Invitrogen NuPAGE Antioxidant running buffer additive greatly minimizes protein oxidation during electrophoresis and keeps reduced protein bands sharp and clear.
A comparison is shown below using a NuPAGE 3–8% Tris-Acetate midi gel, 12+2 well run against a Bio-Rad 7.5% TGX midi gel, 12+2 well. Decreasing amounts of HEK 293 cell lysate prepared in RIPA lysis buffer (48–0.5 µg total protein) were denatured in the respective manufacturer’s sample buffer and subjected to electrophoresis using manufacturer instructions.
NuPAGE Tris-Acetate 3.8% 12+2 well; 0.45 µm PVDF
NuPAGE Tris-Acetate 3.8% 12+2 well; 0.45 µm PVDF
Bio-Rad TGX 7.5% 12+2 well; 0.45 µm PVDF
Bio-Rad TGX 7.5% 12+2 well; 0.45 µm PVDF
Figure 2. Western blots using NuPAGE 3-8% Tris-Acetate midi gels generate sharp bands at high protein and RIPA lysis buffer loads while larger proteins fail to transfer from Bio-Rad 7.5% TGX midi gels. NuPAGE 3-8% Tris-Acetate midi gel, 12+2 well, 12+2 well, was loaded with decreasing total protein amount of HEK293 lysate, subjected to electrophoresis in a SureLock Tandem Midi Gel Tank and transferred onto a 0.45 µm PVDF membrane using the SureLock Tandem Blot Module and the Bio-Rad 7.5 % TGX midi gel, 12+2 well, was subjected to electrophoresis in a Criterion Midi Cell Tank and transferred onto a 0.45 µm PVDF membrane using the Criterion Blotter. For these gels, 10 µL of HiMark Prestained Protein Ladder was used as the protein standard. Both membranes were probed simultaneously for two protein targets (BRCA2 and HDAC1) using chemiluminescent immunodetection. BRCA2 was not detected in the Bio-Rad TGX midi gel, which indicates inefficient transfer of this large protein from the Bio-Rad gel. NuPAGE 3-8% Tris-Acetate precast protein gels were designed for electrophoresis and western blotting of large proteins and large proteins tend to transfer more efficiently from the lower acrylamide concentration near the top of the 3-8% acrylamide gradient gel.
For immunodetection, membranes were blocked for 1 hour in 1X Blocker FL Fluorescent Blocking Buffer. For chemiluminescent detection, the membranes were probed overnight with a mixture of primary antibodies diluted in blocking solution: Rabbit-anti BRCA2 (1:500) and Rabbit anti-HDAC1 (1:5,000,000) followed by an incubation with secondary antibody in 1X Blocker FL: Donkey anti-Rabbit HRP (1:5,000) for 1 hour. Membranes were incubated for 5 minutes with SuperSignal West Dura Extended Duration Substrate and imaged on an iBright Imaging System for the same amount of time and the same image adjustment settings.
Ordering
Catalog # Name Size Price (USD) Qty
EA03755BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
166.65
Online Exclusive
172.00
Save 5.35 (3%)
EA03555BOX NuPAGE™ 7%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA0355BOX NuPAGE™ 7%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA03585BOX NuPAGE™ 7%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA03752BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA0378BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
172.65
Online Exclusive
177.00
Save 4.35 (2%)
EA0375PK2 NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 10-well, 2 Gels/Box Each
36.65
Online Exclusive
37.87
Save 1.22 (3%)
EA03785BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
166.65
Online Exclusive
172.00
Save 5.35 (3%)
EA0358BOX NuPAGE™ 7%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA03552BOX NuPAGE™ 7%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA03752PK2 NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 12-well, 2 Gels/Box Each
36.65
Online Exclusive
37.87
Save 1.22 (3%)
EA0375BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
NuPAGE Tris-Acetate Gel Reagents
Catalog # Name Size Price (USD) Qty
LA0041 NuPAGE™ Tris-Acetate SDS Running Buffer (20X) Each
113.00
LA0050 NuPAGE™ Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels), Contains 1 ea. LA0041, NP0004, NP0005, NP0007 Each
135.65
Online Exclusive
141.00
Save 5.35 (4%)
LC2672 Novex™ Tris-Glycine Native Running Buffer (10X) Each
41.65
Online Exclusive
43.53
Save 1.88 (4%)
LC2673 Novex™ Tris-Glycine Native Sample Buffer (2X) Each
27.48
NP0005 NuPAGE™ Antioxidant Each
41.30
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
19.65
Online Exclusive
20.77
Save 1.12 (5%)
A25977 Mini Gel Tank Each
735.65
Online Exclusive
760.00
Save 24.35 (3%)
B1000 Mini Blot Module Each
480.00
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
80.65
Online Exclusive
84.75
Save 4.10 (5%)
NW2000 Mini Gel Tank and Blot Module Set Each
1,314.65
Online Exclusive
1,368.00
Save 53.35 (4%)
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
185.00
NP0007 NuPAGE™ LDS Sample Buffer (4X), 10mL Each
23.46
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
35.49
Catalog # Name Size Price (USD) Qty
EA03755BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
166.65
Online Exclusive
172.00
Save 5.35 (3%)
EA03555BOX NuPAGE™ 7%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA0355BOX NuPAGE™ 7%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA03585BOX NuPAGE™ 7%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA03752BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA0378BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
172.65
Online Exclusive
177.00
Save 4.35 (2%)
EA0375PK2 NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 10-well, 2 Gels/Box Each
36.65
Online Exclusive
37.87
Save 1.22 (3%)
EA03785BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
166.65
Online Exclusive
172.00
Save 5.35 (3%)
EA0358BOX NuPAGE™ 7%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA03552BOX NuPAGE™ 7%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
EA03752PK2 NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 12-well, 2 Gels/Box Each
36.65
Online Exclusive
37.87
Save 1.22 (3%)
EA0375BOX NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
167.65
Online Exclusive
172.00
Save 4.35 (3%)
NuPAGE Tris-Acetate Gel Reagents
Catalog # Name Size Price (USD) Qty
LA0041 NuPAGE™ Tris-Acetate SDS Running Buffer (20X) Each
113.00
LA0050 NuPAGE™ Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels), Contains 1 ea. LA0041, NP0004, NP0005, NP0007 Each
135.65
Online Exclusive
141.00
Save 5.35 (4%)
LC2672 Novex™ Tris-Glycine Native Running Buffer (10X) Each
41.65
Online Exclusive
43.53
Save 1.88 (4%)
LC2673 Novex™ Tris-Glycine Native Sample Buffer (2X) Each
27.48
NP0005 NuPAGE™ Antioxidant Each
41.30
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
19.65
Online Exclusive
20.77
Save 1.12 (5%)
A25977 Mini Gel Tank Each
735.65
Online Exclusive
760.00
Save 24.35 (3%)
B1000 Mini Blot Module Each
480.00
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
80.65
Online Exclusive
84.75
Save 4.10 (5%)
NW2000 Mini Gel Tank and Blot Module Set Each
1,314.65
Online Exclusive
1,368.00
Save 53.35 (4%)
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
185.00
NP0007 NuPAGE™ LDS Sample Buffer (4X), 10mL Each
23.46
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
35.49
Protein Gel Selection Guide
Protein Gel Selection Guide
Find an excellent mini or midi gel for your protein electrophoresis experiment.
Gel Selection Chart
Protein Gel Conversion Tool
Find an exceptional Invitrogen mini or midi gel to replace your current precast gel from another supplier.
Protein Gel Options Brochure
Gel Migration Chart
Download the Protein Gel Guide for a printable gel migration chart.
Still unsure which precast gel works best for your research needs? Download the Gel Selection Chart
Protocols and Handbooks
Protein Gels Performance Guarantee
Protein Gels Performance Guarantee
We stand behind the performance of our protein gels.
Learn more
Hero banner dark gradient overlay
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.