Search Thermo Fisher Scientific
At Thermo Scientific, we understand the value of your work. Smart-Vue Pro is a precisely engineered remote monitoring and data logging solution to enhance sample security and help prevent unnecessary losses while meeting strict regulatory requirements.
Smart-Vue Pro stands out as a widely-deployed remote monitoring solution for sample management that helps ensure total compliance for regulatory agencies and qualification for GxP protocols. Whether you are eliminating paper processes for the first time or looking for exceptional technology with an easier UI, Smart-Vue Pro will keep your organization in compliance with ease. Smart-Vue Pro’s long range and one-to-many capabilities can help make it smart financial choice for your lab operations. Smart-Vue Pro can be enabled for an on-premise server or with cloud utilization.
Alarms and awareness
Choose from several parameters critical to your laboratory, including temperature, CO2 concentration, differential pressure, any 4-20 mA output or alarm relay and dry contacts. Monitor a whole fleet from multiple OEMs.
Scalable without added complexity
We offer smart digital, wired and wireless sensors and our long-range wireless network (LoRaWAN) is capable of covering multiple labs, floors, or buildings.
Compliance and automation
Save time and improve accuracy by automating data logging. Smart-Vue Pro creates a permanent, secure record of readings, alarms, and acknowledgements to help you meet regulatory requirements according to your SOPs, cGxP, 21 CFR Part 11, ISO 17025, CE, FCC, ISED, WPF (India), CMIIT (China), EMC and electrical safety, among other standards.
Cost effective
Our modular, customizable architecture can mean easy expansion without costly upgrades. New equipment or even buildings can be added to existing Smart-Vue Pro systems and multiple sensors can be connected to a single module.
Robust software
Based on Windows OS client/server architecture, Smart-Vue Pro has excellent management access, control and analysis tools for your collected data with flexibility to set 3 upper and 3 lower limits and create site floor maps.
Advanced hardware
Our module has an impressive 2.4” intuitive color touchscreen and can store 4000 offline measurements per channel. It has easy-to-use plug and play sensor swap capabilities and can provide automatic redundancy with overlapping module zones.
What Smart-Vue Pro can monitor for your lab
Smart-Vue Pro web software is intuitive for the user and friendly to the IT department because it is device and operating system agnostic. The responsive application provides flexible data views, including floorplan overlays, full audit trail compliance (designed according to FDA 21 CFR Part 11 guidelines) and incudes customizable reports, alarms and notifications provided in real-time. Smart-Vue Pro software enables outstanding monitoring and data tools on any device, anywhere, anytime.
The Smart-Vue Pro system can be enabled for an on-premise server solution or with cloud utilization.
We have two options for data management: Cloud or on-premise server solutions. While we recommend cloud-based configurations to enable data access from anywhere, there are certain circumstances where on-premise solutions might be necessary.
Cloud Subscription (annual)
On-Premise Software (single install-one time)
Robust audit trail and documentation
Detailed reporting capabilities including event, sensor and alarm histories enable strong documentation and audit trails for compliance and quality assurance.
Long range wireless connectivity enables a single receiver to cover large areas, replacing the need for repeaters. With minimal IT support needed to set up, the dedicated wireless network requires only an internet connection. Each LoRa gateway can support up to 1000 data logging sensors located up to 9km away.
The final pillar of our Smart-Vue Pro system is our service network. Our specialists are here to help you understand the needs of your facility and work with your IT and network security requirements. We offer plans for installation through upkeep to help your focus remain on your critical science.
For Research Use Only. Not for use in diagnostic procedures.