Use magnetic cell separation for less noise in your final results
Invitrogen MagniSort technology is designed to offer column-free magnetic separation platform for cell enrichment that is simpler, faster, and offers significant cost-savings compared to column-based separation methods. When absolute purity is not necessary, as is often the case with in vitro stimulation of T cells or the derivation of macrophages from peripheral blood monocytes, magnetic cell separation can deliver highly enriched cells without exposure to harsh separation protocols like flow cytometric sorting, or chemical gradients. 
Magnetic Cell Sorting and Separation Process
The MagniSort Assay incorporates an easy-to-follow protocol for isolation of both positive and negative fractions.
Advantages of column-based magnetic cell separation
  • Help save time and money
  • Magnetic bead cell separation treats cells gently with no passage through a column
  • Eliminate cost and waste of disposable columns
  • Process 2 x 109 cells per kit (two times that of leading competitor)
Magnetic cell enrichment kits
Undesired cells in the sample are bound by a specific cocktail of biotinylated antibodies (included in the kits). Streptavidin-coated magnetic beads (included in the kit) are then added to the sample. When the sample is placed in a magnetic field, the undesired cells are sequestered, leaving the desired cells untouched and free in solution. 
The MagniSort Magnet is designed to be used with 12 x 75 mm round-bottom polystyrene tubes and the kits mentioned below.
Catalog # Name Size Price (USD) Qty
8804-6811-71 -
8804-6811-74 MagniSort™ Human CD4 T cell Enrichment Kit Each
1,054.00
8804-6813-74 MagniSort™ Human CD4 Memory T cell Enrichment Kit Each
988.00
8804-6814-71 -
8804-6814-74 MagniSort™ Human CD4 Naïve T cell Enrichment Kit Each
1,004.00
8804-6821-71 -
8804-6821-74 MagniSort™ Mouse CD4 T cell Enrichment Kit Each
819.65
Online Exclusive
878.00
Save 58.35 (7%)
8804-6824-71 -
8804-6824-74 MagniSort™ Mouse CD4 Naïve T cell Enrichment Kit Each
940.00
8802-6831-74 MagniSort™ Human CD4 T cell 2-Step Enrichment Kit Each
1,034.00
8804-6812-74 MagniSort™ Human CD8 T cell Enrichment Kit Each
1,004.00
8804-6815-74 MagniSort™ Human CD8 Memory T cell Enrichment Kit Each
1,016.00
8804-6816-74 MagniSort™ Human CD8 Naïve T cell Enrichment Kit Each
1,004.00
8804-6822-71 -
8804-6822-74 MagniSort™ Mouse CD8 T cell Enrichment Kit Each
788.65
Online Exclusive
850.00
Save 61.35 (7%)
8804-6825-74 MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit Each
902.00
8804-6864-74 MagniSort™ Mouse CD45 Depletion Kit Each
914.00
8804-6827-74 MagniSort™ Mouse B cell Enrichment Kit Each
838.00
8804-6867-74 MagniSort™ Human B Cell Enrichment Kit (2nd Generation) Each
1,010.00
8804-6829-74 MagniSort™ Mouse Hematopoietic Lineage Depletion Kit Each
810.00
8804-6836-74 MagniSort™ Human Hematopoietic Lineage Depletion Kit Each
814.00
8804-6837-74 MagniSort™ Human pan-Monocyte Enrichment Kit Each
1,064.00
8804-6819-74 MagniSort™ Human NK cell Enrichment Kit Each
1,010.00
8804-6828-74 MagniSort™ Mouse NK cell Enrichment Kit Each
836.00
8804-6810-71 -
8804-6810-74 MagniSort™ Human T cell Enrichment Kit Each
896.65
Online Exclusive
1,004.00
Save 107.35 (11%)
8804-6820-71 -
8804-6820-74 MagniSort™ Mouse T cell Enrichment Kit Each
878.00
MSNB-6002-71 MagniSort™ Streptavidin Negative Selection Beads, 25 Tests Each
120.00
Depletion kits
Using a biotinylated antibody and streptavidin-coated magnetic beads, these kits are designed to sequester specific cellular subsets. The unbound cells can then be decanted, and the sample will be depleted of the subset.
The MagniSort Magnet is designed to be used with 12 x 75 mm round-bottom polystyrene tubes and the kits mentioned below.
Catalog # Name Size Price (USD) Qty
8802-6830-74 MagniSort™ Human CD3 Positive Selection Kit Each
916.00
8802-6840-74 MagniSort™ Mouse CD3 Positive Selection Kit Each
920.00
8802-6841-74 MagniSort™ Mouse CD4 Positive Selection Kit Each
920.00
8802-6832-74 MagniSort™ Human CD8 Positive Selection Kit Each
920.00
8802-6842-74 MagniSort™ Mouse CD8 Positive Selection Kit Each
916.00
8802-6860-74 MagniSort™ Mouse CD11b Positive Selection Kit Each
920.00
8802-6861-74 MagniSort™ Mouse CD11c Positive Selection Kit Each
920.00
8802-6834-74 MagniSort™ Human CD14 Positive Selection Kit Each
920.00
8802-6833-74 MagniSort™ Human CD19 Positive Selection Kit Each
874.65
Online Exclusive
902.00
Save 27.35 (3%)
8802-6847-74 MagniSort™ Mouse CD19 Positive Selection Kit Each
902.00
8802-6846-74 MagniSort™ Mouse CD45R (B220) Positive Selection Kit Each
874.65
Online Exclusive
902.00
Save 27.35 (3%)
8802-6848-74 MagniSort™ Mouse CD45.1 Positive Selection Kit Each
920.00
8802-6849-74 MagniSort™ Mouse CD45.2 Positive Selection Kit Each
920.00
8802-6865-74 MagniSort™ Mouse CD45 Positive Selection Kit Each
920.00
8802-6862-74 MagniSort™ Mouse CD49b Positive Selection Kit Each
920.00
8802-6835-74 MagniSort™ Human CD56 Positive Selection Kit Each
920.00
8802-6843-74 MagniSort™ Mouse CD90.1 Positive Selection Kit Each
902.00
8802-6844-74 MagniSort™ Mouse CD90.2 Positive Selection Kit Each
882.00
8802-6838-74 MagniSort™ Mouse CD117 (c-Kit) Positive Selection Kit Each
802.00
8802-6870-74 MagniSort™ Human CD117 (c-Kit) Positive Selection Kit Each
902.00
8802-6817-74 MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit Each
882.00
8802-6863-74 MagniSort™ Mouse F4/80 Positive Selection Kit Each
920.00
MagniSort beads for use with your own antibody
MagniSort Streptavidin Positive Selection Beads may be used with a biotinylated antibody of your choice for either positive selection (desired cells are bound to the bead) or negative selection/depletion (undesired cells are bound to the bead). For negative selection/depletion using a cocktail of antibodies (rather than a single antibody), choose MagniSort Negative Selection Beads.
The MagniSort Magnet is designed to be used with 12 x 75 mm round-bottom polystyrene tubes and the Positive and Negative Selection Beads above.
Learn more about magnetic cell separation
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments