RNA Electrophoresis
Electrophoresis is a common lab technique used to identify, quantify, and purify nucleic acid fragments and assess quality. RNA molecules are negatively charged, and during gel electrophoresis they migrate toward the anode in the presence of an electric current.
The length of RNA generally determines its migration in the gel, since longer RNA molecules move slower than shorter fragments. However, RNA molecules are subject to extensive secondary structure via intramolecular base pairing, and this can affect their gel migration. For most applications, RNA is run using denaturing conditions to disrupt secondary structure. For applications such as resolving different conformations of RNA, native (or nondenaturing) gels are recommended. 
Nondenaturing RNA Electrophoresis
Gel electrophoresis of RNA under nondenaturing conditions maintains the secondary structure of RNA molecules. Agarose is generally preferred to acrylamide because it has lower toxicity and, at the concentrations needed to resolve typical RNA molecules, it is easier to handle. We offer convenient reagents for nondenaturing agarose gel electrophoresis, including hassle-free precast E-Gel™ agarose gels and UltraPure™ reagents to pour your own agarose gels.  
Products for Nondenaturing RNA Electrophoresis:
Denaturing RNA Electrophoresis
To accurately determine the molecular weight of RNA molecules, it is essential to run RNA gels using denaturing conditions. Since northern blots typically seek to characterize RNA molecules based on their size, denaturing electrophoresis is commonly used prior to RNA analysis by northern blotting.. Denaturing conditions disrupt hydrogen bonding so that RNA runs without secondary structure, as single-stranded molecules. There are a number of denaturing agents available, including glyoxal, formamide, and methyl mercury. The disadvantage of these compounds is that they are toxic and therefore should be treated with caution.
Traditionally, formaldehyde has been used as a denaturant for RNA electrophoresis. The NorthernMax™ Kit contains a complete set of RNase-free reagents for running formaldehyde-containing agarose gels. These gels must be poured and run in a fume hood. With the NorthernMax™-Gly Kit, RNA samples are denatured in glyoxal/DMSO loading buffer prior to electrophoresis and run in a formaldehyde-free agarose gel.
For determination of RNA size, a wide range of RNA ladders are available for accurate size and mass estimations, including the Millennium™ Markers.
Products for Nondenaturing RNA Electrophoresis
Catalog # Name Size Price (USD) Qty
AM7150 Millennium™ RNA Markers Each
312.65
Online Exclusive
342.00
Save 29.35 (9%)
AM8676 NorthernMax™ Denaturing Gel Buffer (10X) Each
309.00
AM8552 NorthernMax™ Formaldehyde Load Dye Each
103.00
AM1940 NorthernMax™ Kit Each
788.65
Online Exclusive
812.00
Save 23.35 (3%)
AM8672 NorthernMax™ One-Hour Transfer Buffer Each
302.00
AM8677 NorthernMax™ Prehybridization/Hybridization Buffer Each
400.00
AM8678 NorthernMax™-Gly Gel Prep/Running Buffer (10X) Each
510.65
Online Exclusive
532.00
Save 21.35 (4%)
AM1946 NorthernMax™-Gly Kit Each
908.00
AM8551 NorthernMax™-Gly Sample Loading Dye Each
148.65
Online Exclusive
155.00
Save 6.35 (4%)
750023 UltraPure™ DEPC-Treated Water, 1 L Each
104.65
Online Exclusive
115.00
Save 10.35 (9%)
10977015 UltraPure™ DNase/RNase-Free Distilled Water, 500 mL Each
57.25
16550100 UltraPure™ Agarose-1000 Each
445.65
Online Exclusive
487.00
Save 41.35 (8%)
15575020 UltraPure™ 0.5M EDTA, pH 8.0 Each
103.65
Online Exclusive
115.00
Save 11.35 (10%)
15506017 UltraPure™ Tris Hydrochloride Each
155.65
Online Exclusive
169.00
Save 13.35 (8%)
15557036 UltraPure™ SSC, 20X, 4 L Each
183.65
Online Exclusive
200.00
Save 16.35 (8%)
750018 Denhardt's Solution (50X) Each
211.65
Online Exclusive
235.00
Save 23.35 (10%)
For Research Use Only. Not for human or animal therapeutic or diagnostic use.