Viral RNA/DNA Purification
About viral RNA extraction and viral DNA extraction
Analysis of viruses in biological and environmental samples requires efficient methods for viral nucleic acids that are amenable to a variety of sample types. Viral nucleic acids may need to be isolated from saliva, blood, tissue, and even wastewater or stool samples. Each sample type has unique requirements for optimal nucleic acid extraction and isolation.
The two most common viral nucleic acid isolation technologies are magnetic beads and silica (glass) fiber filters. Both work by taking advantage of their different affinity for nucleic acids based on the concentration of chaotropic salts in the buffer, binding in high-salt buffers and eluting in low-salt buffers.
Silica column isolation of viral RNA and DNA
Silica-containing spin columns for viral RNA/DNA isolation are based on the well-known silica-binding properties of DNA and RNA. The spin column format makes washing the silica easy and efficient.
Our viral spin-column products including PureLink Viral RNA/DNA Mini Kit and GeneJET Viral DNA/RNA Purification Kit are designed for fast and easy isolation of viral RNA or DNA from cell-free samples such as serum, plasma, cerebrospinal fluid, and cell culture supernatant.
  • Performs both viral RNA extraction and viral DNA extraction
  • Yields highly concentrated viral nucleic acids for greater sensitivity in detection
  • Purified nucleic acid is ready for real-time PCR and endpoint analysis
Magnetic bead viral RNA extraction
Magnetic bead-based viral nucleic acid extraction kits utilize small paramagnetic beads that strongly yet reversibly bind to DNA/RNA. Benefits of bead-based MagMAX Viral and Pathogen Nucleic Acid Isolation Kits include:
  • Magnetic beads bind RNA more efficiently than glass fiber filters, resulting in higher and more consistent RNA yields.
  • Magnetic beads can be efficiently transferred and washed by pipette, making the process easily automatable.
  • Magnetic beads are more flexible with regard to sample type, i.e., whole blood and cells
  • Viral nucleic acid can be eluted into a small volume of highly purified material suitable for NGS
Spin column isolation of viral RNA and DNA
All of our viral spin-column kits including PureLink Viral RNA/DNA Mini Kit, Thermo Scientific GeneJET Viral DNA/RNA Purification Kit and PureLink Pro 96 Viral RNA/DNA Kit with 96-well plate are designed for fast and easy isolation of viral RNA or DNA from cell-free samples such as serum, plasma, cerebrospinal fluid, and cell culture supernatant.
  • Complete solution for both viral RNA and DNA
  • Yields highly concentrated viral nucleic acids for greater sensitivity in detection
  • Ideal for real-time PCR and endpoint analysis
Some viral kits are experiencing long lead times but there is another option. See our automated isolation solution using MagMAX Viral/Pathogen Nucleic Acid Isolation kits.
Selection guide: Viral DNA extraction
Viral nucleic extraction kits from Thermo Fisher Scientific are optimized to provide maximum viral DNA/RNA yield, purity, and integrity from a broad range of sample types. Choose the kit that best suits your research needs:
Best forRecovery of low abundance viral DNA/RNAHigh quality DNA/RNA from liquid samplesAutomated extraction of viral DNA/RNA from liquid samplesAutomated extraction of DNA/RNA from range of biological sample typesDNA/RNA extraction from difficult to lyse samples
ProductPureLink Viral RNA/DNA Mini Kit
  PureLink Viral RNA/DNA Mini Kit
GeneJet Viral DNA/RNA Purification KitPureLink Pro 96 Viral RNA/DNA Purification KitMagMAX Prime Viral/Pathogen NA Isolation Kit
MagMAX Prime Viral/Pathogen NA Isolation Kit
MagMAX Microbiome Ultra Nucleic Acid Isolation Kit
  MagMAX Microbiome Ultra Nucleic Acid Isolation Kit
Cat. No.12280050K082112280096AA58145A42357
Starting volume500 µL200–400 µL200 µL200–400 µL200–500 µL or 100–250 mg
Sample typesCell-free plasma, serum, cerebrospinal fluidPlasma, serum, nasal swab, buccal swab, saliva, urineCell-free plasma, serum, cerebrospinal fluidSwabs in transport media (respiratory, genital and lesion swabs), saliva, stool, urine, plasma and serumStool, swabs, transport media, culture media, urine, wastewater, soil
Isolation technology & formatsilica spin columnsilica spin columnsilica- membrane 96-well microplatemagnetic beads/ deep-well platemagnetic beads/ multiwell plate
Protocol time45 mins30–60 mins30 mins30–60 mins (automated)~60 mins
Sample elution volume10–50 µL50 µL50–150 µL60 µL50–200 μL
Automation compatibleNoNoYesYesYes
Number of preps5050384600100
Downstream applications(RT)-PCR, (RT)-qPCR, Northern Blotting(RT)-PCR, (RT)-qPCR, Northern Blotting(RT)-PCR, (RT)-qPCR, Northern Blotting(RT)-PCR, (RT)-qPCR, NGS(RT)-PCR, (RT)-qPCR, NGS
Need to isolate intact virus? Visit SARS-CoV-2 Virus Purification to learn how
Automatic viral nucleic acid purification systems
For fast, easy, and effective high-throughput sample processing, automate your nucleic acid isolation workflow. KingFisher automated sample purification systems can be used to process all MagMAX kits, including MagMAX Viral/Pathogen Nucleic Acid Isolation Kits and MagMAX Prime Viral/Pathogen NA Isolation Kit.
Automated viral DNA extraction protocols for the silica plate-based PureLink Pro 96 Viral RNA/DNA Purification Kit use a vacuum manifold to achieve buffer flow/wash and are available for a variety of XYZ liquid handlers.
Lab essentials for viral RNA extraction
The RNA purification process presents unique challenges, one of which is ribonuclease contamination. Even a trace amount of RNase contamination can sabotage your experiment. Our RNAse-free Reagents and Supplies are designed to be used together to help avoid, detect, and inhibit RNases and nucleases in your lab.
Ordering information for viral nucleic acid isolation kits
Magnetic bead-based kits
Spin columns
Catalog # Name Size Price (USD) Qty
12280050 PureLink™ Viral RNA/DNA Mini Kit Each
364.65
Online Exclusive
389.00
Save 24.35 (6%)
12280096A PureLink™ Pro 96 Viral RNA/DNA Purification Kit Each
2,190.00
K0821 GeneJET Viral DNA/RNA Purification Kit Each
315.00
Resources and support for viral RNA extraction and viral DNA extraction
  • Protocol Videos—Videos to help you isolate nucleic acids using a variety of techniques.
  • DNA & RNA Selection Guide—Find the right purification products.
  • Videos—View our library of DNA & RNA purification and analysis videos.
  • Application Notes—Explore our application notes from scientists sharing data for isolation products
For Research Use Only. Not for use in diagnostic procedures.