Tricine protein gels
The Invitrogen Novex Tricine Gel System is a modification of the traditional tris-glycine gel system that uses a discontinuous buffer system specifically designed for the resolution of low molecular weight proteins.
Interactive selection toolAvailable Novex Tricine Gels
Protein gels performance guarantee ›
Invitrogen precast protein gels are designed and tested to deliver exceptional performance, reliability, reproducibility, and consistency of quality.
Available gel sizesMini: 8 cm x 8 cm (1 mm thick) 
Storage conditions2–8°C
Shelf life1–2 months
Recommended sample bufferTricine SDS sample buffer
Recommended running bufferTricine SDS running buffer
Recommended transfer bufferTris-glycine transfer buffer
Gel chemistry Tricine
Available polyacrylamide concentrations10%, 16%, 10-20%
Separation range2.5-250 kDa
For use with (equipment) mini gelsMini Gel Tank or XCell SureLock Mini-Cell
Mode of separation Molecular weight
Available wells configurations*10, 12, 15
*Not all percentages are available in every well type
Invitrogen Novex Tricine Protein Gels provide separation of low molecular weight proteins and peptides. The Novex Tricine Gel System is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow (Schaegger and von Jagow, 1987), specifically designed for resolving peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and de-stacking of low molecular weight proteins and higher resolution of smaller peptides.
Features of Novex Tricine Protein Gels include:
  • Increased resolution of proteins with molecular weights as low as 2 kDa
  • Improved compatibility with direct sequencing of proteins after transferring to PVDF
  • Minimized protein modification due to the lower pH of the tricine buffering system
  • Minimized protein blow-through during protein transfer
How tricine gels work
In the traditional tris-glycine protein gel system, the resolution of smaller proteins (<10 kDa) is hindered by the continuous accumulation of free dodecyl sulfate (DS) ions from the SDS sample and running buffers in the stacking gel, which causes mixing of the DS ions with smaller proteins and results in fuzzy bands and decreased resolution. The mixing also interferes with the fixing and staining of smaller proteins. The Novex Tricine Gel System uses a low pH in the gel buffer and substitutes tricine for glycine in the running buffer. The smaller proteins and peptides that migrate with the stacked DS ions in the tris-glycine gel system are well separated from DS ions in the Novex Tricine Gel System, offering sharper bands and higher resolution.
Resolution of low molecular weight proteins using Tricine gels
The ability of Novex Tricine gels to resolve low molecular weight proteins was demonstrated in a comparison to Bio-Rad TGX 4-20% Tris-Glycine gels using samples of cleaved caspase-3. Samples were subjected to electrophoresis, followed by protein transfer and western blotting. Cleavage products of 19 kDa and 17 kDa resolved on the Novex 16%  Tricine gel while they did not on the Bio-Rad gel.
Novex Tricine gels
Novex Tricine gels better resolve low molecular weight proteins.
Lane 1: Thermo Scientific PageRuler™ Prestained NIR Protein Ladder;
Lanes 2-9: 1 μL loads per well of a 1.5x dilution series of a Jurkat cell lysate after cytochrome C treatment. After separation on Novex 16% Tricine and Bio-Rad TGX 4-20% Tris-Glycine gels, transfers were probed with primary antibodies labeled with Invitrogen Alexa Fluor™ Plus 680 and Invitrogen Alexa Fluor™ Plus 800 dyes, respectively. The Novex 16% Tricine gel resolved the 17 and 19 kDa bands of cleaved caspase-3, whereas the Bio-Rad TGX Tris-Glycine gel did not provide such resolution.
Catalog # Name Size Price (USD) Qty
EC6695BOX Novex™ 16%, Tricine, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
262.65
Online Exclusive
270.00
Save 7.35 (3%)
EC66252BOX Novex™ 10 to 20%, Tricine, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
262.65
Online Exclusive
270.00
Save 7.35 (3%)
EC66752BOX Novex™ 10%, Tricine, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
258.65
Online Exclusive
269.00
Save 10.35 (4%)
EC6675BOX Novex™ 10%, Tricine, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
258.65
Online Exclusive
269.00
Save 10.35 (4%)
EC66952BOX Novex™ 16%, Tricine, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
262.65
Online Exclusive
270.00
Save 7.35 (3%)
EC6625BOX Novex™ Tricine Mini Protein Gel, 10 to 20%, 1.0 mm, 10-well, 10 Gels/Box Each
262.65
Online Exclusive
270.00
Save 7.35 (3%)
EC66955BOX Novex™ 16%, Tricine, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
258.65
Online Exclusive
270.00
Save 11.35 (4%)
EC66255BOX Novex™ 10 to 20%, Tricine, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
261.65
Online Exclusive
270.00
Save 8.35 (3%)
Catalog # Name Size Price (USD) Qty
LC3675 Tris-Glycine Transfer Buffer (25X) Each
55.25
A25977 Mini Gel Tank Each
735.65
Online Exclusive
760.00
Save 24.35 (3%)
B1000 Mini Blot Module Each
480.00
LC1677 Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675 Each
172.00
LC1676 Novex™ Tricine SDS Sample Buffer (2X) Each
31.55
NW2000 Mini Gel Tank and Blot Module Set Each
1,314.65
Online Exclusive
1,368.00
Save 53.35 (4%)
LC1675 Novex™ Tricine SDS Running Buffer (10X) Each
138.65
Online Exclusive
144.00
Save 5.35 (4%)
Migration chart
Available gel sizesMini: 8 cm x 8 cm (1 mm thick) 
Storage conditions2–8°C
Shelf life1–2 months
Recommended sample bufferTricine SDS sample buffer
Recommended running bufferTricine SDS running buffer
Recommended transfer bufferTris-glycine transfer buffer
Gel chemistry Tricine
Available polyacrylamide concentrations10%, 16%, 10-20%
Separation range2.5-250 kDa
For use with (equipment) mini gelsMini Gel Tank or XCell SureLock Mini-Cell
Mode of separation Molecular weight
Available wells configurations*10, 12, 15
*Not all percentages are available in every well type
Invitrogen Novex Tricine Protein Gels provide separation of low molecular weight proteins and peptides. The Novex Tricine Gel System is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow (Schaegger and von Jagow, 1987), specifically designed for resolving peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and de-stacking of low molecular weight proteins and higher resolution of smaller peptides.
Features of Novex Tricine Protein Gels include:
  • Increased resolution of proteins with molecular weights as low as 2 kDa
  • Improved compatibility with direct sequencing of proteins after transferring to PVDF
  • Minimized protein modification due to the lower pH of the tricine buffering system
  • Minimized protein blow-through during protein transfer
How tricine gels work
In the traditional tris-glycine protein gel system, the resolution of smaller proteins (<10 kDa) is hindered by the continuous accumulation of free dodecyl sulfate (DS) ions from the SDS sample and running buffers in the stacking gel, which causes mixing of the DS ions with smaller proteins and results in fuzzy bands and decreased resolution. The mixing also interferes with the fixing and staining of smaller proteins. The Novex Tricine Gel System uses a low pH in the gel buffer and substitutes tricine for glycine in the running buffer. The smaller proteins and peptides that migrate with the stacked DS ions in the tris-glycine gel system are well separated from DS ions in the Novex Tricine Gel System, offering sharper bands and higher resolution.
Resolution of low molecular weight proteins using Tricine gels
The ability of Novex Tricine gels to resolve low molecular weight proteins was demonstrated in a comparison to Bio-Rad TGX 4-20% Tris-Glycine gels using samples of cleaved caspase-3. Samples were subjected to electrophoresis, followed by protein transfer and western blotting. Cleavage products of 19 kDa and 17 kDa resolved on the Novex 16%  Tricine gel while they did not on the Bio-Rad gel.
Novex Tricine gels
Novex Tricine gels better resolve low molecular weight proteins.
Lane 1: Thermo Scientific PageRuler™ Prestained NIR Protein Ladder;
Lanes 2-9: 1 μL loads per well of a 1.5x dilution series of a Jurkat cell lysate after cytochrome C treatment. After separation on Novex 16% Tricine and Bio-Rad TGX 4-20% Tris-Glycine gels, transfers were probed with primary antibodies labeled with Invitrogen Alexa Fluor™ Plus 680 and Invitrogen Alexa Fluor™ Plus 800 dyes, respectively. The Novex 16% Tricine gel resolved the 17 and 19 kDa bands of cleaved caspase-3, whereas the Bio-Rad TGX Tris-Glycine gel did not provide such resolution.
Catalog # Name Size Price (USD) Qty
EC6695BOX Novex™ 16%, Tricine, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
262.65
Online Exclusive
270.00
Save 7.35 (3%)
EC66252BOX Novex™ 10 to 20%, Tricine, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
262.65
Online Exclusive
270.00
Save 7.35 (3%)
EC66752BOX Novex™ 10%, Tricine, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
258.65
Online Exclusive
269.00
Save 10.35 (4%)
EC6675BOX Novex™ 10%, Tricine, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
258.65
Online Exclusive
269.00
Save 10.35 (4%)
EC66952BOX Novex™ 16%, Tricine, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
262.65
Online Exclusive
270.00
Save 7.35 (3%)
EC6625BOX Novex™ Tricine Mini Protein Gel, 10 to 20%, 1.0 mm, 10-well, 10 Gels/Box Each
262.65
Online Exclusive
270.00
Save 7.35 (3%)
EC66955BOX Novex™ 16%, Tricine, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
258.65
Online Exclusive
270.00
Save 11.35 (4%)
EC66255BOX Novex™ 10 to 20%, Tricine, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
261.65
Online Exclusive
270.00
Save 8.35 (3%)
Catalog # Name Size Price (USD) Qty
LC3675 Tris-Glycine Transfer Buffer (25X) Each
55.25
A25977 Mini Gel Tank Each
735.65
Online Exclusive
760.00
Save 24.35 (3%)
B1000 Mini Blot Module Each
480.00
LC1677 Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675 Each
172.00
LC1676 Novex™ Tricine SDS Sample Buffer (2X) Each
31.55
NW2000 Mini Gel Tank and Blot Module Set Each
1,314.65
Online Exclusive
1,368.00
Save 53.35 (4%)
LC1675 Novex™ Tricine SDS Running Buffer (10X) Each
138.65
Online Exclusive
144.00
Save 5.35 (4%)
Migration chart
For Research Use Only. Not for use in diagnostic procedures.