Scientist working with liquid in cell culture dishes
Thermo Fisher Scientific supplies dependable, high-quality life science research tools, equipment, and materials. Our product offerings include reagents and biochemicals to support academic and biotechnical research. We manufacture and supply solvents for many life science research areas, including genomic and proteomic analysis, cell culture, molecular biology, and imaging.
Contact us
Histological grade solvents are specially prepared for tissue processing and other histological and cytological research procedures. Certificates of Analysis are available online or by scanning the QR code on the label.
Catalog # Name Size Price (USD) Qty
410971000 Glutaric dialdehyde, 8% solution in water Each
135.65
Special offer
Online exclusive
Ends: 26-Sep-2025
159.00
Save 23.35 (15%)
J61984.AP Paraformaldehyde fixative solution Each
158.65
Special offer
Online exclusive
Ends: 26-Sep-2025
186.00
Save 27.35 (15%)
J61984.AK Paraformaldehyde fixative solution Each
90.65
Online Exclusive
101.00
Save 10.35 (10%)
Our research grade solvents for life science applications include DNase-, RNase-, and protease-free molecular biology grade products. Our ethanol solvents can be used with Invitrogen and Thermo Scientific purification kits.
Catalog # Name Size Price (USD) Qty
J67241.AP Chloroform, Molecular Biology Reagent Each -
J67241.K2 Chloroform, Molecular Biology Reagent Each -
J67241.K4 Chloroform, Molecular Biology Reagent Each -
J61059.AP Glycerol, Molecular Biology Grade Each
100.00
J61059.K2 Glycerol, Molecular Biology Grade Each
118.65
Special offer
Online exclusive
Ends: 30-Jun-2025
139.00
Save 20.35 (15%)
T08204K7 80% Ethanol, Molecular Biology Grade Each
187.65
Special offer
Online exclusive
Ends: 30-Jun-2025
220.00
Save 32.35 (15%)
T08204K7CS 80% Ethanol, Molecular Biology Grade Case of 4
776.00
T038181000 Absolute Ethanol, 200 proof, Molecular Biology Grade Each
176.00
T038181000CS Absolute Ethanol, 200 proof, Molecular Biology Grade Case of 6
898.00
T032021000 96% Ethanol, Molecular Biology Grade Each
154.00
T032021000CS 96% Ethanol, Molecular Biology Grade Case of 6
785.00
T036181000 99.9% Isopropanol, Molecular Biology Grade Each
139.65
Online Exclusive
155.00
Save 15.35 (10%)
T036181000CS 99.9% Isopropanol, Molecular Biology Grade Case of 6
822.00
327390010 Water, for molecular biology, DNAse, RNAse and Protease free Each
78.65
Special offer
Online exclusive
Ends: 26-Sep-2025
91.90
Save 13.25 (14%)
327390050 Water, for molecular biology, DNAse, RNAse and Protease free Each
237.65
Special offer
Online exclusive
Ends: 26-Sep-2025
279.00
Save 41.35 (15%)
Thermo Scientific molecular biology grade ethanol is an ultrapure grade ethanol used for the purification and precipitation of biomolecules such as nucleic acids and proteins. It can be used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding.
Key features:
  • Available in several concentrations
  • Tested for DNase, RNase, and protease to ensure absence of these enzymes
  • Meets ACS specifications for sbsolute ethyl alcohol
  • 0.2 micron-filtered
  • Water less than 0.2%
Applications:
  • Purification and precipitation of nucleic acids (DNA and RNA) and proteins
  • Preparation of staining and destaining solutions
  • Dehydration of cells and tissues prior to paraffin wax embedding
  • Extraction medium
  • Chromatographic reagent
Catalog # Name Size Price (USD) Qty
T08204K7 80% Ethanol, Molecular Biology Grade Each
187.65
Special offer
Online exclusive
Ends: 30-Jun-2025
220.00
Save 32.35 (15%)
T08204K7CS 80% Ethanol, Molecular Biology Grade Case of 4
776.00
T032021000 96% Ethanol, Molecular Biology Grade Each
154.00
T032021000CS 96% Ethanol, Molecular Biology Grade Case of 6
785.00
T038181000 Absolute Ethanol, 200 proof, Molecular Biology Grade Each
176.00
T038181000CS Absolute Ethanol, 200 proof, Molecular Biology Grade Case of 6
898.00
High purity isopropanol (IPA) is a staple solvent used in many life science laboratories. Our ultrapure molecular biology grade Thermo Scientific isopropanol can be used in research applications such as purification and precipitation of nucleic acids and proteins, and preservation of biological specimens.
Key features:
  • Ultrapure isopropanol (assay >99.9%) for molecular biology work
  • Tested for DNase, RNase, and protease to ensure absence of these hydrolyzing enzymes
  • Water <0.05%
  • Low UV optical absorbance
  • Variety of product pack sizes to meet various laboratory needs
Applications:
  • Extraction and purification of nucleic acids, proteins, fats, and lipids
  • Preservation of biological specimens (nontoxic alternative to formaldehyde)
  • Disinfectant
  • Chromatography
Catalog # Name Size Price (USD) Qty
T036181000 99.9% Isopropanol, Molecular Biology Grade Each
139.65
Online Exclusive
155.00
Save 15.35 (10%)
T036181000CS 99.9% Isopropanol, Molecular Biology Grade Case of 6
822.00