Artistic rendition of research activities
There are different grades of solvents available on the market to cater to the needs of different laboratory research applications. Normally, the qualities of solvents are set by international organizations such as ASTM International or the American Chemical Society (ACS). Thermo Fisher Scientific offers a wide range of analytical grade solvents for research applications.
Shop now
ACS grade solvents
Our ACS grade solvents are manufactured under specific quality control parameters. The Committee on Analytical Reagents of the ACS is the first of its kind to set the quality requirements for each solvent and develop a validated method to ascertain purity. These solvents are widely used in research laboratories in academic and industrial settings. Certificates of Analysis and MSDS are available upon request.
Catalog # Name Price (USD) Qty
031721.AP Methanol, ACS, absolute, low acetone, 99.8+%
31.30
013455.36 Sodium hydroxide (low chloride), ACS, 97.0% min
55.60
031787.K2 2,2,4-Trimethylpentane, ACS, 99+%
67.80
016371.K7 Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
043894.K2 1-Methyl-2-pyrrolidinone, ACS grade, 99.0+% -
030698.AP Acetone, ACS, 99.5+%
35.50
031344.K7 Ethyl acetate, ACS, 99.5+%
132.65
Special offer
Online exclusive
156.00
Save 23.35 (15%)
043894.K7 1-Methyl-2-pyrrolidinone, ACS grade, 99.0+% -
031733.M6 2-Methoxyethanol, ACS, 99.3+% -
031344.K2 Ethyl acetate, ACS, 99.5+%
46.65
Special offer
Online exclusive
54.20
Save 7.55 (14%)
031787.K7 2,2,4-Trimethylpentane, ACS, 99+%
129.65
Special offer
Online exclusive
152.00
Save 22.35 (15%)
030698.AB Acetone, ACS, 99.5+%
24.70
030760.AP Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT
46.60
036423.M6 Acetonitrile, ACS, 99.5+%
585.65
Special offer
Online exclusive
689.00
Save 103.35 (15%)
016371.AP Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
36.80
043894.AP 1-Methyl-2-pyrrolidinone, ACS grade, 99.0+% -
036423.M1 Acetonitrile, ACS, 99.5+%
207.65
Online Exclusive
231.00
Save 23.35 (10%)
031344.AP Ethyl acetate, ACS, 99.5+%
37.70
031733.AK 2-Methoxyethanol, ACS, 99.3+% -
031068.M6 1-Butanol, ACS, 99.4+%
390.65
Special offer
Online exclusive
459.00
Save 68.35 (15%)
036490.AP Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid
293.00
031068.M1 1-Butanol, ACS, 99.4+%
133.65
Special offer
Online exclusive
157.00
Save 23.35 (15%)
030760.K7 Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT
185.00
030698.M1 Acetone, ACS, 99.5+%
138.00
030760.K2 Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT
71.20
013455.A3 Sodium hydroxide (low chloride), ACS, 97.0% min
84.90
016371.K2 Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
37.65
Special offer
Online exclusive
44.70
Save 7.05 (16%)
031721.M1 Methanol, ACS, absolute, low acetone, 99.8+%
112.00
031787.M1 2,2,4-Trimethylpentane, ACS, 99+%
190.65
Online Exclusive
212.00
Save 21.35 (10%)
031068.AP 1-Butanol, ACS, 99.4+%
41.00
043894.M6 1-Methyl-2-pyrrolidinone, ACS grade, 99.0+% -
036490.AE Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid
79.60
036423.AP Acetonitrile, ACS, 99.5+%
47.65
Special offer
Online exclusive
55.40
Save 7.75 (14%)
031344.M4 Ethyl Acetate, ACS, 99.5+%
556.65
Special offer
Online exclusive
655.00
Save 98.35 (15%)
031733.K2 2-Methoxyethanol, ACS, 99.3+% -
013455.A9 Sodium hydroxide (low chloride), ACS, 97.0% min
326.00
031733.K7 2-Methoxyethanol, ACS, 99.3+% -
033346.AP 4-Methyl-2-pentanone, ACS, 98.5+%
40.50
031344.M1 Ethyl acetate, ACS, 99.5+%
162.65
Online Exclusive
181.00
Save 18.35 (10%)
043894.M1 1-Methyl-2-pyrrolidinone, ACS grade, 99.0+% -
013455.F6 Sodium hydroxide (low chloride), ACS, 97.0% min
601.65
Special offer
Online exclusive
708.00
Save 106.35 (15%)
036423.K2 Acetonitrile, ACS, 99.5+%
70.65
Online Exclusive
78.20
Save 7.55 (10%)
031721.K7 Methanol, ACS, absolute, low acetone, 99.8+%
95.50
013455.22 Sodium hydroxide (low chloride), ACS, 97.0% min
36.65
Special offer
Online exclusive
43.50
Save 6.85 (16%)
031068.K7 1-Butanol, ACS, 99.4+%
119.65
Online Exclusive
133.00
Save 13.35 (10%)
030760.M1 Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT
233.00
031068.K2 1-Butanol, ACS, 99.4+%
48.65
Special offer
Online exclusive
56.50
Save 7.85 (14%)
030698.K7 Acetone, ACS, 99.5+%
108.00
036423.K7 Acetonitrile, ACS, 99.5+%
182.65
Online Exclusive
203.00
Save 20.35 (10%)
033263.AP Perchloric acid, ACS, 60-62%
124.00
030698.K2 Acetone, ACS, 99.5+%
45.90
031721.K2 Methanol, ACS, absolute, low acetone, 99.8+%
37.70
033346.K2 4-Methyl-2-pentanone, ACS, 98.5+%
59.20
031787.AP 2,2,4-Trimethylpentane, ACS, 99+%
48.30
016371.M1 Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Anhydrous solvents
Anhydrous solvents are highly pure solvents made with low water content. Anhydrous solvents produced in laboratories use complex and difficult processes to reduce the moisture content. To minimize the moisture content, we use various dehydration methods, such as column distillation, addition of molecular sieves, and distillation over metallic catalyst.
Commercially available anhydrous solvents can be directly used in reactions, as further purification is not required. These anhydrous solvents are used in moisture-sensitive reactions in both organic and inorganic synthesis and in analytical laboratory applications where yields are maximized by reducing side reactions.
Anhydrous solvents normally have less than 50 ppm of water content. Care should be taken while opening the anhydrous solvents at ambient temperature as they tend to absorb moisture from the atmosphere.
Catalog # Name Size Price (USD) Qty
016767.36 Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT Each
60.60
043799.AE Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles Each
74.65
Online Exclusive
83.00
Save 8.35 (10%)
016799.06 Ethanol-d{6}, anhydrous, 99+% (Isotopic) Each -
043263.K2 n-Hexane, anhydrous Each
86.65
Special offer
Online exclusive
102.00
Save 15.35 (15%)
043263.K7 n-Hexane, anhydrous Each
289.65
Special offer
Online exclusive
341.00
Save 51.35 (15%)
041842.K2 1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles Each
78.65
Special offer
Online exclusive
91.80
Save 13.15 (14%)
042254.AK Tetrahydrofuran, anhydrous, 99.8+%, unstab. Each
41.00
041842.M1 1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles Each
244.65
Online Exclusive
272.00
Save 27.35 (10%)
088488.18 Aluminum chloride, anhydrous, 99.985% (metals basis) Each
93.10
041838.AK Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles Each
36.90
041857.AD tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles Each
39.65
Special offer
Online exclusive
47.00
Save 7.35 (16%)
016799.03 Ethanol-d{6}, anhydrous, 99+% (Isotopic) Each -
088488.30 Aluminum chloride, anhydrous, 99.985% (metals basis) Each
216.65
Online Exclusive
241.00
Save 24.35 (10%)
043997.M1 N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles Each
336.65
Special offer
Online exclusive
396.00
Save 59.35 (15%)
043821.K2 1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under Argon in resealable ChemSeal™ bottles Each
132.00
043821.M1 1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under Argon in resealable ChemSeal™ bottles Each
414.00
041857.AK tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles Each
46.65
Online Exclusive
51.70
Save 5.05 (10%)
043997.K2 N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles Each
152.65
Special offer
Online exclusive
179.00
Save 26.35 (15%)
016767.18 Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT Each
38.00
043799.K2 Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles Each
167.65
Special offer
Online exclusive
197.00
Save 29.35 (15%)
042254.K7 Tetrahydrofuran, anhydrous, 99.8+%, unstab. Each
221.65
Special offer
Online exclusive
260.00
Save 38.35 (15%)
041842.AK 1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles Each
33.10
043263.AK n-Hexane, anhydrous Each
44.70
016767.62 Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT Each
241.00
042254.K2 Tetrahydrofuran, anhydrous, 99.8+%, unstab. Each
111.00
088488.06 Aluminum chloride, anhydrous, 99.985% (metals basis) Each
50.10
041857.M1 tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles Each
337.65
Special offer
Online exclusive
397.00
Save 59.35 (15%)
043821.AE 1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under Argon in resealable ChemSeal™ bottles Each
59.65
Special offer
Online exclusive
70.40
Save 10.75 (15%)
041838.K2 Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles Each
60.90
041857.K2 tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles Each
103.65
Special offer
Online exclusive
122.00
Save 18.35 (15%)