Negishi cross-coupling reaction—a transition metal-catalyzed coupling reaction
In 1972, after the discovery of the nickel-catalyzed cross-coupling of alkenyl and aryl halides with Grignard reagents (Kumada cross-coupling), improvements in functional group tolerance were sought. It was realized that the answer lay in the use of organometallic substrates with metals less electropositive than lithium and magnesium. From the first studies in 1976, extensive research by E. Negishi demonstrated that the best results in terms of reactivity, yield, and stereoselectivity were obtained when organozinc compounds are used in the presence of palladium catalysts. Today, the palladium- or nickel-catalyzed cross-coupling of organozinc compounds with aryl, alkenyl, or alkynyl halides is known as the Negishi cross-coupling reaction.
The use of organozinc reagents allows for a much greater variety of functional groups to be present in both coupling partners than is possible with the Kumada cross-coupling.
Other advantages of the Negishi cross-coupling reaction include high reactivity, high regio- and stereoselectivity, usability in a wide range of applications, few side reactions, and limited toxicity.
The total synthesis of motuporin, a cyclic pentapeptide that is a potent cytotoxin and protein phosphatase-1 inhibitor, utilized the Negishi cross-coupling reaction.
Review available Thermo Scientific products for the Negishi cross-coupling reaction:
Catalog # Name Size Price (USD) Qty
A16281.22 Zinc chloride, anhydrous, 98+% Each
26.65
Special offer
Online exclusive
Ends: 26-Sep-2025
31.00
Save 4.35 (14%)
A16281.36 Zinc chloride, anhydrous, 98+% Each
60.65
Special offer
Online exclusive
Ends: 26-Sep-2025
71.50
Save 10.85 (15%)
A16281.0E Zinc chloride, anhydrous, 98+% Each
218.65
Special offer
Online exclusive
Ends: 30-Jun-2025
257.00
Save 38.35 (15%)
A16281.0C Zinc chloride, anhydrous, 98+% Each
747.65
Special offer
Online exclusive
Ends: 30-Jun-2025
879.00
Save 131.35 (15%)
H32962.AE Zinc chloride, 0.7M soln. in THF Each
118.65
Online Exclusive
132.00
Save 13.35 (10%)
H32962.AP Zinc chloride, 0.7M soln. in THF Each
267.65
Online Exclusive
297.00
Save 29.35 (10%)
H54398.AE Zinc chloride, 1.9M in 2-MeTHF Each -
87908.06 -
14112.14 -
A14089.30 Triphenylphosphine, flake, 99% Each
51.65
Special offer
Online exclusive
Ends: 26-Sep-2025
60.80
Save 9.15 (15%)
A14089.0B Triphenylphosphine, flake, 99% Each
147.65
Special offer
Online exclusive
Ends: 30-Jun-2025
174.00
Save 26.35 (15%)
A14089.0I Triphenylphosphine, flake, 99% Each
528.65
Special offer
Online exclusive
Ends: 30-Jun-2025
622.00
Save 93.35 (15%)
H58659.AD Phenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles Each
212.65
Online Exclusive
236.00
Save 23.35 (10%)
H58544.AD 2-Pyridylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles Each
178.65
Special offer
Online exclusive
Ends: 30-Jun-2025
210.00
Save 31.35 (15%)
H58852.AD Cyclohexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles Each
195.65
Online Exclusive
217.00
Save 21.35 (10%)
44969.06 -
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now