DMEM, low glucose, pyruvate, no glutamine, no phenol red
Save up to 29% on your order of cell culture products. View promotion details >
DMEM, low glucose, pyruvate, no glutamine, no phenol red
Actual product may vary
Gibco™

DMEM, low glucose, pyruvate, no glutamine, no phenol red

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
110540011 L
11054020500 mL
11880036
also known as 11880-036
10 x 500 mL
Catalog number 11054001
Price (USD)
75.65
Online Exclusive
85.25
Save 9.60 (11%)
Each
In stock
Add to cart
Quantity:
1 L
Recurring order eligible. Learn more »
Price (USD)
75.65
Online Exclusive
85.25
Save 9.60 (11%)
Each
Add to cart
Ask our AI about this Product

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: Low Glucose, Sodium Pyruvate

Without: L-glutamine, Phenol Red, HEPES

The complete formulation is available.

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.

Product Use

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineHeLa, 293, Cos-7, and PC-12
Cell TypePrimary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco™
Product TypeDulbecco's Modified Eagle Medium (DMEM)
Quantity1 L
Shelf Life12 Months
ClassificationAnimal Origin-free
Culture TypeAdherent Cell Culture
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesLow Glucose, Sodium Pyruvate, Sodium Bicarbonate
Without AdditivesNo Glutamine, No Phenol Red, No HEPES
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Do you offer media without phenol red (phenol red-free media)?
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3141780Certificate of AnalysisJun 06, 202511054020
3199034Certificate of AnalysisJun 05, 202511054001
3150529Certificate of AnalysisMay 02, 202511054001, 11054020
3079797Certificate of AnalysisApr 25, 202511054001
3126875Certificate of AnalysisMar 09, 202511054001, 11054020
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

Manganese is not present in the formulation of our catalog DMEM media products.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection