DPBS, no calcium, no magnesium
Save up to 29% on your order of cell culture products. View promotion details >
DPBS, no calcium, no magnesium
Gibco™

DPBS, no calcium, no magnesium

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Promo ImagePromotion
Promo code:RCELL25
Save up to 29% across the cell culture workflow - research begins with cells
From now until Dec 31, get up to 29% off media, sera, buffers, reagents and other cell culture products to advance your researchLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging Type
14190359Promo Image10 LBag
14190094
also known as 14190-094
500 mLBottle
14190144Promo Image500 mLBottle
14190250Promo Image10 x 500 mLBottle
14190136Promo Image1000 mLBottle
14190235Promo Image6 x 1000 mLBottle
14190342Promo Image5 LBag
1419036720 LBag
Catalog number 14190359
Price (USD)
610.00
Each
In stock
Add to cart
Quantity:
10 L
Packaging Type:
Bag
Recurring order eligible. Learn more »
Customize this product
Price (USD)
610.00
Each
Add to cart
Ask our AI about this Product

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
AdditiveNo Sodium Pyruvate
ClassificationAnimal Origin-Free
Dilution1 X
For Use With (Application)Mammalian Cell Culture
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 - 300 mOsm/kg
Packaging TypeBag
Phenol Red IndicatorNo Phenol Red
Product LineGibco™
Quantity10 L
Shipping ConditionRoom Temperature
SterilitySterile-filtered
FormLiquid
Product TypeDPBS
Solution TypeDulbecco's Phosphate Buffered Saline
pH7.0 to 7.3
Unit SizeEach
Contents & Storage
Storage conditions: 15°C to 30°C
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What is the shelf life of DPBS after it has been opened?

We cannot provide an official expiration date for open bottles as this depends on the customer's use. Once opened, Thermo Fisher Scientific can no longer guarantee the performance of any product. However, if the storage conditions are followed as recommended, the stated expiration date (36 months from the date of manufacture) should apply to opened bottles as DPBS is extremely stable.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the shelf life of DPBS (Cat. No. 14190136)?

DPBS has a shelf life of 36 months from the date of manufacture, when stored as recommended (15-30 degrees C).

Does Diploid Growth Serum-Reduced Medium (SRM) contain L-glutamine?

The basal Diploid Growth Serum-Reduced Medium already contains 6 mM L-glutamine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do I have to adjust my infection parameters when using Diploid Production Serum-Free Medium (SFM)?

We recommend evaluating performance with Diploid Production Serum-Free Medium using current conditions, however, multiplicity of infection, time of infection, and time of harvest may be different than with conventional media. Closely monitor cells for cytopathic effect and evaluate viral titers at multiple time points.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can Diploid Growth Serum-Reduced Medium (SRM) be supplemented with different types of serum?

Diploid Growth Serum-Reduced Medium can be supplemented with Fetal Bovine Serum, Newborn Calf Serum, Donor Bovine Serum with Iron, or Bovine Serum.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3144480Certificate of AnalysisJul 10, 202514190094, 14190359, 14190144, 14190342, 14190367, 14190136, 14190086, 14190240
3013565Certificate of AnalysisJul 05, 202514190094, 14190359, 14190144, 14190342, 14190367, 14190136, 14190086, 14190240
3166043Certificate of AnalysisJul 05, 202514190094, 14190359, 14190144, 14190342, 14190367, 14190136, 14190086, 14190240
3189212Certificate of AnalysisJul 03, 202514190094, 14190359, 14190144, 14190342, 14190367, 14190136, 14190086, 14190240
3168636Certificate of AnalysisJul 03, 202514190094, 14190359, 14190144, 14190342, 14190367, 14190136, 14190086, 14190240
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (7)

Citations & References
Abstract
A comparison between different human hepatocyte models reveals profound differences in net glucose production, lipid composition and metabolism in vitro.
Authors:Bonanini F,Singh M,Yang H,Kurek D,Harms AC,Mardinoglu A,Hankemeier T
Journal:Experimental cell research
PubMed ID:38499143
Effects of an indole derivative on cell proliferation, transfection, and alternative splicing in production of lentiviral vectors by transient co-transfection.
Authors:Mier NC,Roper DK
Journal:PloS one
PubMed ID:38833479
Lentiviral vectors derived from human immunodeficiency virus type I are widely used to deliver functional gene copies to mammalian cells for research and gene therapies. Post-transcriptional splicing of lentiviral vector transgene in transduced host and transfected producer cells presents barriers to widespread application of lentiviral vector-based therapies. The present study ... More
Method for Large-scale Production of hIPSC Spheroids.
Authors:Lemarié L,Courtial EJ,Sohier J
Journal:Bio-protocol
PubMed ID:38618177
Stem cell spheroids are rapidly becoming essential tools for a diverse array of applications ranging from tissue engineering to 3D cell models and fundamental biology. Given the increasing prominence of biotechnology, there is a pressing need to develop more accessible, efficient, and reproducible methods for producing these models. Various techniques ... More
Macrophage foam cell formation with native low density lipoprotein.
Authors: Kruth Howard S; Huang Wei; Ishii Itsuko; Zhang Wei-Yang;
Journal:J Biol Chem
PubMed ID:12118008
'This investigation has elucidated a mechanism for development of macrophage foam cells when macrophages are incubated with native low density lipoprotein (LDL). LDL is believed to be the main source of cholesterol that accumulates in monocyte-derived macrophages within atherosclerotic plaques, but native LDL has not previously been shown to cause ... More
Microfluidic single-cell real-time PCR for comparative analysis of gene expression patterns.
Authors:Sanchez-Freire V, Ebert AD, Kalisky T, Quake SR, Wu JC,
Journal:Nat Protoc
PubMed ID:22481529
'Single-cell quantitative real-time PCR (qRT-PCR) combined with high-throughput arrays allows the analysis of gene expression profiles at a molecular level in approximately 11 h after cell sample collection. We present here a high-content microfluidic real-time platform as a powerful tool for comparatively investigating the regulation of developmental processes in single ... More
7 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection