N-Acetyl-L-cysteine, 98%
N-Acetyl-L-cysteine, 98%
N-Acetyl-L-cysteine, 98%
N-Acetyl-L-cysteine, 98%
Thermo Scientific Chemicals

N-Acetyl-L-cysteine, 98%

CAS: 616-91-1 | C5H9NO3S | 163.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 160282500
Price (USD)
385.65
Special offer
Online exclusive
454.00
Save 68.35 (15%)
Each
In stock
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
385.65
Special offer
Online exclusive
454.00
Save 68.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS616-91-1
IUPAC Name2-acetamido-3-sulfanylpropanoic acid
Molecular FormulaC5H9NO3S
InChI KeyPWKSKIMOESPYIA-UHFFFAOYNA-N
SMILESCC(=O)NC(CS)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Residue after ignition=<0.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=97.5 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469934Certificate of AnalysisMay 17, 2025160280010, 160282500, 160280500, 160280250
A0469935Certificate of AnalysisFeb 13, 2025160280010, 160282500, 160280500, 160280250
A0469936Certificate of AnalysisFeb 12, 2025160280010, 160282500, 160280500, 160280250
A0468472Certificate of AnalysisJan 31, 2025160280010, 160282500, 160280500, 160280250
A0467557Certificate of AnalysisNov 11, 2024160280010, 160282500, 160280500, 160280250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection