96 Well Black Plate, Coverglass Bottom, CC2 Surface, Pack of 6
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
96 Well Black Plate, Coverglass Bottom, CC2 Surface, Pack of 6
Thermo Scientific™

96 Well Black Plate, Coverglass Bottom, CC2 Surface, Pack of 6

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid; Type: Optical Coverglass Base
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!Learn More
Have Questions?
Catalog NumberQuantity
160376Promo ImageCase of 30
Catalog number 160376
Price (USD)
3,100.00
Case of 30
Estimated availability date 16-Dec-2025
Add to cart
Quantity:
Case of 30
Price (USD)
3,100.00
Case of 30
Add to cart
Ask our AI about this Product
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.

Features:

  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Cell culture surface is ideal for cell-based assays
  • CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
  • Customized bar coding available
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWith Lid
No. per Case30
No. per Pack6
Packaging6/Pk., 30/Cs.
Product TypeOptical Coverglass Base
QuantityCase of 30
Skirt StyleHigh Flange
Array8 x 12
BarcodeCustom Barcoding Available
ColorBlack
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentCC2, Poly-D Lysine Mimic
Volume (Metric) Well400 μL
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 30
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
1299931Certificate of ComplianceDec 11, 2024160376
1314525Certificate of ComplianceDec 11, 2024160376
1361546Certificate of ComplianceDec 11, 2024160376
1292713Certificate of ComplianceDec 11, 2024160376
1274867Certificate of ComplianceDec 11, 2024160376
5 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

Share catalog number, name or link

1x1 image pixel for data collection