Nunc™ F96 MicroWell™ Black and White Polystyrene Plate
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ F96 MicroWell™ Black and White Polystyrene Plate
Nunc™ F96 MicroWell™ Black and White Polystyrene Plate
Nunc™ F96 MicroWell™ Black and White Polystyrene Plate
Nunc™ F96 MicroWell™ Black and White Polystyrene Plate
Thermo Scientific™

Nunc™ F96 MicroWell™ Black and White Polystyrene Plate

Ensure consistent and reliable results using black and white plates optimized for robotics, plate readers and liquid handling systems.
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. per Case
237105Promo ImageBlack50
236105Promo ImageWhite50
236107Promo ImageWhite160
237107Promo ImageBlack160
Catalog number 237105
Price (USD)
347.65
Online Exclusive
378.00
Save 30.35 (8%)
Case of 50
In stock
Add to cart
Color:
Black
No. per Case:
50
Recurring order eligible. Learn more »
Price (USD)
347.65
Online Exclusive
378.00
Save 30.35 (8%)
Case of 50
Add to cart
Ask our AI about this Product

Ensure consistent and reliable results with minimal crosstalk between wells using the Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. With the low flange pinchbar design, these high quality black and white plates are optimized for robotics, plate readers and liquid handling systems. Available with or without cell culture treatment.

  • Black microplates are recommended for fluorescence measurements with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements with maximum reflection and minimal autoluminescence
  • Working volume range: 50 to 250μL/well
  • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
  • Customized bar coding is available
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWith Lid
No. per Case50
No. per Pack1
Packaging50 Cases
Product TypeMicroplate
Quantity50/Cs.
Volume (Metric) Working50 to 250 mL
Array8 x 12
ColorBlack
FormatStandard
MaterialPolystyrene
No. of Wells96
SterilitySterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 50
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
177384Certificate of ComplianceDec 06, 2024237107
179596Certificate of ComplianceDec 06, 2024236107
173287Certificate of ComplianceDec 03, 2024237105
178548Certificate of ComplianceDec 03, 2024237107
162065Certificate of ComplianceDec 03, 2024236107
5 results displayed, search above for a specific certificate

Scientific Resources

Product Information

Share catalog number, name or link

1x1 image pixel for data collection