Liver Digest Medium
Liver Digest Medium
Gibco™

Liver Digest Medium

Liver Digest Medium is a collagenase-dispase medium for dissociation of viable liver cells.Read more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Catalog NumberQuantity
17703034Promo Image500 mL
Catalog number 17703034
Price (USD)
308.00
Each
In stock
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Price (USD)
308.00
Each
Add to cart
Ask our AI about this Product
Liver Digest Medium is a collagenase-dispase medium for dissociation of viable liver cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHepatocyte Cells
Product TypeLiver Digest Medium
Quantity500 mL
Shipping ConditionDry Ice
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
With AdditivesPhenol Red
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2415522Certificate of AnalysisJan 29, 202517703034
2517964Certificate of AnalysisJan 29, 202517703034
2415524Certificate of AnalysisJan 29, 202517703034
2679119Certificate of AnalysisJan 29, 202517703034
2753832Certificate of AnalysisJan 28, 202517703034
5 results displayed, search above for a specific certificate

Safety Data Sheets

Scientific Resources

Product Information

Citations & References (1)

Citations & References
Abstract
Mechanism for peroxisome proliferator-activated receptor-alpha activator-induced up-regulation of UCP2 mRNA in rodent hepatocytes.
Authors: Nakatani Teruyo; Tsuboyama-Kasaoka Nobuyo; Takahashi Mayumi; Miura Shinji; Ezaki Osamu;
Journal:J Biol Chem
PubMed ID:11782473
Peroxisome proliferator-activated receptor-alpha (PPARalpha)activators, fish oil feeding, or fibrate administration up-regulated mitochondrial uncoupling protein (UCP2) mRNA expression in mouse liver by 5-9-fold, whereas tumor necrosis factor-alpha (TNFalpha) also up-regulated UCP2 in liver. In this study, the mechanisms for PPARalpha activators-induced up-regulation of UCP2 mRNA, related to TNFalpha and reactive oxygen ... More
1 total citations

Share catalog number, name or link

1x1 image pixel for data collection