Medium 199, Earle's Salts (10X)
Save up to 29% on your order of cell culture products. View promotion details >
Medium 199, Earle's Salts (10X)
Gibco™

Medium 199, Earle's Salts (10X)

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivationRead more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Catalog NumberQuantity
21180021Promo Image500 mL
Catalog number 21180021
Price (USD)
53.75
Each
In stock
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Customize this product
Price (USD)
53.75
Each
Add to cart
Ask our AI about this Product

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This Medium 199 is modified as follows:
WithWithout
• Phenol Red• HEPES
• L-glutamine• Sodium Bicarbonate


The complete formulation is available.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2.

Medium 199 contains no proteins, lipids, or growth factors. Medium 199 uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Concentrated forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
Concentration10 X
Culture EnvironmentCO2
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality2400 - 2900 mOsm/kg
Product LineGibco™
Product TypeMedium 199
Quantity500 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
ClassificationAnimal Origin
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesGlutamine, Phenol Red, Earle's Balanced Salt Solution (EBSS), Sodium Pyruvate
Without AdditivesNo HEPES, No Sodium Bicarbonate
pH2 to 2.8
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.
Media Formulations
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2549449Certificate of AnalysisJan 29, 202521180021
2487239Certificate of AnalysisJan 29, 202521180021
2549451Certificate of AnalysisJan 29, 202521180021
2650513Certificate of AnalysisJan 29, 202521180021
2687453Certificate of AnalysisJan 29, 202521180021
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection