1x1 image pixel for data collection
UltraPure™ SDS Solution, 10%
UltraPure™ SDS Solution, 10%
Invitrogen™

UltraPure™ SDS Solution, 10%

UltraPure™ 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturantRead more
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
24730020Promo Image1 L
15553027Promo Image4 x 100 mL
Catalog number 24730020
Price (USD)
144.65
Online Exclusive
159.00
Save 14.35 (9%)
Each
In stock
Add to cart
Quantity:
1 L
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
159.00
Save 14.35 (9%)
Each
Add to cart
Ask our AI about this Product
UltraPure™ 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations. It also is used to dissociate nucleic acid-protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and hybridization solutions. UltraPure™ 10% SDS Solution is filtered and ready to use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.1
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting
FormLiquid
Product LineUltraPure™
Product TypeSDS Solution
Quantity1 L
Reagent TypeSDS
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
1 bottle containing 1 L of Ultrapure SDS at 10% concentration (w/v). Store at room temperature.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Why do solutions containing SDS form precipitates?

SDS is a detergent used to denature proteins leading to disruption of cell membranes and dissociation of nucleic acid-protein complexes in DNA and RNA extractions. It tends to come out of solution at concentrations of 10% or greater, especially at low temperatures or in high-salt buffers. SDS can be resolubilized by slowly heating the solution to 65°C with slight agitation.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3094084Certificate of AnalysisMay 23, 202515553027
3177329Certificate of AnalysisApr 20, 202524730020
3118296Certificate of AnalysisApr 20, 202524730020
2938968Certificate of AnalysisFeb 08, 202515553027
2451914Certificate of AnalysisJan 29, 202515553027
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (6)

Citations & References
Abstract
Diversity of human small intestinal Streptococcus and Veillonella populations.
Authors:van den Bogert B, Erkus O, Boekhorst J, de Goffau M, Smid EJ, Zoetendal EG, Kleerebezem M
Journal:FEMS Microbiol Ecol
PubMed ID:23614882
'Molecular and cultivation approaches were employed to study the phylogenetic richness and temporal dynamics of Streptococcus and Veillonella populations in the small intestine. Microbial profiling of human small intestinal samples collected from four ileostomy subjects at four time points displayed abundant populations of Streptococcus spp. most affiliated with S. salivarius, S. thermophilus, ... More
Determine the effect of p53 on chemosensitivity.
Authors:Senturk E, Manfredi JJ
Journal:Methods Mol Biol
PubMed ID:23150441
'The p53 tumor suppressor protein plays a central role in mediating the cellular response to a variety of stresses. Activation of p53 signaling will trigger cell cycle arrest or apoptosis in normal cells, depending on such factors as cell type and genetic context. The ability of a cell to circumvent ... More
Visualization of fas-mediated death-inducing signaling complex formation by immunoprecipitation.
Authors:Cordeiro N, Bidère N
Journal:Methods Mol Biol
PubMed ID:23397387
The prototypical death receptor Fas (also known as CD95 or Apo-1) plays an essential role in the maintenance of lymphocyte homeostasis. Propagation of cell death through Fas relies on the formation of a multiprotein complex at the receptor level known as the death-inducing signaling complex (DISC). Here, we describe an ... More
Collagen I confers gamma radiation resistance.
Authors:Azorin E, González-Martínez PR, Azorin J
Journal:Appl Radiat Isot
PubMed ID:22705234
The effect of collagen on the response of somatomammotroph tumor cells (GH3) to gamma, radiation therapy was studied in vitro. After incubating confluent GH3 cell monolayers in a serum-free, maintaining medium, either with or without collagen, the monolayers were irradiated with 137Cs, gamma radiation. Collagen reduces cell mortality via ERK1/2 ... More
Microarray-based CGH and copy number analysis of FFPE samples.
Authors:Al-Mulla F
Journal:Methods Mol Biol
PubMed ID:21370011
Over the past decade, utilization of microarray technology has flourished in biomedical research. It has evolved rapidly into a revolutionary tool that offers deeper insight into the molecular basis associated with complex diseases, especially in the field of cancer. Specifically, array-based Comparative Genomic Hybridization (aCGH) permits the detection of genome-wide ... More
6 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection