Pierce™ Disposable Columns, 10 mL
Product Image
Thermo Scientific™

Pierce™ Disposable Columns, 10 mL

Thermo Scientific Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety ofRead more
Have Questions?
Catalog NumberQuantity
29924100 columns
Catalog number 29924
Price (USD)
314.65
Online Exclusive
349.00
Save 34.35 (10%)
Each
-
Add to cart
Quantity:
100 columns
Recurring order eligible. Learn more »
Price (USD)
314.65
Online Exclusive
349.00
Save 34.35 (10%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety of resin-based affinity or desalting techniques.

These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10 mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.

Features of these disposable plastic columns:

• For benchtop gravity-flow affinity purification and size exclusion chromatography
• Supplied complete with porous polyethylene discs, stoppers and end caps
• Columns can be pre-packed and stored until needed
• Compatible with most types of aqueous buffer eluents commonly used in chromatography
• Plastic funnels can be attached to expand the buffer reservoir for increased flow rates

Applications:
• Affinity purification/Affinity chromatography
• Immunodepletion
• Size-exclusion chromatography

Related Products
Pierce™ Disposable Column Trial Pack
Pierce™ Disposable Columns, 2 mL
Pierce™ Disposable Columns, 5 mL
Column Funnels for Pierce™ Disposable Columns
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeEmpty Column, Polyethylene Filter
DescriptionDisposable Columns, 10mL
FormatGravity Flow Column
Quantity100 columns
Capacity (Metric)10 mL
Product LinePierce™
TypeSpin Column
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3215339Certificate of AnalysisJun 26, 202529924
3213723Certificate of AnalysisJun 11, 202529924
AC403625Certificate of AnalysisMay 06, 202529924
AB403626Certificate of AnalysisMar 05, 202529924
AB403624Certificate of AnalysisMar 04, 202529924
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection