Bond-Breaker™ TCEP Solution, Neutral pH
Bond-Breaker™ TCEP Solution, Neutral pH
Thermo Scientific™

Bond-Breaker™ TCEP Solution, Neutral pH

Thermo Scientific Bond-Breaker TCEP Solution, Neutral pH is a stable, 0.5M solution of the thiol-free, phosphine-based TCEP compound, useful asRead more
Have Questions?
Catalog NumberQuantity
777205 mL
Catalog number 77720
Price (USD)
204.00
Each
-
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (USD)
204.00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Bond-Breaker TCEP Solution, Neutral pH is a stable, 0.5M solution of the thiol-free, phosphine-based TCEP compound, useful as a 10X stock for addition to SDS-PAGE sample loading buffers to reduce protein disulfide bonds.

Features of Bond-Breaker TCEP Solution, Neutral pH:

Odorless—unlike DTT and β-ME, TCEP is odor-free, contributing to a healthier lab environment
Efficient—5 to 50 mM TCEP thoroughly reduces most peptide or protein disulfide bonds within a few minutes (i.e., just as effective as DTT)
Specific—selective and complete reduction of even the most stable water-soluble alkyl disulfides
Fast—reduces protein disulfides at room temperature and pH 5 in less than five minutes
Stable—resistant to air oxidation; nonvolatile and nonreactive toward other functional groups found in proteins
Versatile—reduces peptides and proteins over a broad range of pH, salt, detergent and temperature conditions
Compatible—removal of the reducing agent is not necessary before most applications, (e.g. histidine-tagged protein purification, maleimide conjugations), because TCEP does not contain sulfhydryl groups

Bond-Breaker TCEP Solution is a potent, odorless, thiol-free reducing agent with broad application to protein and other research involving reduction of disulfide bonds. This product is an effective and convenient replacement for B-mercaptoethanol or DTT in SDS-PAGE sample buffers. The neutral pH of this reagent provides sharp bands and avoids exposing proteins to the strong acid of TCEP·HCl, which can result in hydrolysis and carbohydrate modification.

Considerations for use of Bond-Breaker TCEP Solution:
• Reduction occurs over a wide range of pH (pH 4.0-9.0) and temperature (5 to 95°C).
• Most proteins are reduced efficiently without a denaturant. However, adding a denaturant such as guanidine·HCl may aid in exposing internal disulfides to the Immobilized TCEP.
• Urea is not recommended as a denaturant as it forms cyanates that react with sulfhydryl groups.
• Do not allow metals to contact the TCEP solution as this will decrease TCEP activity.
• Including 5 to 20 mM EDTA in the sample buffer during reduction helps prevent reoxidation of the sulfhydryl groups by divalent metals such as Zn 2+, Cu 2+ and Mg 2+.
• The reduced sample should be used immediately after reduction because disulfides will reform over time.

Related Products
Pierce™ TCEP-HCl
Pierce™ DTT (Dithiothreitol)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product LineBond-Breaker™
Quantity5 mL
Product TypeDisulfide Reducing Reagent
Unit SizeEach
Contents & Storage
Store at room temperature.
four-plex-image-COS7-live-cell zoomed
PG2624-PJT8477-COL022341-Media-Promo-Pod-Graphic-Global-F-278x123
Have questions about this product? Ask our AI assisted search.
I seem to be getting bubbles from the polymer pouch on my Applied Biosystems 3500/3500xL Genetic Analyzer but the polymer level is normal. What is causing it?
I have an Applied Biosystems 3500/3500xL Genetic Analyzer. When do my consumables expire?
Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?
What is the white residue around the Applied Biosystems 3500/3500xL Genetic Analyzer array knob?
What is the difference between the compositions of POP-7 in POP-7 Polymer for Protein Quality Analysis (Cat. Nos. A30936, A31122) and POP-7 Polymer, for 3500/SeqStudio Flex (Cat. Nos. 4393708, A26703, 4393714)?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3214958Certificate of AnalysisJun 16, 202577720
3214631Certificate of AnalysisMay 06, 202577720
AC411363Certificate of AnalysisApr 15, 202577720
AB405897Certificate of AnalysisMar 17, 202577720
ZL402857Certificate of AnalysisFeb 12, 202577720, DNU-77720
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

TCEP, Tris Carboxy Ethyl Phosphene is an alternative sulfhydryl reducing agent for protein samples. It is an extremely potent and effective reducing agent for particularly ‘difficult' proteins. It is compatible with the Tris-Glycine gels and NuPAGE gels. It should be added to the sample buffer for these systems. 20 mM final (maximum) concentration is sufficient for samples. You may add alkylating agents, e.g. Iodine (50 mM Iodoacetic acid), to prevent re-forming of S-S bonds but it is not necessary. Do not heat because this will hydrolyze much of your sample. Instead let the sample sit for several minutes at RT and then load.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection