1x1 image pixel for data collection
D-Luciferin, Potassium Salt
D-Luciferin, Potassium Salt
Invitrogen™
D-Luciferin, Potassium Salt
The ATP-dependent oxidation of luciferin by luciferase produces light, and this system is employed as a reporter in plants, bacteria,Read more
Have Questions?
Catalog NumberQuantity
L291625 mg
Catalog number L2916
Price (USD)
318.00
Each
In stock
Add to cart
Quantity:
25 mg
Recurring order eligible. Learn more »
Price (USD)
318.00
Each
Add to cart
Ask our AI about this Product
The ATP-dependent oxidation of luciferin by luciferase produces light, and this system is employed as a reporter in plants, bacteria, and mammalian cells. Because chemiluminescent techniques are virtually background-free, this reporter gene system is ideal for detecting low-level gene expression.
Molecular Probes™ highly purified synthetic luciferin exhibits physical properties identical to those of natural luciferin. We offer the luciferin free acid (L2911), as well as its water-soluble sodium and potassium salts (L2912, L2916). Our DMNPE-caged D-luciferin (L7085) offers an efficient way to deliver luciferin into intact cells and can be used to supply a continuous source of active luciferin. DMNPE-caged luciferin readily crosses cell membranes. Once inside the cell, a pulse of luciferin can be released by UV photolysis, or a continuous supply of active luciferin can be slowly formed by the action of intracellular esterases found in most eukaryotic cells.
Luciferase Specifications:
• Chemiluminescent after oxidation by luciferase
• Luciferin potassium salt is readily soluble in aqueous buffers
Find More Fluorogenic and Photoactivatable Probes
We offer a number of fluorogenic substrates, photoactivatable probes, and caged probes. Review Substrates for Microsomal Dealkylases, Acetyltransferases, Luciferases and Other Enzymes—Section 10.6 and Photoactivatable Reagents, Including Photoreactive Crosslinkers and Caged Probes—Section 5.3 in the Molecular Probes™ Handbook for more information on these products.
For Research Use. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodChemiluminescence
Quantity25 mg
Shipping ConditionRoom Temperature
SubstrateLuciferin
Substrate PropertiesChemical Substrate
Substrate TypeLuciferase Substrate
TypeD-Luciferin
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Figures
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
3118571Certificate of AnalysisFeb 27, 2025L2916
3080056Certificate of AnalysisNov 14, 2024L2916
2833456Certificate of AnalysisFeb 29, 2024L2916
2685399Certificate of AnalysisJun 16, 2023L2916
2591219Certificate of AnalysisFeb 01, 2023L2916
5 results displayed, search above for a specific certificate
Safety Data Sheets
Citations & References (232)
Citations & References
Abstract
Endogenous endothelial cell nitric-oxide synthase modulates apoptosis in cultured breast cancer cells and is transcriptionally regulated by p53.
Authors:Mortensen K,Skouv J,Hougaard DM,Larsson LI
Journal:The Journal of biological chemistry
PubMed ID:10608825
5-methyltetrahydrofolate, the active form of folic acid, restores endothelial function in familial hypercholesterolemia.
Authors:Verhaar MC, Wever RM, Kastelein JJ, van Dam T, Koomans HA, Rabelink TJ
Journal:Circulation
PubMed ID:9462523
BACKGROUND: Impaired nitric oxide (NO) activity is an early event in the pathogenesis of cardiovascular disease, resulting from either reduced NO formation or increased NO degradation. Administration of tetrahydrobiopterin (BH4), an essential cofactor for NO production, could restore NO activity in familial hypercholesterolemia (FH). Because folates have been suggested to ... More
Sarco/endoplasmic reticulum calcium ATPase-2 expression is regulated by ATF6 during the endoplasmic reticulum stress response: intracellular signaling of calcium stress in a cardiac myocyte model system.
Authors:Thuerauf DJ, Hoover H, Meller J, Hernandez J, Su L, Andrews C, Dillmann WH, McDonough PM, Glembotski CC
Journal:J Biol Chem
PubMed ID:11595740
The recently described transcription factor, ATF6, mediates the expression of proteins that compensate for potentially stressful changes in the endoplasmic reticulum (ER), such as reduced ER calcium. In cardiac myocytes the maintenance of optimal calcium levels in the sarcoplasmic reticulum (SR), a specialized form of the ER, is required for ... More
A novel zinc finger protein TReP-132 interacts with CBP/p300 to regulate human CYP11A1 gene expression.
Authors:Gizard F, Lavallée B, DeWitte F, Hum DW
Journal:J Biol Chem
PubMed ID:11349124
'The human CYP11A1 gene is expressed specifically in steroidogenic tissues and encodes cytochrome P450scc, which catalyzes the first step in steroid synthesis. A region of the 5'-flanking DNA of the gene from nucleotides -155 to -131 (-155/-131) is shown to activate transcription in steroidogenic human placental JEG-3 (1) and adrenal ... More
Tec kinase signaling in T cells is regulated by phosphatidylinositol 3-kinase and the Tec pleckstrin homology domain.
Authors:Yang WC, Ching KA, Tsoukas CD, Berg LJ
Journal:J Immunol
PubMed ID:11123316
'Tec, the prototypical member of the Tec family of tyrosine kinases, is abundantly expressed in T cells and other hemopoietic cell types. Although the functions of Itk and Txk have recently been investigated, little is known about the role of Tec in T cells. Using antisense oligonucleotide treatment to deplete ... More
232 total citations
Other products to consider
Share catalog number, name or link
1x1 image pixel for data collection