1x1 image pixel for data collection
DMEM for SILAC
DMEM for SILAC
Thermo Scientific™

DMEM for SILAC

Analyze protein expression by mass spectrometry using these media or additives for stable isotope labeling with amino acids in cell culture (SILAC).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
88364500 mL
A338226 x 500 mL
Catalog number 88364
Price (USD)
70.97
Special offer
Online exclusive
Ends: 15-Aug-2025
83.50
Save 12.53 (15%)
Each
In stock
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Price (USD)
70.97
Special offer
Online exclusive
Ends: 15-Aug-2025
83.50
Save 12.53 (15%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific™ DMEM for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

Features of DMEM for SILAC:

  • Flexible—liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
  • High quality—medium is sterile, endotoxin-free, and cell culture-compatible

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization, and quantitation of proteins.

This SILAC medium is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis. SILAC media supplemented with heavy amino acids are capable of nearly 100% label incorporation into proteins of living cells.

Related products

  • NeuCode™ Lysine -080, 50 mg (A33613)
  • Fetal Bovine Serum, dialyzed, US origin (26400036)
  • RPMI 1640 Media for SILAC (88365)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeLiquid Medium
For Use With (Equipment)Mass Spectrometer
Label or DyeMass Isotope of Amino Acid
Quantity500 mL
Shipping ConditionRoom Temperature
Workflow StepCell Culture
Detection MethodMass Spectrometry
FormLiquid
Product TypeMedia
Starting MaterialProtein Samples
Unit SizeEach
Contents & Storage
1 x 500 mL, store at 2–8°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Does DMEM for SILAC (Cat. No. 88364, A33822) have the labeled amino acids in it or do those have to be added separately?

DMEM for SILAC (Cat. No. 88364, A33822) is a liquid medium deficient in both L-lysine and L-arginine. It is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
4534381Certificate of AnalysisJul 02, 202588364
AB4534381Certificate of AnalysisMar 11, 202588364
ZJ4375581Certificate of AnalysisOct 15, 202488364
ZB4114631Certificate of AnalysisFeb 17, 202488364
YJ3982312Certificate of AnalysisNov 08, 202388364
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link