Primary Rat Cortex Neurons
Primary Rat Cortex Neurons
Gibco™

Primary Rat Cortex Neurons

Gibco Primary Rat Cortex Neurons are isolated and cryopreserved from day-18 rat embryos. A flexible, ready-to-use and quality alternative toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A10840011 x 106 cells
A10840024 x 106
Catalog number A1084001
Price (USD)
365.00
Each
In stock
Add to cart
Quantity:
1 x 106 cells
Recurring order eligible. Learn more »
Price (USD)
365.00
Each
Add to cart
Ask our AI about this Product
Gibco Primary Rat Cortex Neurons are isolated and cryopreserved from day-18 rat embryos. A flexible, ready-to-use and quality alternative to fresh neurons, each vial contains 1 million cells per mL. These neurons consistently yield post-thaw viability in the range of 50–80% and have high neuronal purity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Donor AttributesNormal
No. of Cells1 x 10^6
Product TypeRat Cortex Neuron
Quantity1 x 106 cells
Tested ForSterility, Viability, Mycoplasma
AgeFetal
Cell TypeNeural Cells
Culture TypeMammalian Cell Culture
FormCryopreserved
SpeciesRat
TissueCortex
Unit SizeEach
Contents & Storage
Store in liquid nitrogen, vapor-phase.
Have questions about this product? Ask our AI assisted search.
Why does the manual for primary rat neurons have the instruction "Do not expose neurons to air at any time"?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What is the difference between Primary Rat Cortex Neurons, Sprague Dawley and Primary Rat Cortex Neurons?

These neurons differ in the rat strain from which they were isolated: Sprague Dawley vs. Fisher 344. These neurons don't expand anymore. They just grow neurites. The manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0001574-PrimaryRatCortex-HippocampusNeurons-UG.pdf) has instructions for culturing the cells. You can use Neurobasal Plus/B-27 Plus/GlutaMAX to culture them, or just Neurobasal/B-27/GlutaMAX. However, the former combination provides much superior performance than the latter and can maintain neurons longer.

I'm currently culturing neurons. Which media system do you recommend using in conjunction with the Poly-D-Lysine matrix?

We recommend using the B-27 Plus Neuronal Culture System when culturing neurons on Poly-D-Lysine. The B-27 Plus Neuronal Culture System is a complete media system that includes the B-27 Plus Supplement and the Neurobasal Plus Medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What cell types will Poly-D-Lysine support as an attachment matrix?

The Poly-D-Lysine matrix has successfully supported the attachment of primary rodent neurons, iPSC-derived neurons and PC12 cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can primary rat neurons be cryopreserved again after they are cultured?

Primary rat neurons cannot be cryopreserved again because they will not revive.

Can primary rat neurons proliferate?

No, primary rat neurons do not proliferate.

Videos

EVOS M7000 microscope Z-stack of rat cortical neurons using a 20X S-Apo objective, DAPI, GFP and RFP LED cubes.

Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
 
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Rat cortical neurons (A1084001) transfected with CellLight??? Golgi-RFP, BacMam 2.0 (C10593). At 48 hours cells were fixed and stained with anti-Tau (MN1000), followed by Goat anti-Mouse Alexa Fluor Plus 488 (A32723) and NucBlue??? Fixed Cell ReadyProbes??? Reagent (R37606). Cells were imaged on an M7000 (AMF7000) using an Olympus 20X objective (AMEP4734) with DAPI (AMEP4650), GFP (AMEP4651) and RFP (AMEP4652) LED cubes.

      Customers who viewed this item also viewed



      Documents & Downloads

      Certificates

      Lot #Certificate TypeDateCatalog Number(s)
      3107118Certificate of AnalysisApr 09, 2025A1084002
      3043931Certificate of AnalysisFeb 12, 2025A1084001
      2494783Certificate of AnalysisJan 29, 2025A1084001
      2494785Certificate of AnalysisJan 29, 2025A1084001
      2530700Certificate of AnalysisJan 29, 2025A1084001
      5 results displayed, search above for a specific certificate

      Safety Data Sheets

      Other products to consider



      Share catalog number, name or link

      1x1 image pixel for data collection