B-27™ Plus Neuronal Culture System
Culturing neurons? Try our new Gibco™ Poly-D-Lysine Solution.
B-27™ Plus Neuronal Culture System
Gibco™
B-27™ Plus Neuronal Culture System
The B-27 Plus Neuronal Culture System is a next-generation media system that provides the highest rate of in vitro survivalRead more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Promo ImagePromotion
Promo code:RACCS25
Save on advanced culture solutions
Have Questions?
Catalog NumberQuantity
A3653401Promo Image1 System
Catalog number A3653401
Price (USD)
292.00
Each
In stock
Add to cart
Quantity:
1 System
Recurring order eligible. Learn more »
Price (USD)
292.00
Each
Add to cart
Ask our AI about this Product
The B-27 Plus Neuronal Culture System is a next-generation media system that provides the highest rate of in vitro survival of primary rodent and human stem cell-derived neurons. Comprised of B-27 Plus Supplement (50X) and Neurobasal Plus Medium, this system is an evolution of the most cited neuronal cell culture products, B-27 Supplement and Neurobasal Medium. It features an optimized formulation, upgraded manufacturing process, and more stringent quality control of raw materials and final product. These improvements enable the highest neuronal cell survival, improved electrophysiological activity, and enhanced functional maturity versus other neuronal cell culture media systems.
The B-27 Plus Neuronal Culture System offers:
• At least 50% higher survival rate of primary rat neurons compared to classic B-27 Supplement and Neurobasal Medium
Highest survival rate of primary rat, mouse, and human stem cell-derived neurons compared to other commercially available serum-free neuronal media systems
Improved electrophysiological activity and maturation—more consistent, stable, and synchronized spontaneous activity in primary rodent neurons after 10 days compared to BrainPhys™ Neuronal Medium
Accelerates neurite outgrowth and increases the number of synaptic-positive puncta
Available as a 50X, 10 mL supplement with 500 mL basal medium, the B-27 Plus Neuronal Culture System can seamlessly replace other neuronal cell culture systems, such as classic B-27 Supplement and Neurobasal Medium. The individual components of the system are also available for purchase separately.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeural Cells
Product LineB-27™, Neurobasal™
Product TypeNeuronal Culture System
Quantity1 System
Shelf Life12 Months
Shipping ConditionAmbient
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
Unit SizeEach
Contents & Storage
• 10 mL B-27 Plus Supplement (50X), store at –5 to –20°C, protect from light
• 500 mL Neurobasal Plus Medium, store at 2–8°C, protect from light
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Figures
Customers who viewed this item also viewed
Frequently asked questions (FAQs)
We recommend using Neurobasal Plus Medium with B-27 Plus Supplement for long-term culture of mixed hippocampal cells. For short-term culture of a pure population of hippocampal neurons, we would recommend using Neurobasal-A Medium with B-27 Supplement.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
We recommend using the B-27 Plus Neuronal Culture System when culturing neurons on Poly-D-Lysine. The B-27 Plus Neuronal Culture System is a complete media system that includes the B-27 Plus Supplement and the Neurobasal Plus Medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Yes, we have shown improved electrophysiological activity with the B-27 Plus Neuronal Culture System
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
We recommend performing half medium exchanges every 2-3 days post-plating. Make sure that neurons are not completely exposed to air by not removing all of the medium during exchanges.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Other products to consider
Share catalog number, name or link
1x1 image pixel for data collection