Fetal Bovine Serum, exosome-depleted
Fetal Bovine Serum, exosome-depleted
Gibco™

Fetal Bovine Serum, exosome-depleted

Gibco™ Exosome-Depleted FBS streamLines and standardizes cell culture workflows for exosome research. It is ready to use and performance tested to help save time and enable optimal exosome preparations.
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A2720803Promo Image50 mLOne Shot
A2720801Promo Image500 mLBottle
Catalog number A2720803
Price (USD)
247.00
Each
In stock
Add to cart
Quantity:
50 mL
Format:
One Shot
Recurring order eligible. Learn more »
Price (USD)
247.00
Each
Add to cart
Ask our AI about this Product

Gibco™ Exosome-Depleted FBS streamLines and standardizes cell culture workflows for exosome research. It is ready to use and performance tested to help save time and enable optimal exosome preparations.

  • Lowest exosome background
  • Enables consistent results: each lot tested for cell culture performance
  • No preparation needed: ultracentrifugation is not required

Exosome-Depleted FBS has been depleted of ≥90% of endogenous exosomes and is intended for use in cell culture exosome isolation assays. Every lot is tested for exosome depletion and culture performance, and our standard quality control assays are conducted to help ensure optimal and consistent results. This ready-to-use FBS for cell culture exosome isolations helps eliminate the risks associated with FBS reagent preparation by ultracentrifugation, for optimal assay results and increased experimental throughput.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Product LineOne Shot™
Shipping ConditionFrozen
SpeciesBovine
AgeFetal
Country of OriginUnited States
FormLiquid
FormatOne Shot
Product TypeFetal Bovine Serum
Quantity50 mL
Serum TreatmentExosome-depleted
SterilitySterile
Unit SizeEach
Contents & Storage
Storage conditions: ≤-10°C
Shipping conditions: Frozen
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
U3151924RPCertificate of AnalysisMay 25, 2025A2720801, A2720803
U3196886RPCertificate of AnalysisMay 23, 2025A2720801, A2720803
U3189883RPCertificate of AnalysisMay 18, 2025A2720801, A2720803
U3151921RPCertificate of AnalysisMay 11, 2025A2720801, A2720803
U3151918RPCertificate of AnalysisApr 27, 2025A2720801, A2720803
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

  1. Heating inactivates complement. Active complement can participate in cytolytic events, contract smooth muscle, release histamine from mast cells and platelets, and activate lymphocytic and macrophage cells. Applications where heat-inactivated serum is recommended include immunological studies and culturing of embryonic stem cells (ESCs), insect cells, and smooth muscle cells.
  2. Heat inactivation helps to achieve bottle-to-bottle and lot-to-lot stability by neutralizing many factors that can vary largely from lot to lot.
  3. There aren't necessarily disadvantages to heat inactivation of FBS, but there is some evidence that suggests there may be no added benefit to it unless you are carrying out immune studies.

Note: Heat inactivation is performed in a 56 degrees C water bath for 30 min with swirling every 10 min or so for heat distribution and to lower the degree of protein aggregation/flocculant precipitation. Note: If the time or temperature is exceeded, the serum may thicken to a gel. If this occurs, the serum is no longer usable. Unnecessary heat inactivation can take up time and potentially lead to wasted reagents if a mistake is made during the protocol1.

1. Pellerin, et al., Bioengineering, published in 2021.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Yes, the material undergoes USP sterility testing, which includes mycoplasma testing.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Exosome-Depleted FBS is made from Gibco FBS (qualified, US-origin) in the Value category.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Serum can be kept at 2-8 degrees C for up to 4 weeks.

Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.

Yes, we also offer a 10 X 50 mL package, which is equivalent to a 500 mL bottle of FBS.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection