Orion™ AQUAfast™ II Chemistries Test Kits
Orion™ AQUAfast™ II Chemistries Test Kits
Thermo Scientific™

Orion™ AQUAfast™ II Chemistries Test Kits

Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludes
AC2030HydrazineKit: Hydrazine Powder, Spoon
AC2002Alkalinity100 Alkalinity-M Tablets
AC3002PCaCO3
AC2027AluminumSet of 2: 50 Aluminum No 1 Tablets, 50 Aluminum No
AC4P27Aluminum
AC2012AmmoniaSet of 2: 50 Ammonia No 1 Tablets, 50 Ammonia No 2
AC2035Reagent TabletBromine
AC2017ChlorideSet of 2: 50 Chloride T1 Tablets, 50 Chloride T2 T
AC2099Chlorine Dioxide100 DPD No 1 Tablets
AC2071Chlorine100 DPD No 1 Tablets
AC2070ChlorineSet of 2: 50 DPD No 1 Tablets, 50 DPD No 3 Tablets
AC2072Chlorine100 DPD No 4 Tablets
AC3072Chlorine, Sodium Chlorate
AC2029CopperSet of 2: 50 Copper No 1 Tablets, 50 Copper No 2 Tablets
AC2065ZincSet of 2: 50 Copper/Zinc LR Tablets, 50 EDTA Table
AC2098Cyanuric Acid
AC3032C2Calcium
AC3032CCalcium
AC3032TMetallphthalein Method
AC3069Hydrogen Peroxide
AC2078Iron100 Iron LR Tablets
AC2055ManganeseSet of 2: 50 Manganese LR1 Tablets, 50 Manganese L
AC2007Nitrate50 x Nitrite HR No. 1 Tablets, 50 x Nitrite HR No. 2 Tablets
AC2046Nitrite100 Nitrite LR Tablets
AC2048Ozone100 Indigo Blue Tablets
AC2001pH100 Phenol Red Tablets
AC3001BPBromocresol Purple, pH, Low Range
AC3001TBThymol Blue, pH, High Range
AC2095-WAPhosphateSet of 2: 50 Phosphate No 1 LR Tablets, 50 Phosphate No 2 LR Tablet
AC2096PhosphateSet of 2: 50 Phosphate No 1 HR Tablets, 50 Phosphate No 2 HR Tablets
AC3019Potassium
AC2060SilicaSet of 2: 50 Silica No 1 Tablets, 50 Silica No 2 T
AC2061Silica100 Silica Phosphate Removal Tablets
AC2082Sulfate100 Sulfate Tablets
AC2016SulfideSet of 2: 100 Sulfide No 1 Tablets, 100 Sulfide No 2
Catalog number AC2030
Price (USD)
60.30
Online Exclusive
67.00
Save 6.70 (10%)
Each
Add to cart
Type:
Hydrazine
Includes:
Kit: Hydrazine Powder, Spoon
Price (USD)
60.30
Online Exclusive
67.00
Save 6.70 (10%)
Each
Add to cart
Ask our AI about this Product

Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100

Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionAQUAfast hydrazine reagent powder kit, 30 tests
For Use With (Application)Colorimetric water and wastewater testing equipment
IncludesKit: Hydrazine Powder, Spoon
Method4-(Dimethyl-amino)-Benzaldehyde
Quantity30 Tests
Range0.05 to 0.5 mg/L N2H4
Recommended AccessoriesAQUAfast 24mm round vials (Cat. No. AC2V24)
Test Reagent FormatsPowder Pack
For Use With (Equipment)Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
TypeHydrazine
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Share catalog number, name or link