CellEvent™ Caspase-3/7 Detection Reagents
Invitrogen™

CellEvent™ Caspase-3/7 Detection Reagents

CellEvent Caspase-3/7 detection reagents are fluorogenic no-wash substrates for apoptosis detection. These substrates detect activated caspase-3/7 in live cells that can be fixed and multiplexed with reporters, dyes, or antibodies.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityColorExcitation/EmissiondimFormat
C104231 x 100 μL VialGreen∼502/530
C104301 VialRed∼590/610
C104315 VialsRed∼590/610
C104321 VialGreen∼502/530
C104335 VialsGreen∼502/530
C104342 Vials (1 Green, 1 Red)Red, Green∼590/610, ∼502/530
C107231 x 25 μL VialGreen∼502/530
Catalog number C10423
Price (USD)
826.65
Online exclusive
870.00
Save 43.35
Each
Quantity:
1 x 100 μL Vial
Color:
Green
Excitation/Emission:
∼502/530
Price (USD)
826.65
Online exclusive
870.00
Save 43.35
Each

CellEvent Caspase-3/7 Green and CellEvent Caspase-3/7 Red detection reagents are novel fluorogenic substrates for activated caspase-3/7 in live cells that can be fixed and multiplexed with imaging dyes or antibodies. Activation of caspase-3 is a critical event during apoptosis, making these optimized reagents for analysis of apoptotic cells. CellEvent Caspase-3/7 Red Detection Reagent can also be used to detect apoptosis in GFP expressing cells.

Features of these indicators include:

  • Optimized caspase-3/7 substrate for apoptosis analysis
  • Easy to use—add and read; no cell wash or lysis required
  • Use for time course measurements; easily select the time point of interest up to and beyond 72 hours
  • Compatible with both live cell fluorescence-imaging and formaldehyde-based fixation methods
  • Multiplex enabled—combine with other fluorescent reagents to confirm apoptosis in the same cell or cell population

CellEvent™ Caspase-3/7 Green Detection Reagent: Absorption/emission maxima of ∼502/530 nm. Detectable with FITC/GFP traditional filter set.

CellEvent™ Caspase-3/7 Red Detection Reagent: Absorption/emission maxima of ∼590/610 nm. Detectable with traditional Texas red filter set.

CellEvent™ Caspase-3/7 Red Detection Reagent and CellEvent™ Caspase-3/7 Green is provided as Dry down powder. Just add 100 μL of PBS to dry down powder to generate 100X Stock solution. Dilute this 100X stock solution 1:100 in complete media and stain cells for 30 to 60 minutes and analyze with fluorescence microscope.

CellEvent™ Caspase-3/7 Green Detection Reagent is also provided as an already prepared DMSO solution. DMSO solution is 400X stock solution, which is ready to use. Just dilute in growth media 1:400 and stain the cells for 30 to 60 minutes.

To prevent the apoptotic cells from washing away, no wash is needed at the end of staining. Stained cells can be analyzed live or can be fixed with 4% Paraformaldehyde solution such as Image-IT™ Paraformaldehyde 4% in 0.1M Phosphate Buffer pH 7.4, Cat. No. I28800. These fixed cells can be further processed using fixed cell compatible dyes or ICC workflows.

Substrate specifics

CellEvent Caspase-3/7 Detection Reagents are a four-amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye. The DEVD peptide sequence is a cleavage site for caspase-3/7, and the conjugated dye is non-fluorescent until cleaved from the peptide and bound to DNA.

Principles of action

CellEvent Caspase-3/7 Green Detection Reagent is intrinsically non-fluorescent as the DEVD peptide inhibits the ability of the dye to bind to DNA. However, after activation of caspase-3/7 in apoptotic cells, the DEVD peptide is cleaved, enabling the dye to bind to DNA and produce a bright, fluorogenic response.

Simple three-step protocol

To use CellEvent Caspase 3/7 Detection Reagent, simply add it to cells, incubate 30 to 60 minutes, and visualize over time. Apoptotic cells with activated caspase-3/7 will have bright green or red nuclei, while cells without activated caspase 3/7 will have minimal fluorescent signal.

Assay versatility allows for live cell detection and fixation

This robust assay enables the examination of caspase-3/7 activation in live cells. Furthermore, since wash steps are not necessary for detection, fragile apoptotic cells typically lost during wash steps are preserved. Importantly, the fluorescent signal from CellEvent Caspase 3/7 Detection Reagent can survive fixation and permeabilization. This allows for the flexibility to perform end-point assays and probe for other proteins of interest using reporters, dyes, or immunocytochemistry.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Excitation/Emission∼502/530
For Use With (Equipment)Fluorescence Microscope, High Content Instrument
Formulation2 mM in DMSO
Incubation Time30 min
Label TypeOther Label(s) or Dye(s)
Product LineCellEvent
Product TypeCaspase 3/7 Green Detection Reagent
Quantity1 x 100 μL Vial
Reaction Time30 min
Shipping ConditionRoom Temperature
ConjugateCellEvent Caspase 3/7 Green
Detection MethodFluorescence
Unit SizeEach
Contents & Storage
100 μL, 2 mM solution in DMSO. Store (at ≤-20°C, desiccate and protect from light.)

Frequently asked questions (FAQs)

How many tests can I perform with each Invitrogen CellEvent Caspase-3/7 Green product?

The number of tests depends on the final assay volume. At 100 µL per well and 5 µM working solution, Invitrogen CellEvent Caspase-3/7 Green Detection Reagent, Cat. No. C10723, yields approximately 100 tests, while Cat. No. C10423 yields approximately 400 tests. Practical yield may be lower because of pipetting loss, dead volume, and concentration optimization.

How should Invitrogen CellEvent Caspase-3/7 Green Detection Reagent be stored, and what formats are supplied?

Prepare the working staining solution fresh each day, protect the reagents from light, and avoid repeated freeze-thaw cycles. The shipped products are stable for six months from receipt when stored as directed. Invitrogen CellEvent Caspase-3/7 Green Detection Reagent solutions, Cat. Nos. C10723 and C10423, are supplied as 400X, 2 mM solutions in DMSO.

Can cleaved Invitrogen CellEvent Caspase-3/7 Detection Reagents stain cytoplasmic RNA?

Cytoplasmic RNA staining may occur after Invitrogen CellEvent Caspase Detection Reagents are cleaved. The intended assay readout is nuclear fluorescence from released dye binding to nucleic acids in caspase-positive cells.

How long can Invitrogen CellEvent Caspase Detection Reagents remain in continuous culture?

For continuous culture experiments the Invitrogen CellEvent Caspase Detection Reagents can be maintained in complete cell culture medium at its working concentration.

Can I use a nuclear counterstain with Invitrogen CellEvent Caspase Detection Reagents?

Yes, but add the nuclear counterstain after the Invitrogen CellEvent Caspase reagent signal has developed. For kinetic or continuous-culture experiments, add a Hoechst nuclear stain at the endpoint to reduce potential effects on normal cell physiology. Nuclear counterstain dyes may compete for nucleic acid binding, so optimize the combination.

Citations & References (35)

Citations & References
Abstract
Combination Small Molecule MEK and PI3K Inhibition Enhances Uveal Melanoma Cell Death in a Mutant GNAQ- and GNA11-Dependent Manner.
Authors:Khalili JS, Yu X, Wang J, Hayes BC, Davies MA, Lizee G, Esmaeli B, Woodman SE,
Journal:Clin Cancer Res
PubMed ID:22733540
Activating Q209L/P mutations in GNAQ or GNA11 (GNAQ/11) are present in approximately 80% of uveal melanomas. Mutant GNAQ/11 are not currently therapeutically targetable. Inhibiting key down-stream effectors of GNAQ/11 represents a rational therapeutic approach for uveal melanomas that harbor these mutations. The mitogen-activated protein/extracellular signal-regulated kinase/mitogen-activated protein kinase (MEK/MAPK) and ... More
Perforin rapidly induces plasma membrane phospholipid flip-flop.
Authors:Metkar SS, Wang B, Catalan E, Anderluh G, Gilbert RJ, Pardo J, Froelich CJ,
Journal:PLoS One
PubMed ID:21931672
'The cytotoxic cell granule secretory pathway is essential for host defense. This pathway is fundamentally a form of intracellular protein delivery where granule proteases (granzymes) from cytotoxic lymphocytes are thought to diffuse through barrel stave pores generated in the plasma membrane of the target cell by the pore forming protein ... More
Membrane sialidase NEU3 is highly expressed in human melanoma cells promoting cell growth with minimal changes in the composition of gangliosides.
Authors:Miyata M, Kambe M, Tajima O, Moriya S, Sawaki H, Hotta H, Kondo Y, Narimatsu H, Miyagi T, Furukawa K, Furukawa K,
Journal:Cancer Sci
PubMed ID:21895867
'NEU3 is a membrane sialidase specific for gangliosides. Its increased expression and implication in some cancers have been reported. Here, we analyzed NEU3 expression in malignant melanoma cell lines and its roles in the cancer phenotypes. Quantitative RT-PCR revealed that high levels of the NEU3 gene were expressed at almost ... More
Pardaxin, an Antimicrobial Peptide, Triggers Caspase-Dependent and ROS-Mediated Apoptosis in HT-1080 Cells.
Authors:Huang TC, Lee JF, Chen JY,
Journal:Mar Drugs
PubMed ID:22073006
Pardaxin is an antimicrobial peptide (AMP) that was first isolated from secretions of the Red Sea Moses sole. The role of pardaxin in inducing apoptosis for preventing cancer has not yet been investigated. In the present study, we examined the antitumor activity of pardaxin against human fibrosarcoma HT-1080 cells; pardaxin ... More
Development of a highly sensitive in vitro endothelial cell toxicity assay for evaluating ricin toxin A chain-based vaccines or therapeutics.
Authors:Machesky NJ, Rusnak JM, Moore EH, Dorsey CB, Ward LA
Journal:Toxicon
PubMed ID:31207351
'The ricin toxin A chain (RTA) is responsible for ricin intoxication due to inhibition of protein synthesis. RTA is also known to cause endothelial toxicity [via a 3 amino acid sequence (x)D(y) motif that acts as a natural disintegrin] resulting in vascular leak syndrome (VLS) in humans. An in vitro ... More