Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), Texas Red
Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), Texas Red
Invitrogen™

Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), Texas Red

Lycopersicon esculentum (Tomato) Lectin (LEL, TL), Texas Red, is a bright-red LEL-fluorophore conjugate. The tomato lectin LEL is widely usedWeitere Informationen
Have Questions?
KatalognummerMenge
L324821 mg
Katalognummer L32482
Preis (EUR)
200,00
Each
-
Zum Warenkorb hinzufügen
Menge:
1 mg
Recurring order eligible. Learn more »
Preis (EUR)
200,00
Each
Zum Warenkorb hinzufügen
Ask our AI about this Product
Lycopersicon esculentum (Tomato) Lectin (LEL, TL), Texas Red, is a bright-red LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-Texas Red can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.

View complete list of fluorescent dye-conjugated lectins ›

• Very bright Texas Red conjugate detected with traditional FITC filter set, Ex/Em: 595/615 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
• Inhibiting/eluting sugar: chitin hydrolysate
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 71 KDa.
• Immunofluorescence working concentration: 5–20 μg/mL

LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Protein-SubtypLectins
Menge1 mg
QuelleTomato
KonjugatTexas Red
FormSolution
Unit SizeEach
Inhalt und Lagerung
When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Dokumente und Downloads

Zertifikate

Chargen-Nr.Certificate TypeDateCatalog Number(s)
3136477Certificate of Analysis10. Apr. 2025L32482
3118698Certificate of Analysis18. März 2025L32482
3091972Certificate of Analysis07. Nov. 2024L32482
2936372Certificate of Analysis21. Juli 2024L32482
2738293Certificate of Analysis27. Feb. 2024L32482
5 Ergebnisse angezeigt, oben nach einem bestimmten Zertifikat suchen

Sicherheitsdatenblätter

Geben Sie die Katalognummer, den Namen oder Link frei.

1x1 image pixel for data collection