Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 488
Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 488
Invitrogen™

Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 488

Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 488, is a bright-green LEL- fluorophore conjugate. The tomato lectin LEL is widelyRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
L32470Promo Image1 mg
Catalog number L32470
Price (USD)
211.65
Online Exclusive
218.00
Save 6.35 (3%)
Each
In stock
Add to cart
Quantity:
1 mg
Recurring order eligible. Learn more »
Price (USD)
211.65
Online Exclusive
218.00
Save 6.35 (3%)
Each
Add to cart
Ask our AI about this Product
Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 488, is a bright-green LEL- fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-DyLight 488 can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.

View complete list of fluorescent dye-conjugated lectins ›

• Very bright DyLight 488 conjugate detected with traditional FITC filter set, Ex/Em: 493/518 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
• Inhibiting/eluting sugar: chitin hydrolysate
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 71 KDa.
• Immunofluorescence working concentration: 5–20 μg/mL

LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeDyLight Fluors
Product LineDyLight™
Protein SubtypeLectins
Quantity1 mg
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
SourceTomato
ConjugateDyLight 488
FormSolution
Unit SizeEach
Contents & Storage
When stored at or below 2–8°C in the dark, the product is stable for 6 months after receipt.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3168071Certificate of AnalysisJun 01, 2025L32470
3136315Certificate of AnalysisApr 10, 2025L32470
3080119Certificate of AnalysisJan 16, 2025L32470
3023488Certificate of AnalysisSep 11, 2024L32470
2956387Certificate of AnalysisJun 10, 2024L32470
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection