1x1 image pixel for data collection
Qubit™ Flex Assay Tube Strips
Qubit™ Flex Assay Tube Strips
Invitrogen™

Qubit™ Flex Assay Tube Strips

Qubit Flex Assay Tube Strips are strips of 8 x 200-μL thin-walled polypropylene tubes with very low fluorescence ideal forRead more
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Catalog NumberQuantity
Q33252Promo Image125 tube strips
Catalog number Q33252
Price (USD)
286.00
125 tube strips
In stock
Add to cart
Quantity:
125 tube strips
Recurring order eligible. Learn more »
Price (USD)
286.00
125 tube strips
Add to cart
Ask our AI about this Product
Qubit Flex Assay Tube Strips are strips of 8 x 200-μL thin-walled polypropylene tubes with very low fluorescence ideal for optimal performance of the Qubit Flex Fluorometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)200 μL
Product LineQubit™
Quantity125 tube strips
Shipping ConditionRoom Temperature
For Use With (Equipment)Qubit Flex Fluorometer
TypeAssay Tube Strip
Unit Size125 tube strips

Enhance your Qubit Fluorometer with the latest firmware

New Qubit 4 and Qubit Flex firmware features include importing sample IDs, renaming files before exporting, a countdown timer, and on-board troubleshooting guidance. Get the most out of your Qubit assays with these powerful new tools.

Download the latest Qubit firmware ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can the Qubit Flex Assay Tube Strips (Cat. No. Q33252) be cut to run 1 sample or does the full tube strip need to be placed in the Qubit Flex Fluorometer (Cat. No. Q33327)?

Qubit Flex Assay Tube Strips (Cat. No. Q33252) can be cut to run 1 sample with the Qubit Flex Fluorometer (Cat. No. Q33327).

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

What would be a suitable alternative for the Qubit Flex Assay Tube Strips (Cat. No. Q33252)?

There is no official alternative to Qubit Flex Assay Tube Strips (Cat. No. Q33252). Some Qubit Flex users have had success with MicroAmp Optical 8-Tube Strips (Cat. No. A30588) and Sarstedt 0.2 mL PCR tube strips (PN 72.991.102), but we do not guarantee their performance. Sarstedt 0.2 mL PCR tube strips can be ordered from FisherSci (Cat. No. NC1880432) or directly from Sarstedt.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection