Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
R1223 | 70 mL |
Thermo Scientific GeneJET Lysis Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502, K0503) and may be purchased separately.
Related Products
Yes, the Lysis Solution from the GeneJET Plasmid Miniprep Kit is available as a stand-alone product (Cat. No. R1223).
Share catalog number, name or link