Contact Us
Soy Allergy Nuance: Differentiating between a clinical soy allergy and a pollen-related soy allergy
Anna and Oliver, both 8 years old, have a history of birch and soy allergy, both visit their healthcare provider after experiencing rhinitis during pollen season and dry skin.
Anna and OIiver's healthcare provider conducts a full clinical history and physical examination of them both and decides to test using soy components.
Two kids have a history of birch and soy allergy and visit their healthcare provider after experiencing dry skin and rhinitis during pollen season.
Patient History
Family History
  • None
Anna and Oliver's Personal History
  • Eczema during infancy, rhinitis.
Skin Prick Test
Test
Type
Anna's Results
Oliver's Results
Egg
Whole Allergen
0
0
Milk
Whole Allergen
0
0
Soy
Whole Allergen
+2
+2
Wheat
Whole Allergen
0
0
Fish
Whole Allergen
0
0
Birch
Whole Allergen
+2
+1
Timothy grass
Whole Allergen
0
0
Anna and Oliver's previous healthcare provider diagnosed them with birch and soy allergies. She recommended the use of oral antihistamines during birch pollen season and directed them to avoid soy and soy products.
Anna and Oliver’s ImmunoCAP™ Test Results
These results together with this patient's case history and symptoms, help confirm the diagnosis.
ImmunoCAP Test Results (kUA/I)
Test
Type
Anna’s Results
Oliver’s Results
Soy
Whole Allergen
0.8
24
Gly m 4
Allergen Component
15
2.5
Gly m 5
Allergen Component
<0.1
16.2
Gly m 6
Allergen Component
<0.1
6.7
Birch
Whole Allergen
58
20
Differential Diagnosis
Anna
Anna’s results indicate a clinically relevant cross-reaction between birch pollen and soy. Specific IgE antibodies to Gly m 4 are often associated with local reactions. However, systemic reactions may occur, particularly when consuming high amounts of low processed soy, e.g., soy milk. If clinical history is unclear, it may be advisable to proceed with a food challenge to confirm the diagnosis of soy allergy.1
REFINED DIAGNOSIS
  • Birch allergy is confirmed, with a likely pollen-related allergy to soy.
HEALTHCARE PROVIDER MANAGEMENT PLAN
Anna's healthcare provider advises her:
  • To avoid large amounts of soy, e.g., soy milk.
  • That she has no strict dietary restrictions on soy and should not be stressed about the risk of eating “hidden” soy.
  • To continue using antihistamine and nasal steroids during pollen season.
FOLLOW UP
  • Anna with her mother visit her healthcare provider one year later. Anna is doing well.
Oliver
Oliver’s results indicate his level of sensitization to Gly m 5 put him at an increased risk of severe reaction if soy is ingested, even small amounts.1
REFINED DIAGNOSIS
  • Soy and birch allergy was confirmed.
HEALTHCARE PROVIDER MANAGEMENT PLAN
Oliver's healthcare provider advises him:
  • To continue to avoid soy in all forms and always carry his emergency adrenaline autoinjector.
  • To get follow-up education and training on autoinjector use for Oliver and his family.
  • To continue using antihistamine and nasal steroids during pollen season.
FOLLOW UP
  • Oliver with his mother visit his healthcare provider one year later. He is fit and well. Oliver's healthcare provider checks his autoinjector technique and is satisfied.
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.
References
  1. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.