Case Study
Chronic Renal Failure: Rule Out Peritonitis, Peritoneal Dialysis
Michael R. Broyles
PharmD – Director of Medical Affairs, Thermo Fisher Scientific
Learn more about implementing procalcitonin testing
in your hospital.
Explore similar topics
Key Takeaways
Takeaway #1
Takeaway #1
The normal baseline PCT in renal patients is typically a bit higher—we use 0.5 μg/L at my facility.1
Takeaway #2
Takeaway #2
PCT is useful in patients on peritoneal dialysis, and trending is quite useful.
Takeaway #3
Takeaway #3
There was a steady decrease in PCT, by approximately 50% per day.
Share your Clinical Experience
Submit your own PCT Case Study and you may be selected to be featured on procalcitonin.com. 
Submit a case study here
Share your experience
Related Resources
References
  1. Meisner M. Procalcitonin-biochemistry and clinical diagnosis. Dresden (Germany): UNI-MED-Verlag; 2010.
Back to Top Arrow