Collibri DNA Whole Genome Sequencing Kits
Invitrogen Collibri whole-genome library prep kits improve library prep success for high-throughput Illumina™ next-generation sequencing (NGS) systems.
Accommodates a wide range of sample types and inputs:
  • Intact DNA
  • Degraded DNA, including FFPE or cfDNA
  • Suitable for genomes of all sizes
  • Consistent gene coverage from 1 to 500–1,000 ng
Variant detection is consistent among low abundance (1 ng) and high abundance (500–1,000 ng) samples to confidently analyze variable samples.
Sequence SARS-CoV-2 samples
High-quality genomic coverage from nasopharyngeal, oropharyngeal, and diverse samples increases accuracy in SARS-CoV-2 research such as the study of viral evolution and transmission patterns. The optimized workflow for SARS-CoV-2 reduces time and sequencing costs while providing flexibility to use available reagents.
Review the optimized protocol ›
Physically sheared DNA
Sequencing studies can now accept a greater diversity of samples and retain confidence in genomic interpretation. Variant detection rates by the Invitrogen Collibri PS DNA Library Prep Kits for Illumina Systems are similar between amplified (1–100 ng) libraries and PCR-free (500–1,000 ng) libraries.
Physically sheared DNA
Figure 1. Coriell NA12878 DNA was converted into a library and sequenced on an Illumina™ NovaSeq™ 6000 system at 372M reads per sample. SNP recall calculated using FreeBayes v1.2.0.
Enzymatically sheared DNA
Consistency of variant detection provides more consistent, predictable results from diverse samples. Variant detection rates by the Invitrogen Collibri ES DNA Library Prep Kits for Illumina Systems are similar between amplified (100 ng) libraries and PCR-free (100 ng) libraries.
Enzymatically sheared DNA
Figure 2. Coriell NA12878 DNA was converted into a library and sequenced on an Illumina NovaSeq 6000 system at 258M reads per sample. SNP recall calculated using FreeBayes v1.2.0.
Product comparison
 Kit view
Collibri PS DNA Library Prep Kits for Illumina Systems
Kit view
Collibri ES DNA Library Prep Kits for Illumina Systems
PCR-free input500 ng100 ng
Input1–1,000 ng1–500 ng
Lowest GC bias  
DNA shearing methodPhysical shearing (e.g., Covaris)Enzymatic shearing
Species supportedAllAll
PCR-free assay time (hr)1.51.5
Assay time (hr)2.752.5
PCR-free hands-on time (hr)0.50.5
Hands-on time (hr)11
Target insert size (bp)350-550350-550
All components included  
Visual feedback  
 View performance dataView performance data
Technical Note: DNA fragmentation strategies for next-generation sequencing library preparation
Choose between DNA fragmentation options based upon the need for even genomic coverage, high yields, and ease-of-automation.
Download
Ordering information
Catalog # Name Size Price (USD) Qty
A38602024 Collibri™ PCR-free ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set A, 1-24), 24 preps Each
1,288.00
A43602024 Collibri™ PCR-free ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set B, 25-48), 24 preps Each
1,288.00
A43603024 Collibri™ PCR-free ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set C, 49-72), 24 preps Each
1,306.00
A43604024 Collibri™ PCR-free ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set D, 73-96), 24 preps Each
1,306.00
A38545024 Collibri™ PCR-free ES DNA Library Prep Kit for Illumina Systems, with CD indexes, 24 preps Each
1,152.00
A38603096 Collibri™ PCR-free ES DNA Library Prep Kit for Illumina Systems, with CD indexes, 96 preps Each
4,245.00
A38606024 Collibri™ ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set A, 1-24), 24 preps Each
1,364.00
A43605024 Collibri™ ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set B, 25-48), 24 preps Each
1,350.00
A43606024 Collibri™ ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set C, 49-72), 24 preps Each
1,376.00
A43607024 Collibri™ ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set D, 73-96), 24 preps Each
1,376.00
A38607196 Collibri™ ES DNA Library Prep Kit for Illumina Systems, with UD indexes (Set A-D, 1-96), 96 preps Each
4,115.00
A38605024 Collibri™ ES DNA Library Prep Kit for Illumina Systems, with CD indexes, 24 preps Each
1,246.00
A38607096 Collibri™ ES DNA Library Prep Kit for Illumina Systems, with CD indexes, 96 preps Each
4,275.00
Catalog # Name Size Price (USD) Qty
A38609024 Collibri™ PCR-free PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set A, 1-24), 24 preps Each
1,232.00
A43608024 Collibri™ PCR-free PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set B, 25-48), 24 preps Each
1,232.00
A43609024 Collibri™ PCR-free PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set C, 49-72), 24 preps Each
1,232.00
A43610024 Collibri™ PCR-free PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set D, 73-96), 24 preps Each
1,232.00
A38608024 Collibri™ PCR-free PS DNA Library Prep Kit for Illumina Systems, with CD indexes, 24 preps Each
1,132.00
A38610096 Collibri™ PCR-free PS DNA Library Prep Kit for Illumina Systems, with CD indexes, 96 preps Each
4,130.00
A38613024 Collibri™ PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set A, 1-24), 24 preps Each
1,308.00
A43611024 Collibri™ PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set B, 25-48), 24 preps Each
1,300.00
A43612024 Collibri™ PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set C, 49-72), 24 preps Each
1,300.00
A43613024 Collibri™ PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set D, 73-96), 24 preps Each
1,300.00
A38614196 Collibri™ PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set A-D, 1-96), 96 preps Each
4,285.00
A38612024 Collibri™ PS DNA Library Prep Kit for Illumina Systems, with CD indexes, 24 preps Each
1,202.00
A38614096 Collibri™ PS DNA Library Prep Kit for Illumina Systems, with CD indexes, 96 preps Each
4,190.00
For Research Use Only. Not for use in diagnostic procedures.