Whether you are leveraging stem cells for basic research, drug discovery, or future therapeutic applications, we offer an extensive range of Gibco media, supplements, and reagents for all cell types to meet your stem cell culture needs. Stem cell culture involves growing stem cells under controlled laboratory conditions using specialized media and supplements formulated to maintain their viability and properties. These culture systems (including feeder-free, serum-free, and xeno-free media) are designed to support stem cell self-renewal or directed differentiation as needed, while helping ensure consistency and reproducibility in experimental results.
Request information
Pluripotent stem cell (PSC) research requires careful attention to culture conditions. From feeder-based media to feeder-free media to media for stem cell therapy research, Gibco products enable you to culture with confidence.
Gibco media and supplements are highly cited reagents for use in neural stem cell (NSC) culture. Specifically formulated for serum-free growth and expansion of human NSCs, choose between B-27, N-2, StemPro Neural Supplement, Neurobasal Medium, and more.
We offer a wide range of specialty products optimized for mesenchymal stem cell (MSC) populations. These media are designed to help minimize adaptation time, enhance cell performance, and help meet regulatory requirements.
Hematopoietic stem cells expressing CD34 (CD34+ cells) are defined by their ability to replenish all blood cell types and to self-renew. Gibco StemPro-34 SFM is a serum-free medium specifically formulated to support the culture of these stem cells and offers excellent expansion and differentiation of HSCs isolated from bone marrow, peripheral blood, or neonatal cord blood over traditional culture systems.
Stem cell therapy culture system
As you move from basic cell therapy research to the clinic, high-quality products and proper documentation are essential to getting it right the first time. Cell Therapy Systems (CTS) products help minimize the risk of contamination and variability in your research and offer documentation to support regulatory review, making them an excellent choice as you transition from the bench to the clinic.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments