Search Thermo Fisher Scientific
The alamarBlue HS and alamarBlue Cell Viability Reagents are ready-to-use resazurin-based reagents that function as cell health indicators by using the reducing power of living cells to quantitatively measure viability. Upon entering living cells, resazurin is reduced to resorufin, a compound that is red in color and highly fluorescent.
![]() |
![]() |
![]() |
![]() |
![]() |
Note regarding subsequent culture
After viability determination, the diluted alamarBlue HS or alamarBlue reagent can be replaced with complete media and returned to the incubator. The cells will continue to proliferate normally.
alamarBlue HS or alamarBlue cell viability reagents | |
---|---|
Excitation/Emission | 560/590 nm |
Absorbance | 570 nm (use 600 nm reference wavelength) |
Storage conditions | 4°C |
Both alamarBlue Cell Viability Reagents are supplied as 10X solutions. Add alamarBlue reagent directly to cells in culture medium. See below for example volumes:
Format | Volume of cells + medium | Volume of alamarBlue HS or alamarBlue reagent |
---|---|---|
Cuvette | 900 μL | 100 μL |
96-well plate | 90 μL | 10 μL |
384-well plate | 36 μL | 4 μL |