NovaFluor Red 700 excitation shown as dotted line and emission shown as solid red histogram
5640695/40637701(in buffer) 5
flow cytometry
Invitrogen NovaFluor Red 700 dye is ~6 times brighter than APC-Cyanine 5.5 and Alexa Fluor 680 dye. In complex multicolor flow cytometry panels, NovaFluor Red 700 dye can be used to fill the spectral gap between Alexa Fluor 647 and Alexa Fluor 700 dyes.
The macromolecule-based NovaFluor Red 700 dye produces highly stable fluorescence, and stained samples retain their fluorescence intensity and spectral signature when stored at 4°C. Use NovaFluor dyes with CellBlox Plus Blocking Buffer to reduce background and to block non-specific binding of NovaFluor labels, PE and APC tandems observed with macrophages and monocytes.
We offer NovaFluor dyes conjugated to primary antibodies for use in flow cytometry, as well as NovaFluor Antibody Conjugation Kits, NovaFluor CD4 Label Characterization Kits, and custom conjugation services.
Search NovaFluor Red 700 primary antibodies
  • Narrow emission spectrum and minimal cross-laser excitation for better resolution
  • Bright fluorescence allows it to be paired with medium-to-low abundance antigens
  • Can be used to fill the spectra gap between Alexa Fluor 647 and Alexa Fluor 700 dyes
  • Can be used in a flow cytometry panel with NovaFluor Red 660 (or APC-Alexa Fluor 647) and Alexa Fluor 700 dyes
  • Produces highly stable fluorescence and is compatible with all buffers tested
Spectral signature of NovaFluor Red 700 dye. Normal human peripheral blood cells stained with CD4 Monoclonal Antibody (SK3 (SK-3)), NovaFluor Red 700 were used for analysis. Data was acquired on a 5-laser Cytek Aurora system.
Side scatter plot with unstained and CD3 NovaFluor Red 700 dye-stained normal human PBMCs
Staining of human peripheral blood cells with NovaFluor Red 700 conjugated antibody. Normal human peripheral blood cells (PBMCs) were unstained (left) or stained with CD3 Monoclonal Antibody, NovaFluor Red 700 (right). Data was acquired in the R3 channel on a 5-laser Cytek Aurora system, and singlet cells were used for analysis.
NovaFluor dyes
NovaFluor dyes are built using Phiton technology and are compatible with both spectral flow cytometry and traditional flow cytometry. NovaFluor dyes exhibit narrow emission spectra and minimal cross-laser excitation, which helps reduce spectral spillover for better marker resolution. In addition, their unique spectral signatures can provide the opportunity to detect additional markers in flow cytometry panels by opening up previously unusable channels.
Additional resources
Flow cytometry protocols handbook
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
Request the handbook
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.