Scientist holding a flask with pink liquid
Achieve maximum performance in chromatography with Thermo Scientific chromatography solvents. Our solvents meet the challenges of chromatography for a wide range of laboratory research applications, from LC to GC- and UHPLC-MS and more. We supply solvents, blends, and reagents in grades, sizes, and packaging that best fit your needs.
Contact us   Download brochure
We offer a range of chromatography solutions for research and development applications in pharmaceuticals, biotechnology, environmental analysis, food and beverage testing, forensic toxicology, proteomics, metabolomics, and clinical research.
Manufacturing capabilities
Our experience in manufacturing, processing, and testing high purity solvents enables us to fully customize solvents to meet your specifications. Our dedicated solvent mixing facilities produce high purity blends, and our long-standing expertise and experience in distillation, processing, testing, and packaging high purity solvents make Thermo Scientific your brand of choice for chromatography lab applications.
Explore more chromatography solutions
Our chromatography solvents are made specifically for HPLC instrumentation. They are prepared by a distillation process to remove volatile impurities that can contaminate the analytical columns and reduce their lifetimes, followed by filtration to achieve maximum purity. These solvents have low UV absorption and low peak interference and are free of particles up to 0.02 micron. Thus, usage of our high purity HPLC solvents extends column lifetime. Our HPLC grade solvents are also designed to significantly reduce detector noise and baseline drifts, thereby enhancing data quality and helping ensure reproducibility.
Quality control laboratories that employ UHPLC-MS systems will find that our ultra-high purity solvents help improve mass spectrometer sensitivity to chromatographic interferences. Developed for mobile phases targeting trace analysis by UHPLC-MS, our ultra-pure solvents provide a very low mass noise level in both positive and negative mode ionization, minimal metal ion content, and very low UHPLC-UV response using photo diode array detection. Acetonitrile, methanol, and water are among the high-purity solvents that are specifically qualified for UHPLC-MS.
Catalog # Name Size Price (USD) Qty
396650250 1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous Each
176.65
Special offer
Online exclusive
208.00
Save 31.35 (15%)
396640250 1-Butanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous Each
127.65
Special offer
Online exclusive
150.00
Save 22.35 (15%)
396690250 1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous Each
331.65
Online Exclusive
368.00
Save 36.35 (10%)
396681000 1-Heptanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous Each
867.65
Special offer
Online exclusive
1,020.00
Save 152.35 (15%)
396670250 1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous Each
331.65
Special offer
Online exclusive
390.00
Save 58.35 (15%)
446882500 Acetic acid, sodium salt, 99%, for HPLC, anhydrous Each
160.65
Special offer
Online exclusive
189.00
Save 28.35 (15%)
396710250 1-Decanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous Each
349.65
Special offer
Online exclusive
411.00
Save 61.35 (15%)
396720250 1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous Each -
447362500 Potassium phosphate, dibasic, 99%, for HPLC, anhydrous Each
226.65
Special offer
Online exclusive
266.00
Save 39.35 (15%)
Catalog # Name Size Price (USD) Qty
28905 Formic Acid, LC-MS Grade, 10 x 1 mL Each
140.00
85183 Pierce™ Trifluoroacetic Acid (TFA), LC-MS Grade Each
210.00
047146.K7 Water, LC-MS Grade Each
111.65
Special offer
Online exclusive
131.00
Save 19.35 (15%)
25104 Pierce™ Pyridine, LC-MS Grade Each
118.00
T111101000 ChromaCare™ LC-MS Instrument Flush Solution Each
80.65
Special offer
Online exclusive
95.10
Save 14.45 (15%)
MB1241 ChromaCare™ LC-MS Biologics Flush Solution Each
124.00
53104 Pierce™ Heptafluorobutyric Acid (HFBA), HPLC Grade 10 x 1 mL
194.00
Catalog # Name Size Price (USD) Qty
W81 Water, UHPLC-MS Each
45.65
Online Exclusive
50.00
Save 4.35 (9%)
A9561 Acetonitrile, UHPLC-MS Each
64.65
Special offer
Online exclusive
76.10
Save 11.45 (15%)
A4581 Methanol, UHPLC-MS Each
68.65
Special offer
Online exclusive
80.00
Save 11.35 (14%)
UHPLCMSKIT UHPLC-MS Reagents Installation Kit Each
311.65
Online Exclusive
346.00
Save 34.35 (10%)