Organism TypeHuman293-F cells are GIBCO brand cells. These are clonal isolates derived from transformed human embryonal kidney (HEK) cells. The 293-F strain is a fast-growing variant of the 293 cell line. They have been adapted to suspension culture in serum-free media, 293 SFM II.
TissuesKidney
PhenotypeEither
Primaryno
ApplicationCell Culture / Growth Conditions,Transfection: Gene Expression,Transient Transfection
Products
Catalog # Name Size Price (USD) Qty
10566016 DMEM, high glucose, GlutaMAX™ Supplement, 500 mL Each
32.16
Special offer
Online exclusive
Ends: 01-May-2025
45.95
Save 13.79 (30%)
11668019 Lipofectamine™ 2000 Transfection Reagent, 1.5 mL Each
804.65
Online exclusive
882.00
Save 77.35 (9%)
11668027 Lipofectamine™ 2000 Transfection Reagent, 0.75 mL Each
519.65
Online exclusive
562.00
Save 42.35 (8%)
11686029 293 SFM II Each
163.00
11913019 CD 293 Medium (1X) Each
151.00
11965084 DMEM, high glucose, 1000 mL Each
52.65
Online exclusive
57.75
Save 5.10 (9%)
11965092 DMEM, high glucose, 500 mL Each
32.65
Online exclusive
35.34
Save 2.69 (8%)
11965118 DMEM, high glucose, 10 x 500 mL Each
276.65
Online exclusive
304.00
Save 27.35 (9%)
11965126 DMEM, high glucose, 6 x 1000 mL Each
266.65
Online exclusive
293.00
Save 26.35 (9%)
15140122 Penicillin-Streptomycin (10,000 U/mL), 100 mL Each
31.45
16447100 FreeStyle™ MAX Reagent, 1 mL Each
840.65
Online exclusive
886.00
Save 45.35 (5%)
18324012 Lipofectamine™ Transfection Reagent, 1 mL Each
650.00
25030081 L-Glutamine (200 mM), 100 mL Each
35.65
Online exclusive
38.90
Save 3.25 (8%)
51985034 Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement, 500 mL 500 mL
51.63
Special offer
Online exclusive
Ends: 01-May-2025
73.75
Save 22.12 (30%)
K278001 Calcium Phosphate Transfection Kit Each
700.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.